Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47668632-47668993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
UniGene
Hs.147377
Gene Chromosome Location
Chr.11: 47659168-47715397 on Build GRCh38
Transcripts
NM_024783.3 XM_005253138.4 XM_005253139.4 XM_005253140.4 XM_006718327.3 XM_006718328.3 XM_011520372.2 XM_011520373.2 XM_011520374.2 XM_011520375.2 XM_011520376.2 XM_011520377.2 XM_011520378.2 XM_011520379.2 XM_011520381.2 XM_017018331.1 XM_017018332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4660325 c.2176G>A p.D726N Chr.11: 47668879-47668879 on Build GRCh38
3447857 c.2179G>A p.G727S Chr.11: 47668876-47668876 on Build GRCh38
7115451 c.2148-7G>A p.? Chr.11: 47668914-47668914 on Build GRCh38
5140987 c.2192A>C p.H731P Chr.11: 47668863-47668863 on Build GRCh38
158956 c.2209G>A p.D737N Chr.11: 47668846-47668846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778438728 Chr.11: 47668929-47668929 on Build GRCh38
rs757640792 Chr.11: 47668927-47668927 on Build GRCh38
rs998731436 Chr.11: 47668768-47668768 on Build GRCh38
rs779667726 Chr.11: 47668962-47668962 on Build GRCh38
rs907791180 Chr.11: 47668875-47668875 on Build GRCh38
rs770539820 Chr.11: 47668844-47668844 on Build GRCh38
rs768600821 Chr.11: 47668874-47668874 on Build GRCh38
rs184416393 Chr.11: 47668656-47668656 on Build GRCh38
rs776327427 Chr.11: 47668879-47668879 on Build GRCh38
rs529421193 Chr.11: 47668714-47668714 on Build GRCh38
rs758082115 Chr.11: 47668826-47668827 on Build GRCh38
rs781712490 Chr.11: 47668793-47668793 on Build GRCh38
rs561969049 Chr.11: 47668815-47668815 on Build GRCh38
rs140006264 Chr.11: 47668851-47668851 on Build GRCh38
rs751252201 Chr.11: 47668925-47668928 on Build GRCh38
rs761643461 Chr.11: 47668887-47668887 on Build GRCh38
rs143896953 Chr.11: 47668898-47668898 on Build GRCh38
rs778231562 Chr.11: 47668920-47668920 on Build GRCh38
rs375374926 Chr.11: 47668923-47668923 on Build GRCh38
rs767525366 Chr.11: 47668904-47668904 on Build GRCh38
rs1037070368 Chr.11: 47668950-47668950 on Build GRCh38
rs751801689 Chr.11: 47668892-47668892 on Build GRCh38
rs756656574 Chr.11: 47668915-47668915 on Build GRCh38
rs867537019 Chr.11: 47668899-47668899 on Build GRCh38
rs778073994 Chr.11: 47668824-47668824 on Build GRCh38
rs761860851 Chr.11: 47668852-47668852 on Build GRCh38
rs745441049 Chr.11: 47668849-47668849 on Build GRCh38
rs753134429 Chr.11: 47668810-47668810 on Build GRCh38
rs766567065 Chr.11: 47668890-47668890 on Build GRCh38
rs921784935 Chr.11: 47668778-47668778 on Build GRCh38
rs760638276 Chr.11: 47668869-47668869 on Build GRCh38
rs541766540 Chr.11: 47668718-47668718 on Build GRCh38
rs768202253 Chr.11: 47668881-47668882 on Build GRCh38
rs974535272 Chr.11: 47668715-47668715 on Build GRCh38
rs150818119 Chr.11: 47668841-47668841 on Build GRCh38
rs753236343 Chr.11: 47668910-47668910 on Build GRCh38
rs771732922 Chr.11: 47668958-47668958 on Build GRCh38
rs752383175 Chr.11: 47668796-47668796 on Build GRCh38
rs368341488 Chr.11: 47668956-47668956 on Build GRCh38
rs749633475 Chr.11: 47668835-47668835 on Build GRCh38

View Full Product Details