Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85957650-85958189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

SNP Detail

SNP ID Chromosome Location
rs906488449 Chr.11: 85958069-85958069 on Build GRCh38
rs1045457800 Chr.11: 85957769-85957769 on Build GRCh38
rs562451968 Chr.11: 85957990-85957990 on Build GRCh38
rs1041851209 Chr.11: 85958083-85958083 on Build GRCh38
rs985331209 Chr.11: 85957676-85957678 on Build GRCh38
rs930104594 Chr.11: 85958023-85958023 on Build GRCh38
rs531461791 Chr.11: 85957825-85957825 on Build GRCh38
rs1009957149 Chr.11: 85957816-85957816 on Build GRCh38
rs895959094 Chr.11: 85957822-85957822 on Build GRCh38
rs937421936 Chr.11: 85957810-85957810 on Build GRCh38
rs1025989117 Chr.11: 85957956-85957963 on Build GRCh38
rs867882955 Chr.11: 85958048-85958048 on Build GRCh38
rs1018517113 Chr.11: 85957706-85957706 on Build GRCh38
rs138705131 Chr.11: 85958154-85958154 on Build GRCh38
rs534789688 Chr.11: 85957857-85957857 on Build GRCh38
rs540314632 Chr.11: 85957780-85957780 on Build GRCh38
rs10927 Chr.11: 85957801-85957801 on Build GRCh38
rs1056951670 Chr.11: 85958143-85958143 on Build GRCh38
rs965355323 Chr.11: 85957772-85957772 on Build GRCh38
rs77556759 Chr.11: 85957675-85957675 on Build GRCh38
rs560065389 Chr.11: 85958160-85958160 on Build GRCh38
rs544153970 Chr.11: 85957985-85957985 on Build GRCh38
rs937456073 Chr.11: 85958033-85958033 on Build GRCh38
rs551179450 Chr.11: 85957912-85957912 on Build GRCh38
rs999195463 Chr.11: 85957858-85957858 on Build GRCh38
rs985174108 Chr.11: 85957908-85957908 on Build GRCh38
rs888466032 Chr.11: 85957718-85957718 on Build GRCh38
rs1042379402 Chr.11: 85957826-85957826 on Build GRCh38
rs976585583 Chr.11: 85957932-85957932 on Build GRCh38
rs556314944 Chr.11: 85958026-85958027 on Build GRCh38
rs1018353366 Chr.11: 85957909-85957909 on Build GRCh38
rs909995918 Chr.11: 85958046-85958046 on Build GRCh38
rs610916 Chr.11: 85957687-85957687 on Build GRCh38
rs918717748 Chr.11: 85957674-85957674 on Build GRCh38
rs1018132503 Chr.11: 85957921-85957921 on Build GRCh38
rs1045112123 Chr.11: 85958060-85958060 on Build GRCh38
rs950276362 Chr.11: 85958081-85958081 on Build GRCh38
rs918771868 Chr.11: 85958019-85958019 on Build GRCh38
rs891597494 Chr.11: 85957809-85957809 on Build GRCh38
rs185162245 Chr.11: 85958142-85958142 on Build GRCh38
rs757491737 Chr.11: 85957962-85957973 on Build GRCh38
rs533066609 Chr.11: 85958145-85958145 on Build GRCh38
rs951259611 Chr.11: 85958016-85958016 on Build GRCh38
rs948102131 Chr.11: 85957726-85957726 on Build GRCh38
rs991964049 Chr.11: 85958045-85958045 on Build GRCh38
rs984269921 Chr.11: 85958026-85958026 on Build GRCh38
rs1032009003 Chr.11: 85957861-85957861 on Build GRCh38
rs775350662 Chr.11: 85957798-85957798 on Build GRCh38
rs968422185 Chr.11: 85957872-85957872 on Build GRCh38
rs769559663 Chr.11: 85957686-85957686 on Build GRCh38
rs1056001160 Chr.11: 85957814-85957814 on Build GRCh38
rs773609423 Chr.11: 85958137-85958137 on Build GRCh38
rs765254297 Chr.11: 85958022-85958022 on Build GRCh38
rs748778764 Chr.11: 85957799-85957799 on Build GRCh38
rs965607764 Chr.11: 85957923-85957923 on Build GRCh38
rs3088276 Chr.11: 85957689-85957689 on Build GRCh38
rs903402806 Chr.11: 85957856-85957856 on Build GRCh38
rs1053478155 Chr.11: 85958164-85958164 on Build GRCh38
rs893594334 Chr.11: 85957868-85957868 on Build GRCh38
rs929993601 Chr.11: 85957791-85957791 on Build GRCh38
rs762345946 Chr.11: 85958000-85958000 on Build GRCh38
rs977058427 Chr.11: 85957775-85957775 on Build GRCh38
rs767919632 Chr.11: 85958131-85958131 on Build GRCh38

View Full Product Details