Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111685746-111686326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
-
UniGene
Hs.331141
Gene Chromosome Location
Chr.12: 111686053-111757107 on Build GRCh38
Transcripts
NM_001136538.1 NM_025247.5 NM_006768.4 XM_005253944.4 XM_011538789.2 XM_017019992.1 XM_017019993.1 XM_017019994.1

Gene Symbol
BRAP
Entrez Gene ID
8315
Gene Name
BRCA1 associated protein
Gene Aliases
BRAP2, IMP, RNF52
UniGene
Hs.530940
Gene Chromosome Location
Chr.12: 111642146-111686023 on Build GRCh38
Transcripts
NM_001136538.1 NM_025247.5 NM_006768.4 XM_005253944.4 XM_011538789.2 XM_017019992.1 XM_017019993.1 XM_017019994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298919 c.26G>C p.R9P Chr.12: 111685767-111685767 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035662110 Chr.12: 111685979-111685979 on Build GRCh38
rs562270000 Chr.12: 111685805-111685805 on Build GRCh38
rs1006801644 Chr.12: 111686104-111686104 on Build GRCh38
rs1021243989 Chr.12: 111685907-111685907 on Build GRCh38
rs1003805405 Chr.12: 111686033-111686033 on Build GRCh38
rs543173295 Chr.12: 111685944-111685944 on Build GRCh38
rs1023454821 Chr.12: 111685952-111685952 on Build GRCh38
rs893868217 Chr.12: 111686003-111686003 on Build GRCh38
rs927963067 Chr.12: 111686146-111686146 on Build GRCh38
rs888894561 Chr.12: 111685876-111685876 on Build GRCh38
rs975841580 Chr.12: 111686188-111686188 on Build GRCh38
rs913846928 Chr.12: 111685987-111685987 on Build GRCh38
rs972360199 Chr.12: 111685957-111685957 on Build GRCh38
rs993887745 Chr.12: 111686249-111686249 on Build GRCh38
rs112071868 Chr.12: 111686267-111686267 on Build GRCh38
rs371857332 Chr.12: 111685807-111685807 on Build GRCh38
rs897285742 Chr.12: 111686213-111686213 on Build GRCh38
rs923046154 Chr.12: 111686204-111686204 on Build GRCh38
rs979328730 Chr.12: 111685999-111685999 on Build GRCh38
rs910387154 Chr.12: 111686163-111686163 on Build GRCh38
rs554711092 Chr.12: 111685897-111685897 on Build GRCh38
rs114642524 Chr.12: 111686141-111686141 on Build GRCh38
rs983899745 Chr.12: 111685925-111685925 on Build GRCh38
rs1049542083 Chr.12: 111685874-111685874 on Build GRCh38
rs926393812 Chr.12: 111685815-111685815 on Build GRCh38
rs987958356 Chr.12: 111685852-111685852 on Build GRCh38
rs1016713954 Chr.12: 111686079-111686079 on Build GRCh38
rs928948495 Chr.12: 111686208-111686208 on Build GRCh38
rs988483127 Chr.12: 111685984-111685984 on Build GRCh38
rs1043852364 Chr.12: 111685869-111685869 on Build GRCh38
rs1005753520 Chr.12: 111686184-111686184 on Build GRCh38
rs1048435608 Chr.12: 111686090-111686090 on Build GRCh38
rs1023591659 Chr.12: 111686160-111686160 on Build GRCh38
rs376859494 Chr.12: 111685816-111685816 on Build GRCh38
rs758091237 Chr.12: 111685924-111685925 on Build GRCh38
rs1036971205 Chr.12: 111685861-111685861 on Build GRCh38
rs796549306 Chr.12: 111686114-111686114 on Build GRCh38
rs576168995 Chr.12: 111685954-111685954 on Build GRCh38
rs190918934 Chr.12: 111685981-111685981 on Build GRCh38
rs922556423 Chr.12: 111685961-111685961 on Build GRCh38
rs776958367 Chr.12: 111685767-111685767 on Build GRCh38
rs970020004 Chr.12: 111685820-111685820 on Build GRCh38
rs759619713 Chr.12: 111686067-111686067 on Build GRCh38
rs1003753042 Chr.12: 111685914-111685916 on Build GRCh38
rs943105347 Chr.12: 111686021-111686021 on Build GRCh38
rs113364574 Chr.12: 111686180-111686180 on Build GRCh38
rs895336536 Chr.12: 111686036-111686036 on Build GRCh38
rs770724086 Chr.12: 111686005-111686005 on Build GRCh38
rs1042406858 Chr.12: 111686006-111686006 on Build GRCh38
rs964432636 Chr.12: 111686165-111686165 on Build GRCh38
rs777609801 Chr.12: 111685978-111685978 on Build GRCh38
rs774585593 Chr.12: 111686027-111686027 on Build GRCh38
rs985306504 Chr.12: 111685974-111685974 on Build GRCh38
rs1022938149 Chr.12: 111685779-111685779 on Build GRCh38
rs913728513 Chr.12: 111685854-111685854 on Build GRCh38
rs955061922 Chr.12: 111686270-111686270 on Build GRCh38
rs956054030 Chr.12: 111686127-111686127 on Build GRCh38
rs751817733 Chr.12: 111685813-111685813 on Build GRCh38
rs772485386 Chr.12: 111686126-111686126 on Build GRCh38
rs541355946 Chr.12: 111686016-111686016 on Build GRCh38
rs1009963074 Chr.12: 111685906-111685906 on Build GRCh38
rs1040021508 Chr.12: 111686034-111686034 on Build GRCh38
rs543662014 Chr.12: 111685967-111685967 on Build GRCh38
rs559780923 Chr.12: 111686258-111686258 on Build GRCh38
rs1026458208 Chr.12: 111685932-111685932 on Build GRCh38
rs1055408388 Chr.12: 111686169-111686169 on Build GRCh38
rs942305314 Chr.12: 111686269-111686269 on Build GRCh38
rs527688294 Chr.12: 111686064-111686064 on Build GRCh38
rs559383968 Chr.12: 111686043-111686043 on Build GRCh38
rs911063950 Chr.12: 111685977-111685977 on Build GRCh38
rs780987768 Chr.12: 111685817-111685817 on Build GRCh38
rs371357979 Chr.12: 111685996-111685996 on Build GRCh38
rs752536476 Chr.12: 111685818-111685818 on Build GRCh38
rs939305849 Chr.12: 111685846-111685846 on Build GRCh38
rs1038941072 Chr.12: 111686195-111686195 on Build GRCh38
rs561062298 Chr.12: 111686111-111686111 on Build GRCh38
rs756368602 Chr.12: 111686156-111686156 on Build GRCh38
rs536254027 Chr.12: 111685887-111685887 on Build GRCh38
rs959105906 Chr.12: 111685817-111685817 on Build GRCh38
rs968026768 Chr.12: 111686157-111686157 on Build GRCh38
rs1010260027 Chr.12: 111686135-111686135 on Build GRCh38
rs751943488 Chr.12: 111685823-111685823 on Build GRCh38
rs750525758 Chr.12: 111685801-111685801 on Build GRCh38
rs1033850136 Chr.12: 111685922-111685922 on Build GRCh38
rs761910725 Chr.12: 111686002-111686002 on Build GRCh38
rs943471131 Chr.12: 111685889-111685889 on Build GRCh38
rs946457620 Chr.12: 111685855-111685855 on Build GRCh38
rs765160587 Chr.12: 111685787-111685787 on Build GRCh38
rs531666774 Chr.12: 111686125-111686125 on Build GRCh38
rs906773867 Chr.12: 111685913-111685914 on Build GRCh38
rs529374424 Chr.12: 111685896-111685896 on Build GRCh38
rs961410928 Chr.12: 111686097-111686097 on Build GRCh38
rs777630125 Chr.12: 111685826-111685826 on Build GRCh38
rs922507214 Chr.12: 111685866-111685866 on Build GRCh38
rs902059831 Chr.12: 111685898-111685898 on Build GRCh38
rs145117804 Chr.12: 111685769-111685769 on Build GRCh38
rs775839134 Chr.12: 111686131-111686131 on Build GRCh38
rs955009104 Chr.12: 111686107-111686107 on Build GRCh38
rs929544089 Chr.12: 111686066-111686066 on Build GRCh38
rs748828361 Chr.12: 111685833-111685833 on Build GRCh38
rs1047431511 Chr.12: 111686247-111686247 on Build GRCh38
rs889240272 Chr.12: 111685875-111685875 on Build GRCh38
rs756923770 Chr.12: 111685821-111685821 on Build GRCh38
rs887957990 Chr.12: 111685940-111685940 on Build GRCh38
rs542573463 Chr.12: 111686100-111686100 on Build GRCh38
rs993751469 Chr.12: 111685915-111685915 on Build GRCh38
rs1044605020 Chr.12: 111686108-111686108 on Build GRCh38
rs375417855 Chr.12: 111685928-111685928 on Build GRCh38
rs1042461421 Chr.12: 111686144-111686144 on Build GRCh38

View Full Product Details