Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94173368-94173964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP29
Entrez Gene ID
9411
Gene Name
Rho GTPase activating protein 29
Gene Aliases
PARG1
UniGene
Hs.483238
Gene Chromosome Location
Chr.1: 94168907-94275068 on Build GRCh38
Transcripts
NM_004815.3 XM_006711048.3 XM_011542438.2 XM_011542439.2 XM_011542440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5010577 c.3718C>A p.Q1240K Chr.1: 94173937-94173937 on Build GRCh38
6064495 c.3697C>G p.P1233A Chr.1: 94173958-94173958 on Build GRCh38
1296812 c.3760G>A p.E1254K Chr.1: 94173895-94173895 on Build GRCh38
913479 c.3737G>A p.R1246Q Chr.1: 94173918-94173918 on Build GRCh38
913478 c.3759C>T p.L1253L Chr.1: 94173896-94173896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915535985 Chr.1: 94173768-94173768 on Build GRCh38
rs533201177 Chr.1: 94173797-94173797 on Build GRCh38
rs144300371 Chr.1: 94173445-94173445 on Build GRCh38
rs946280093 Chr.1: 94173458-94173458 on Build GRCh38
rs539532569 Chr.1: 94173526-94173526 on Build GRCh38
rs750684448 Chr.1: 94173850-94173850 on Build GRCh38
rs1007243257 Chr.1: 94173622-94173622 on Build GRCh38
rs964533751 Chr.1: 94173434-94173434 on Build GRCh38
rs199687794 Chr.1: 94173902-94173902 on Build GRCh38
rs1029950848 Chr.1: 94173400-94173400 on Build GRCh38
rs757192020 Chr.1: 94173490-94173490 on Build GRCh38
rs35865570 Chr.1: 94173666-94173667 on Build GRCh38
rs759401331 Chr.1: 94173911-94173911 on Build GRCh38
rs375753775 Chr.1: 94173896-94173896 on Build GRCh38
rs1021796299 Chr.1: 94173661-94173661 on Build GRCh38
rs560724872 Chr.1: 94173734-94173736 on Build GRCh38
rs768040389 Chr.1: 94173706-94173706 on Build GRCh38
rs956925801 Chr.1: 94173715-94173715 on Build GRCh38
rs777099561 Chr.1: 94173844-94173844 on Build GRCh38
rs914770069 Chr.1: 94173446-94173446 on Build GRCh38
rs1330858 Chr.1: 94173781-94173781 on Build GRCh38
rs369260500 Chr.1: 94173679-94173679 on Build GRCh38
rs564905932 Chr.1: 94173484-94173484 on Build GRCh38
rs764560723 Chr.1: 94173832-94173833 on Build GRCh38
rs1005110860 Chr.1: 94173530-94173530 on Build GRCh38
rs544158335 Chr.1: 94173722-94173722 on Build GRCh38
rs922612241 Chr.1: 94173849-94173849 on Build GRCh38
rs1999272 Chr.1: 94173891-94173891 on Build GRCh38
rs541671376 Chr.1: 94173615-94173615 on Build GRCh38
rs759862562 Chr.1: 94173894-94173894 on Build GRCh38
rs957796914 Chr.1: 94173630-94173630 on Build GRCh38
rs1016215748 Chr.1: 94173532-94173532 on Build GRCh38
rs1037478867 Chr.1: 94173531-94173531 on Build GRCh38
rs901135627 Chr.1: 94173455-94173455 on Build GRCh38
rs370165174 Chr.1: 94173868-94173868 on Build GRCh38
rs779545466 Chr.1: 94173647-94173647 on Build GRCh38
rs1042027056 Chr.1: 94173475-94173475 on Build GRCh38
rs780232939 Chr.1: 94173572-94173572 on Build GRCh38
rs772383920 Chr.1: 94173895-94173895 on Build GRCh38
rs146535456 Chr.1: 94173610-94173610 on Build GRCh38
rs372477728 Chr.1: 94173877-94173877 on Build GRCh38
rs114681812 Chr.1: 94173408-94173408 on Build GRCh38
rs575181267 Chr.1: 94173688-94173688 on Build GRCh38
rs769645920 Chr.1: 94173918-94173918 on Build GRCh38
rs894572694 Chr.1: 94173505-94173505 on Build GRCh38
rs1012077472 Chr.1: 94173597-94173597 on Build GRCh38
rs762433605 Chr.1: 94173843-94173844 on Build GRCh38
rs879105019 Chr.1: 94173843-94173843 on Build GRCh38
rs761956442 Chr.1: 94173819-94173819 on Build GRCh38
rs556186813 Chr.1: 94173395-94173395 on Build GRCh38
rs529470383 Chr.1: 94173851-94173851 on Build GRCh38
rs796978297 Chr.1: 94173481-94173481 on Build GRCh38
rs776627518 Chr.1: 94173910-94173910 on Build GRCh38
rs746581824 Chr.1: 94173651-94173651 on Build GRCh38
rs981904960 Chr.1: 94173649-94173649 on Build GRCh38
rs183018490 Chr.1: 94173635-94173635 on Build GRCh38
rs1039946022 Chr.1: 94173795-94173795 on Build GRCh38
rs748541171 Chr.1: 94173892-94173892 on Build GRCh38
rs529456105 Chr.1: 94173943-94173943 on Build GRCh38
rs755542411 Chr.1: 94173890-94173890 on Build GRCh38
rs764148433 Chr.1: 94173436-94173436 on Build GRCh38
rs762753584 Chr.1: 94173919-94173919 on Build GRCh38
rs1006746262 Chr.1: 94173524-94173524 on Build GRCh38
rs146805532 Chr.1: 94173833-94173835 on Build GRCh38
rs760533209 Chr.1: 94173835-94173836 on Build GRCh38
rs745605372 Chr.1: 94173624-94173624 on Build GRCh38
rs771369523 Chr.1: 94173878-94173878 on Build GRCh38
rs767442660 Chr.1: 94173841-94173841 on Build GRCh38
rs954435448 Chr.1: 94173401-94173401 on Build GRCh38
rs112984569 Chr.1: 94173545-94173545 on Build GRCh38
rs41292685 Chr.1: 94173431-94173431 on Build GRCh38
rs756253443 Chr.1: 94173865-94173865 on Build GRCh38

View Full Product Details