Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99864375-99864938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGL
Entrez Gene ID
178
Gene Name
amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase
Gene Aliases
GDE
UniGene
Hs.904
Gene Chromosome Location
Chr.1: 99850077-99924023 on Build GRCh38
Transcripts
NM_000028.2 NM_000642.2 NM_000643.2 NM_000644.2 NM_000645.2 NM_000646.2 XM_005270557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182224 c.628G>A p.V210I Chr.1: 99864553-99864553 on Build GRCh38
4004610 c.581G>A p.W194* Chr.1: 99864506-99864506 on Build GRCh38
5968703 c.595G>T p.V199F Chr.1: 99864571-99864571 on Build GRCh38
4572077 c.588T>A p.I196I Chr.1: 99864564-99864564 on Build GRCh38
423314 c.526G>C p.A176P Chr.1: 99864451-99864451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538389 Chr.1: 99864681-99864681 on Build GRCh38
rs780694207 Chr.1: 99864520-99864520 on Build GRCh38
rs794729208 Chr.1: 99864536-99864536 on Build GRCh38
rs758711507 Chr.1: 99864453-99864454 on Build GRCh38
rs756329937 Chr.1: 99864510-99864510 on Build GRCh38
rs752316561 Chr.1: 99864839-99864839 on Build GRCh38
rs913513569 Chr.1: 99864863-99864863 on Build GRCh38
rs945073165 Chr.1: 99864905-99864905 on Build GRCh38
rs61811105 Chr.1: 99864590-99864590 on Build GRCh38
rs1042265944 Chr.1: 99864906-99864906 on Build GRCh38
rs1053405327 Chr.1: 99864557-99864557 on Build GRCh38
rs947618516 Chr.1: 99864865-99864865 on Build GRCh38
rs767630899 Chr.1: 99864614-99864614 on Build GRCh38
rs1033729074 Chr.1: 99864643-99864643 on Build GRCh38
rs939137007 Chr.1: 99864910-99864910 on Build GRCh38
rs758219105 Chr.1: 99864602-99864602 on Build GRCh38
rs893488693 Chr.1: 99864581-99864581 on Build GRCh38
rs1016927218 Chr.1: 99864669-99864669 on Build GRCh38
rs540051403 Chr.1: 99864565-99864565 on Build GRCh38
rs906449125 Chr.1: 99864909-99864909 on Build GRCh38
rs766094427 Chr.1: 99864662-99864662 on Build GRCh38
rs955534228 Chr.1: 99864749-99864749 on Build GRCh38
rs537245173 Chr.1: 99864729-99864729 on Build GRCh38
rs370792293 Chr.1: 99864592-99864592 on Build GRCh38
rs765188709 Chr.1: 99864573-99864573 on Build GRCh38
rs747508907 Chr.1: 99864637-99864638 on Build GRCh38
rs149552795 Chr.1: 99864641-99864641 on Build GRCh38
rs781491884 Chr.1: 99864498-99864498 on Build GRCh38
rs528260114 Chr.1: 99864529-99864529 on Build GRCh38
rs963608925 Chr.1: 99864736-99864736 on Build GRCh38
rs775752564 Chr.1: 99864735-99864737 on Build GRCh38
rs1042768052 Chr.1: 99864892-99864892 on Build GRCh38
rs565950895 Chr.1: 99864740-99864740 on Build GRCh38
rs533806773 Chr.1: 99864451-99864451 on Build GRCh38
rs749851190 Chr.1: 99864412-99864412 on Build GRCh38
rs794727706 Chr.1: 99864460-99864463 on Build GRCh38
rs1053384869 Chr.1: 99864841-99864841 on Build GRCh38
rs916422016 Chr.1: 99864840-99864855 on Build GRCh38
rs765927790 Chr.1: 99864556-99864556 on Build GRCh38
rs905645820 Chr.1: 99864663-99864666 on Build GRCh38
rs987143206 Chr.1: 99864475-99864475 on Build GRCh38
rs959829837 Chr.1: 99864716-99864716 on Build GRCh38
rs778728216 Chr.1: 99864461-99864461 on Build GRCh38
rs748570736 Chr.1: 99864543-99864543 on Build GRCh38
rs750715340 Chr.1: 99864639-99864639 on Build GRCh38
rs910311069 Chr.1: 99864546-99864546 on Build GRCh38
rs943105883 Chr.1: 99864816-99864816 on Build GRCh38
rs200123416 Chr.1: 99864713-99864714 on Build GRCh38
rs539157 Chr.1: 99864739-99864739 on Build GRCh38
rs547384384 Chr.1: 99864886-99864886 on Build GRCh38
rs985503184 Chr.1: 99864803-99864803 on Build GRCh38
rs550589094 Chr.1: 99864558-99864558 on Build GRCh38
rs774542133 Chr.1: 99864535-99864535 on Build GRCh38
rs759509204 Chr.1: 99864571-99864571 on Build GRCh38
rs562042076 Chr.1: 99864538-99864538 on Build GRCh38
rs1005532114 Chr.1: 99864666-99864666 on Build GRCh38
rs559596492 Chr.1: 99864490-99864490 on Build GRCh38
rs985607474 Chr.1: 99864752-99864752 on Build GRCh38
rs202084554 Chr.1: 99864547-99864547 on Build GRCh38
rs954197282 Chr.1: 99864784-99864784 on Build GRCh38
rs760243154 Chr.1: 99864437-99864437 on Build GRCh38
rs914907240 Chr.1: 99864895-99864895 on Build GRCh38
rs912743645 Chr.1: 99864825-99864825 on Build GRCh38
rs772920146 Chr.1: 99864548-99864548 on Build GRCh38
rs910280890 Chr.1: 99864761-99864761 on Build GRCh38
rs753835367 Chr.1: 99864444-99864444 on Build GRCh38
rs766234642 Chr.1: 99864440-99864440 on Build GRCh38
rs757895966 Chr.1: 99864862-99864862 on Build GRCh38
rs767233801 Chr.1: 99864402-99864402 on Build GRCh38
rs186709388 Chr.1: 99864534-99864534 on Build GRCh38
rs751772979 Chr.1: 99864480-99864480 on Build GRCh38
rs760169396 Chr.1: 99864549-99864549 on Build GRCh38

View Full Product Details