Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111164196-111164776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L11
Entrez Gene ID
10018
Gene Name
BCL2 like 11
Gene Aliases
BAM, BIM, BOD
UniGene
Hs.469658
Gene Chromosome Location
Chr.2: 111120914-111168445 on Build GRCh38
Transcripts
NM_001204106.1 NM_001204107.1 NM_001204108.1 NM_001204109.1 NM_001204110.1 NM_001204111.1 NM_001204112.1 NM_006538.4 NM_138621.4 NM_138622.3 NM_138623.3 NM_138624.3 NM_138625.3 NM_138626.3 NM_138627.3 NM_207003.2 XM_005263551.2 XM_005263553.2 XM_005263554.2 XM_005263559.2 XM_017003101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2983157 c.563G>A p.R188H Chr.2: 111164197-111164197 on Build GRCh38
389356 c.585G>C p.W195C Chr.2: 111164219-111164219 on Build GRCh38
5930828 c.569T>A p.I190N Chr.2: 111164203-111164203 on Build GRCh38
7003210 c.561_562insA p.R188fs*>12 Chr.2: 111164195-111164196 on Build GRCh38
6969938 c.597A>T p.*199C Chr.2: 111164231-111164231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116174114 Chr.2: 111164530-111164530 on Build GRCh38
rs375598912 Chr.2: 111164223-111164223 on Build GRCh38
rs932294355 Chr.2: 111164488-111164488 on Build GRCh38
rs745731464 Chr.2: 111164254-111164254 on Build GRCh38
rs926826130 Chr.2: 111164416-111164416 on Build GRCh38
rs1002322433 Chr.2: 111164660-111164660 on Build GRCh38
rs114776970 Chr.2: 111164363-111164363 on Build GRCh38
rs914055558 Chr.2: 111164438-111164438 on Build GRCh38
rs993579085 Chr.2: 111164352-111164352 on Build GRCh38
rs996928814 Chr.2: 111164307-111164307 on Build GRCh38
rs1033676919 Chr.2: 111164702-111164702 on Build GRCh38
rs367628733 Chr.2: 111164527-111164527 on Build GRCh38
rs576839037 Chr.2: 111164330-111164330 on Build GRCh38
rs993367603 Chr.2: 111164622-111164622 on Build GRCh38
rs755418047 Chr.2: 111164242-111164243 on Build GRCh38
rs376786434 Chr.2: 111164249-111164249 on Build GRCh38
rs529861281 Chr.2: 111164485-111164485 on Build GRCh38
rs372164447 Chr.2: 111164219-111164219 on Build GRCh38
rs1014183726 Chr.2: 111164543-111164543 on Build GRCh38
rs577007904 Chr.2: 111164750-111164750 on Build GRCh38
rs964237302 Chr.2: 111164664-111164664 on Build GRCh38
rs572614693 Chr.2: 111164482-111164482 on Build GRCh38
rs1036935058 Chr.2: 111164539-111164539 on Build GRCh38
rs922881324 Chr.2: 111164704-111164704 on Build GRCh38
rs970725531 Chr.2: 111164374-111164374 on Build GRCh38
rs202136894 Chr.2: 111164244-111164244 on Build GRCh38
rs73954977 Chr.2: 111164258-111164258 on Build GRCh38
rs911222512 Chr.2: 111164652-111164652 on Build GRCh38
rs574285623 Chr.2: 111164643-111164643 on Build GRCh38
rs567035722 Chr.2: 111164342-111164343 on Build GRCh38
rs774026066 Chr.2: 111164250-111164250 on Build GRCh38
rs980787718 Chr.2: 111164395-111164395 on Build GRCh38
rs751424147 Chr.2: 111164658-111164658 on Build GRCh38
rs10199276 Chr.2: 111164263-111164263 on Build GRCh38
rs989769418 Chr.2: 111164430-111164430 on Build GRCh38
rs757169450 Chr.2: 111164707-111164707 on Build GRCh38
rs976961236 Chr.2: 111164686-111164686 on Build GRCh38
rs371903732 Chr.2: 111164638-111164638 on Build GRCh38
rs558804007 Chr.2: 111164271-111164271 on Build GRCh38
rs563321558 Chr.2: 111164575-111164575 on Build GRCh38
rs12996100 Chr.2: 111164318-111164318 on Build GRCh38
rs73954978 Chr.2: 111164339-111164339 on Build GRCh38
rs900820666 Chr.2: 111164286-111164286 on Build GRCh38
rs961858990 Chr.2: 111164615-111164615 on Build GRCh38
rs569726997 Chr.2: 111164529-111164529 on Build GRCh38
rs562014378 Chr.2: 111164332-111164332 on Build GRCh38
rs935767516 Chr.2: 111164743-111164743 on Build GRCh38
rs149549441 Chr.2: 111164567-111164567 on Build GRCh38
rs111743396 Chr.2: 111164299-111164299 on Build GRCh38
rs1041154135 Chr.2: 111164396-111164396 on Build GRCh38
rs552119326 Chr.2: 111164605-111164605 on Build GRCh38
rs777713287 Chr.2: 111164606-111164607 on Build GRCh38
rs546134766 Chr.2: 111164338-111164338 on Build GRCh38
rs115106683 Chr.2: 111164610-111164610 on Build GRCh38
rs958268112 Chr.2: 111164423-111164423 on Build GRCh38
rs984163795 Chr.2: 111164359-111164359 on Build GRCh38
rs894373659 Chr.2: 111164541-111164541 on Build GRCh38
rs1041214499 Chr.2: 111164278-111164278 on Build GRCh38
rs548535302 Chr.2: 111164441-111164441 on Build GRCh38
rs72948380 Chr.2: 111164742-111164742 on Build GRCh38
rs200016050 Chr.2: 111164243-111164243 on Build GRCh38
rs945143035 Chr.2: 111164275-111164275 on Build GRCh38
rs781678726 Chr.2: 111164278-111164282 on Build GRCh38
rs946205621 Chr.2: 111164712-111164712 on Build GRCh38
rs541295129 Chr.2: 111164533-111164533 on Build GRCh38
rs1005309532 Chr.2: 111164333-111164333 on Build GRCh38
rs7595612 Chr.2: 111164310-111164310 on Build GRCh38
rs901355868 Chr.2: 111164402-111164402 on Build GRCh38
rs12989846 Chr.2: 111164358-111164358 on Build GRCh38
rs961546852 Chr.2: 111164348-111164348 on Build GRCh38
rs989715745 Chr.2: 111164735-111164735 on Build GRCh38
rs574625366 Chr.2: 111164256-111164256 on Build GRCh38
rs945562895 Chr.2: 111164452-111164452 on Build GRCh38
rs904294480 Chr.2: 111164523-111164523 on Build GRCh38
rs115158848 Chr.2: 111164456-111164456 on Build GRCh38
rs554369951 Chr.2: 111164639-111164639 on Build GRCh38

View Full Product Details