Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55158347-55158944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7137824 c.1797G>T p.V599V Chr.14: 55158598-55158598 on Build GRCh38
1707362 c.1715C>T p.A572V Chr.14: 55158680-55158680 on Build GRCh38
1370157 c.1757G>T p.R586I Chr.14: 55158638-55158638 on Build GRCh38
6645029 c.1757_1758delGA p.R586fs*2 Chr.14: 55158637-55158638 on Build GRCh38
6645028 c.1730C>T p.A577V Chr.14: 55158665-55158665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778125741 Chr.14: 55158477-55158477 on Build GRCh38
rs140729390 Chr.14: 55158630-55158630 on Build GRCh38
rs549173191 Chr.14: 55158672-55158672 on Build GRCh38
rs770323930 Chr.14: 55158666-55158668 on Build GRCh38
rs781312686 Chr.14: 55158685-55158685 on Build GRCh38
rs777845595 Chr.14: 55158723-55158723 on Build GRCh38
rs766073374 Chr.14: 55158512-55158514 on Build GRCh38
rs967624560 Chr.14: 55158826-55158830 on Build GRCh38
rs139031197 Chr.14: 55158568-55158568 on Build GRCh38
rs769773432 Chr.14: 55158702-55158702 on Build GRCh38
rs758520984 Chr.14: 55158600-55158600 on Build GRCh38
rs552675454 Chr.14: 55158902-55158902 on Build GRCh38
rs773805210 Chr.14: 55158653-55158653 on Build GRCh38
rs778975358 Chr.14: 55158496-55158496 on Build GRCh38
rs12881211 Chr.14: 55158409-55158409 on Build GRCh38
rs764520620 Chr.14: 55158586-55158586 on Build GRCh38
rs372390277 Chr.14: 55158649-55158649 on Build GRCh38
rs754238015 Chr.14: 55158590-55158590 on Build GRCh38
rs1006866071 Chr.14: 55158404-55158405 on Build GRCh38
rs901204844 Chr.14: 55158797-55158805 on Build GRCh38
rs952791015 Chr.14: 55158402-55158402 on Build GRCh38
rs756425328 Chr.14: 55158572-55158572 on Build GRCh38
rs372360305 Chr.14: 55158511-55158511 on Build GRCh38
rs947769645 Chr.14: 55158779-55158779 on Build GRCh38
rs778922367 Chr.14: 55158596-55158596 on Build GRCh38
rs569948136 Chr.14: 55158818-55158818 on Build GRCh38
rs139156235 Chr.14: 55158491-55158492 on Build GRCh38
rs570390931 Chr.14: 55158873-55158873 on Build GRCh38
rs764513265 Chr.14: 55158850-55158850 on Build GRCh38
rs773872088 Chr.14: 55158722-55158722 on Build GRCh38
rs746686691 Chr.14: 55158500-55158500 on Build GRCh38
rs746040103 Chr.14: 55158500-55158500 on Build GRCh38
rs534670922 Chr.14: 55158901-55158901 on Build GRCh38
rs993836067 Chr.14: 55158450-55158450 on Build GRCh38
rs1046708727 Chr.14: 55158615-55158615 on Build GRCh38
rs542092291 Chr.14: 55158452-55158452 on Build GRCh38
rs780251913 Chr.14: 55158604-55158604 on Build GRCh38
rs978873145 Chr.14: 55158832-55158832 on Build GRCh38
rs145664806 Chr.14: 55158510-55158510 on Build GRCh38
rs757782338 Chr.14: 55158594-55158594 on Build GRCh38
rs372884646 Chr.14: 55158671-55158671 on Build GRCh38
rs529935301 Chr.14: 55158733-55158733 on Build GRCh38
rs1003363104 Chr.14: 55158854-55158854 on Build GRCh38
rs766110350 Chr.14: 55158567-55158567 on Build GRCh38
rs185926266 Chr.14: 55158650-55158650 on Build GRCh38
rs57075984 Chr.14: 55158417-55158417 on Build GRCh38
rs763346369 Chr.14: 55158731-55158733 on Build GRCh38
rs368248560 Chr.14: 55158486-55158486 on Build GRCh38
rs780198207 Chr.14: 55158679-55158679 on Build GRCh38
rs776508223 Chr.14: 55158637-55158637 on Build GRCh38
rs138277054 Chr.14: 55158652-55158652 on Build GRCh38
rs546157515 Chr.14: 55158501-55158501 on Build GRCh38
rs200878291 Chr.14: 55158508-55158508 on Build GRCh38
rs888519264 Chr.14: 55158714-55158714 on Build GRCh38
rs764314388 Chr.14: 55158666-55158666 on Build GRCh38
rs906390391 Chr.14: 55158847-55158847 on Build GRCh38
rs747260725 Chr.14: 55158618-55158618 on Build GRCh38
rs562731400 Chr.14: 55158896-55158896 on Build GRCh38
rs530740891 Chr.14: 55158821-55158821 on Build GRCh38
rs998207351 Chr.14: 55158797-55158797 on Build GRCh38
rs375526239 Chr.14: 55158574-55158574 on Build GRCh38
rs138214529 Chr.14: 55158782-55158782 on Build GRCh38
rs773930677 Chr.14: 55158762-55158762 on Build GRCh38
rs769826494 Chr.14: 55158645-55158645 on Build GRCh38
rs768242490 Chr.14: 55158515-55158518 on Build GRCh38
rs1020917343 Chr.14: 55158801-55158804 on Build GRCh38
rs545178100 Chr.14: 55158406-55158406 on Build GRCh38
rs754259895 Chr.14: 55158667-55158667 on Build GRCh38
rs374278216 Chr.14: 55158565-55158565 on Build GRCh38
rs776177444 Chr.14: 55158590-55158590 on Build GRCh38
rs747999185 Chr.14: 55158638-55158638 on Build GRCh38
rs991009469 Chr.14: 55158872-55158872 on Build GRCh38
rs143015786 Chr.14: 55158866-55158866 on Build GRCh38
rs749868690 Chr.14: 55158478-55158478 on Build GRCh38
rs953412354 Chr.14: 55158583-55158583 on Build GRCh38
rs888174777 Chr.14: 55158457-55158457 on Build GRCh38
rs774007246 Chr.14: 55158542-55158542 on Build GRCh38
rs755262255 Chr.14: 55158683-55158683 on Build GRCh38
rs767025301 Chr.14: 55158660-55158660 on Build GRCh38
rs1036614701 Chr.14: 55158389-55158389 on Build GRCh38
rs1018720255 Chr.14: 55158442-55158442 on Build GRCh38
rs929652399 Chr.14: 55158686-55158686 on Build GRCh38
rs1012049689 Chr.14: 55158466-55158466 on Build GRCh38
rs761298387 Chr.14: 55158595-55158596 on Build GRCh38
rs761160665 Chr.14: 55158810-55158810 on Build GRCh38
rs1048024748 Chr.14: 55158720-55158720 on Build GRCh38
rs1031358091 Chr.14: 55158864-55158864 on Build GRCh38
rs747938429 Chr.14: 55158700-55158700 on Build GRCh38
rs377135023 Chr.14: 55158772-55158772 on Build GRCh38
rs745916502 Chr.14: 55158597-55158597 on Build GRCh38
rs369650574 Chr.14: 55158636-55158636 on Build GRCh38
rs12882439 Chr.14: 55158408-55158408 on Build GRCh38
rs553139401 Chr.14: 55158375-55158375 on Build GRCh38
rs761825301 Chr.14: 55158509-55158509 on Build GRCh38
rs190904740 Chr.14: 55158651-55158651 on Build GRCh38
rs896847919 Chr.14: 55158445-55158445 on Build GRCh38
rs111324853 Chr.14: 55158475-55158475 on Build GRCh38

View Full Product Details