Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10488124-10488615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4

SNP Detail

SNP ID Chromosome Location
rs78647248 Chr.18: 10488180-10488180 on Build GRCh38
rs879469647 Chr.18: 10488268-10488268 on Build GRCh38
rs961383295 Chr.18: 10488407-10488407 on Build GRCh38
rs904213757 Chr.18: 10488526-10488526 on Build GRCh38
rs934334812 Chr.18: 10488535-10488535 on Build GRCh38
rs571884835 Chr.18: 10488510-10488510 on Build GRCh38
rs75833303 Chr.18: 10488167-10488167 on Build GRCh38
rs1043962915 Chr.18: 10488521-10488521 on Build GRCh38
rs754107893 Chr.18: 10488184-10488184 on Build GRCh38
rs371805137 Chr.18: 10488583-10488583 on Build GRCh38
rs957797637 Chr.18: 10488324-10488324 on Build GRCh38
rs386801139 Chr.18: 10488335-10488343 on Build GRCh38
rs953951531 Chr.18: 10488308-10488308 on Build GRCh38
rs545186727 Chr.18: 10488340-10488340 on Build GRCh38
rs59177178 Chr.18: 10488342-10488342 on Build GRCh38
rs981024769 Chr.18: 10488304-10488304 on Build GRCh38
rs1045648 Chr.18: 10488453-10488453 on Build GRCh38
rs1031404754 Chr.18: 10488286-10488286 on Build GRCh38
rs747524617 Chr.18: 10488505-10488505 on Build GRCh38
rs752068746 Chr.18: 10488222-10488222 on Build GRCh38
rs138936528 Chr.18: 10488437-10488437 on Build GRCh38
rs1017368999 Chr.18: 10488398-10488398 on Build GRCh38
rs201343606 Chr.18: 10488339-10488340 on Build GRCh38
rs990952665 Chr.18: 10488493-10488493 on Build GRCh38
rs117564812 Chr.18: 10488233-10488233 on Build GRCh38
rs755195456 Chr.18: 10488191-10488191 on Build GRCh38
rs141508733 Chr.18: 10488537-10488537 on Build GRCh38
rs527343497 Chr.18: 10488352-10488352 on Build GRCh38
rs190711946 Chr.18: 10488569-10488569 on Build GRCh38
rs748126047 Chr.18: 10488293-10488293 on Build GRCh38
rs764392330 Chr.18: 10488197-10488197 on Build GRCh38
rs771858805 Chr.18: 10488471-10488471 on Build GRCh38
rs568975850 Chr.18: 10488323-10488323 on Build GRCh38
rs11558327 Chr.18: 10488576-10488576 on Build GRCh38
rs866053729 Chr.18: 10488546-10488546 on Build GRCh38
rs510337 Chr.18: 10488303-10488303 on Build GRCh38
rs746939603 Chr.18: 10488567-10488567 on Build GRCh38
rs946986009 Chr.18: 10488511-10488511 on Build GRCh38
rs878862118 Chr.18: 10488182-10488182 on Build GRCh38
rs1051393252 Chr.18: 10488409-10488409 on Build GRCh38
rs772919649 Chr.18: 10488473-10488476 on Build GRCh38
rs576530395 Chr.18: 10488192-10488192 on Build GRCh38
rs995529711 Chr.18: 10488159-10488159 on Build GRCh38
rs987877040 Chr.18: 10488326-10488326 on Build GRCh38
rs57479247 Chr.18: 10488343-10488343 on Build GRCh38
rs965621911 Chr.18: 10488290-10488290 on Build GRCh38
rs898649109 Chr.18: 10488448-10488448 on Build GRCh38
rs969727718 Chr.18: 10488541-10488541 on Build GRCh38
rs145170053 Chr.18: 10488428-10488428 on Build GRCh38
rs913604888 Chr.18: 10488345-10488345 on Build GRCh38
rs898590065 Chr.18: 10488153-10488155 on Build GRCh38
rs186139406 Chr.18: 10488441-10488441 on Build GRCh38
rs547931549 Chr.18: 10488565-10488565 on Build GRCh38
rs545041021 Chr.18: 10488335-10488337 on Build GRCh38

View Full Product Details