Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104783177-104783730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

SNP Detail

SNP ID Chromosome Location
rs79123530 Chr.9: 104783675-104783675 on Build GRCh38
rs866428233 Chr.9: 104783447-104783447 on Build GRCh38
rs917061398 Chr.9: 104783689-104783689 on Build GRCh38
rs886063301 Chr.9: 104783638-104783638 on Build GRCh38
rs755419992 Chr.9: 104783462-104783462 on Build GRCh38
rs539558014 Chr.9: 104783441-104783442 on Build GRCh38
rs898285983 Chr.9: 104783638-104783638 on Build GRCh38
rs971912904 Chr.9: 104783427-104783427 on Build GRCh38
rs950621960 Chr.9: 104783650-104783650 on Build GRCh38
rs183535818 Chr.9: 104783474-104783474 on Build GRCh38
rs993174687 Chr.9: 104783663-104783663 on Build GRCh38
rs1037309867 Chr.9: 104783284-104783284 on Build GRCh38
rs1024381427 Chr.9: 104783423-104783423 on Build GRCh38
rs954094495 Chr.9: 104783310-104783310 on Build GRCh38
rs555000737 Chr.9: 104783492-104783492 on Build GRCh38
rs1012936854 Chr.9: 104783402-104783402 on Build GRCh38
rs556998758 Chr.9: 104783700-104783700 on Build GRCh38
rs4149338 Chr.9: 104783622-104783622 on Build GRCh38
rs1056929833 Chr.9: 104783417-104783417 on Build GRCh38
rs758655998 Chr.9: 104783611-104783611 on Build GRCh38
rs552787469 Chr.9: 104783620-104783620 on Build GRCh38
rs189826506 Chr.9: 104783259-104783259 on Build GRCh38
rs1029762063 Chr.9: 104783584-104783584 on Build GRCh38
rs995391814 Chr.9: 104783332-104783332 on Build GRCh38
rs962672002 Chr.9: 104783357-104783357 on Build GRCh38
rs886063300 Chr.9: 104783440-104783440 on Build GRCh38
rs954224850 Chr.9: 104783592-104783592 on Build GRCh38
rs929247205 Chr.9: 104783407-104783407 on Build GRCh38
rs1007039989 Chr.9: 104783602-104783602 on Build GRCh38
rs765718505 Chr.9: 104783548-104783548 on Build GRCh38
rs1031911804 Chr.9: 104783249-104783249 on Build GRCh38
rs534504547 Chr.9: 104783609-104783609 on Build GRCh38
rs1000828107 Chr.9: 104783220-104783220 on Build GRCh38
rs753919065 Chr.9: 104783235-104783235 on Build GRCh38
rs958673244 Chr.9: 104783399-104783399 on Build GRCh38
rs912309547 Chr.9: 104783616-104783616 on Build GRCh38
rs565619592 Chr.9: 104783383-104783383 on Build GRCh38
rs1036792572 Chr.9: 104783625-104783625 on Build GRCh38
rs569604699 Chr.9: 104783441-104783441 on Build GRCh38
rs1033601541 Chr.9: 104783360-104783360 on Build GRCh38
rs1053238248 Chr.9: 104783532-104783532 on Build GRCh38
rs898402597 Chr.9: 104783311-104783311 on Build GRCh38
rs879092419 Chr.9: 104783701-104783701 on Build GRCh38
rs142039624 Chr.9: 104783577-104783577 on Build GRCh38
rs998362887 Chr.9: 104783269-104783269 on Build GRCh38
rs932304625 Chr.9: 104783599-104783599 on Build GRCh38
rs374568251 Chr.9: 104783615-104783615 on Build GRCh38
rs977615117 Chr.9: 104783428-104783428 on Build GRCh38

View Full Product Details