Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41598846-41599433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAT4
Entrez Gene ID
137964
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7
UniGene
Hs.355753
Gene Chromosome Location
Chr.8: 41578188-41625001 on Build GRCh38
Transcripts
NM_178819.3 XM_011544392.2 XM_011544393.1 XM_011544394.1 XM_017013042.1 XM_017013043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182229 c.92T>C p.I31T Chr.8: 41599231-41599231 on Build GRCh38
5014015 c.52T>A p.S18T Chr.8: 41599191-41599191 on Build GRCh38
5017336 c.70_71insC p.L25fs*76 Chr.8: 41599209-41599210 on Build GRCh38
7347930 c.141delG p.M47fs*5 Chr.8: 41599280-41599280 on Build GRCh38
3649162 c.80T>A p.L27H Chr.8: 41599219-41599219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201982831 Chr.8: 41599165-41599165 on Build GRCh38
rs188299415 Chr.8: 41599093-41599093 on Build GRCh38
rs35914617 Chr.8: 41599054-41599055 on Build GRCh38
rs771369212 Chr.8: 41599039-41599039 on Build GRCh38
rs776052125 Chr.8: 41599325-41599325 on Build GRCh38
rs578189498 Chr.8: 41598992-41598992 on Build GRCh38
rs1006313148 Chr.8: 41598966-41598966 on Build GRCh38
rs750845516 Chr.8: 41599238-41599238 on Build GRCh38
rs536341749 Chr.8: 41598993-41598993 on Build GRCh38
rs534990402 Chr.8: 41599144-41599144 on Build GRCh38
rs947449973 Chr.8: 41599372-41599372 on Build GRCh38
rs963780853 Chr.8: 41599384-41599384 on Build GRCh38
rs942231695 Chr.8: 41599217-41599217 on Build GRCh38
rs927913739 Chr.8: 41598985-41598985 on Build GRCh38
rs905943273 Chr.8: 41599002-41599002 on Build GRCh38
rs757200224 Chr.8: 41599287-41599287 on Build GRCh38
rs771676659 Chr.8: 41599205-41599205 on Build GRCh38
rs916450915 Chr.8: 41598943-41598943 on Build GRCh38
rs193289264 Chr.8: 41599130-41599130 on Build GRCh38
rs753834903 Chr.8: 41599286-41599286 on Build GRCh38
rs765600184 Chr.8: 41599231-41599231 on Build GRCh38
rs368558123 Chr.8: 41599106-41599106 on Build GRCh38
rs761299619 Chr.8: 41599342-41599342 on Build GRCh38
rs969162986 Chr.8: 41599263-41599263 on Build GRCh38
rs780311042 Chr.8: 41599135-41599135 on Build GRCh38
rs559700587 Chr.8: 41599355-41599355 on Build GRCh38
rs770829363 Chr.8: 41599096-41599096 on Build GRCh38
rs369654595 Chr.8: 41599132-41599132 on Build GRCh38
rs72554082 Chr.8: 41599060-41599060 on Build GRCh38
rs779620977 Chr.8: 41599313-41599313 on Build GRCh38
rs554740180 Chr.8: 41599177-41599177 on Build GRCh38
rs868541337 Chr.8: 41598951-41598951 on Build GRCh38
rs749192194 Chr.8: 41599173-41599173 on Build GRCh38
rs907594739 Chr.8: 41598891-41598891 on Build GRCh38
rs368137766 Chr.8: 41599006-41599006 on Build GRCh38
rs758595297 Chr.8: 41599188-41599189 on Build GRCh38
rs749797939 Chr.8: 41598937-41598937 on Build GRCh38
rs372797523 Chr.8: 41599115-41599115 on Build GRCh38
rs774174022 Chr.8: 41599099-41599099 on Build GRCh38
rs375431670 Chr.8: 41599128-41599128 on Build GRCh38
rs374092780 Chr.8: 41599312-41599312 on Build GRCh38
rs751787240 Chr.8: 41599256-41599256 on Build GRCh38
rs138968347 Chr.8: 41599353-41599353 on Build GRCh38
rs1047907328 Chr.8: 41598918-41598918 on Build GRCh38
rs1004936234 Chr.8: 41599121-41599121 on Build GRCh38
rs79557732 Chr.8: 41599321-41599321 on Build GRCh38
rs1010164956 Chr.8: 41599347-41599347 on Build GRCh38
rs777623777 Chr.8: 41599172-41599172 on Build GRCh38
rs762211959 Chr.8: 41599129-41599129 on Build GRCh38
rs770353727 Chr.8: 41599344-41599344 on Build GRCh38
rs1053144842 Chr.8: 41599001-41599001 on Build GRCh38
rs774801575 Chr.8: 41599109-41599109 on Build GRCh38
rs866666749 Chr.8: 41599267-41599267 on Build GRCh38
rs763373728 Chr.8: 41599241-41599241 on Build GRCh38
rs963184156 Chr.8: 41598900-41598900 on Build GRCh38
rs755038220 Chr.8: 41599266-41599266 on Build GRCh38
rs778977102 Chr.8: 41599303-41599303 on Build GRCh38
rs900710669 Chr.8: 41599036-41599036 on Build GRCh38
rs947972990 Chr.8: 41598950-41598950 on Build GRCh38
rs145171978 Chr.8: 41599351-41599351 on Build GRCh38
rs547752246 Chr.8: 41599377-41599377 on Build GRCh38
rs139630566 Chr.8: 41599304-41599304 on Build GRCh38
rs963747917 Chr.8: 41599120-41599120 on Build GRCh38
rs933537716 Chr.8: 41598915-41598915 on Build GRCh38
rs902131341 Chr.8: 41598892-41598892 on Build GRCh38
rs762913166 Chr.8: 41598929-41598929 on Build GRCh38
rs1018033566 Chr.8: 41599381-41599381 on Build GRCh38
rs1043238999 Chr.8: 41599034-41599034 on Build GRCh38
rs574733240 Chr.8: 41599178-41599178 on Build GRCh38
rs760142023 Chr.8: 41599218-41599218 on Build GRCh38
rs375002166 Chr.8: 41599221-41599221 on Build GRCh38
rs768188787 Chr.8: 41599220-41599220 on Build GRCh38
rs766550823 Chr.8: 41599253-41599253 on Build GRCh38
rs1035680822 Chr.8: 41599028-41599028 on Build GRCh38
rs527586046 Chr.8: 41599357-41599357 on Build GRCh38
rs745525354 Chr.8: 41599200-41599200 on Build GRCh38
rs764572267 Chr.8: 41599124-41599124 on Build GRCh38
rs781295731 Chr.8: 41599279-41599279 on Build GRCh38
rs752831623 Chr.8: 41599103-41599106 on Build GRCh38
rs1011697621 Chr.8: 41599020-41599020 on Build GRCh38
rs775200023 Chr.8: 41599208-41599208 on Build GRCh38
rs922245732 Chr.8: 41599138-41599138 on Build GRCh38
rs886565884 Chr.8: 41598921-41598921 on Build GRCh38
rs762266933 Chr.8: 41599227-41599227 on Build GRCh38
rs150813531 Chr.8: 41599148-41599148 on Build GRCh38
rs768082923 Chr.8: 41599322-41599322 on Build GRCh38
rs768242581 Chr.8: 41599240-41599240 on Build GRCh38
rs959959197 Chr.8: 41599035-41599035 on Build GRCh38

View Full Product Details