Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243502970-243503561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2 XM_011544014.2 XM_016999985.1

SNP Detail

SNP ID Chromosome Location
rs915805696 Chr.1: 243503428-243503428 on Build GRCh38
rs954174200 Chr.1: 243503130-243503130 on Build GRCh38
rs559552412 Chr.1: 243503489-243503489 on Build GRCh38
rs957290949 Chr.1: 243503122-243503122 on Build GRCh38
rs911983788 Chr.1: 243503240-243503240 on Build GRCh38
rs761715712 Chr.1: 243503492-243503492 on Build GRCh38
rs538216688 Chr.1: 243503395-243503396 on Build GRCh38
rs564075522 Chr.1: 243503179-243503179 on Build GRCh38
rs577602285 Chr.1: 243503321-243503321 on Build GRCh38
rs907025612 Chr.1: 243503490-243503490 on Build GRCh38
rs185750644 Chr.1: 243503102-243503102 on Build GRCh38
rs549605049 Chr.1: 243503482-243503482 on Build GRCh38
rs113768661 Chr.1: 243503285-243503285 on Build GRCh38
rs760274781 Chr.1: 243503422-243503422 on Build GRCh38
rs372540763 Chr.1: 243503052-243503052 on Build GRCh38
rs1010960837 Chr.1: 243503419-243503419 on Build GRCh38
rs899206795 Chr.1: 243503269-243503269 on Build GRCh38
rs529465390 Chr.1: 243503381-243503381 on Build GRCh38
rs557453245 Chr.1: 243503374-243503374 on Build GRCh38
rs763872282 Chr.1: 243503458-243503458 on Build GRCh38
rs920007017 Chr.1: 243503421-243503421 on Build GRCh38
rs1014821867 Chr.1: 243503048-243503048 on Build GRCh38
rs954308013 Chr.1: 243503392-243503393 on Build GRCh38
rs570624987 Chr.1: 243503138-243503138 on Build GRCh38
rs531329460 Chr.1: 243503217-243503217 on Build GRCh38
rs1011170037 Chr.1: 243503520-243503520 on Build GRCh38
rs748250019 Chr.1: 243503003-243503003 on Build GRCh38
rs1014440470 Chr.1: 243503334-243503334 on Build GRCh38
rs961874247 Chr.1: 243503403-243503403 on Build GRCh38
rs34770086 Chr.1: 243503393-243503394 on Build GRCh38
rs1024387221 Chr.1: 243503352-243503352 on Build GRCh38
rs1001942208 Chr.1: 243503362-243503362 on Build GRCh38
rs943401551 Chr.1: 243503247-243503247 on Build GRCh38
rs1033272588 Chr.1: 243503118-243503118 on Build GRCh38
rs1024026532 Chr.1: 243503050-243503050 on Build GRCh38
rs7521027 Chr.1: 243503034-243503034 on Build GRCh38
rs911763765 Chr.1: 243503488-243503488 on Build GRCh38
rs540642563 Chr.1: 243503279-243503279 on Build GRCh38
rs879188187 Chr.1: 243503101-243503102 on Build GRCh38
rs1031874058 Chr.1: 243503322-243503322 on Build GRCh38
rs972839248 Chr.1: 243503406-243503406 on Build GRCh38
rs957750741 Chr.1: 243503415-243503415 on Build GRCh38
rs1048002666 Chr.1: 243503310-243503310 on Build GRCh38
rs769595591 Chr.1: 243503177-243503177 on Build GRCh38
rs1039130545 Chr.1: 243503267-243503267 on Build GRCh38
rs555005002 Chr.1: 243503205-243503205 on Build GRCh38
rs35197966 Chr.1: 243503278-243503279 on Build GRCh38
rs948597839 Chr.1: 243503441-243503441 on Build GRCh38
rs1024690912 Chr.1: 243503063-243503063 on Build GRCh38
rs539325565 Chr.1: 243503154-243503154 on Build GRCh38
rs1052835521 Chr.1: 243503496-243503496 on Build GRCh38
rs774630157 Chr.1: 243503270-243503270 on Build GRCh38
rs190950461 Chr.1: 243503186-243503186 on Build GRCh38
rs1019722598 Chr.1: 243503394-243503394 on Build GRCh38
rs760118113 Chr.1: 243503440-243503440 on Build GRCh38
rs957934327 Chr.1: 243503126-243503126 on Build GRCh38
rs931345244 Chr.1: 243503218-243503218 on Build GRCh38
rs529091593 Chr.1: 243503516-243503516 on Build GRCh38
rs940139371 Chr.1: 243503163-243503163 on Build GRCh38
rs535312241 Chr.1: 243503202-243503202 on Build GRCh38
rs970426157 Chr.1: 243503092-243503092 on Build GRCh38
rs551766297 Chr.1: 243503103-243503103 on Build GRCh38
rs760538042 Chr.1: 243503059-243503061 on Build GRCh38
rs966990985 Chr.1: 243503423-243503423 on Build GRCh38
rs1026982631 Chr.1: 243503031-243503031 on Build GRCh38
rs912618614 Chr.1: 243503145-243503145 on Build GRCh38
rs1048170948 Chr.1: 243503527-243503527 on Build GRCh38
rs551610301 Chr.1: 243503499-243503499 on Build GRCh38
rs771886035 Chr.1: 243503329-243503329 on Build GRCh38
rs183400807 Chr.1: 243503491-243503491 on Build GRCh38
rs552114137 Chr.1: 243503126-243503129 on Build GRCh38
rs376781950 Chr.1: 243503529-243503529 on Build GRCh38
rs1048967642 Chr.1: 243503228-243503228 on Build GRCh38
rs151083954 Chr.1: 243503359-243503359 on Build GRCh38

View Full Product Details