Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57473775-57474373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP9
Entrez Gene ID
64333
Gene Name
Rho GTPase activating protein 9
Gene Aliases
10C, RGL1
UniGene
Hs.437126
Gene Chromosome Location
Chr.12: 57472255-57488814 on Build GRCh38
Transcripts
NM_001080156.2 NM_001080157.1 NM_001319850.1 NM_001319851.1 NM_001319852.1 NM_032496.3 XM_005269083.2 XM_005269084.2 XM_005269085.2 XM_011538656.2 XM_011538657.2 XM_011538658.2 XM_011538659.2 XM_017019800.1 XM_017019801.1 XM_017019802.1 XM_017019803.1 XM_017019804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6073283 c.1915C>A p.R639R Chr.12: 57474102-57474102 on Build GRCh38
1183467 c.1928A>C p.Q643P Chr.12: 57474089-57474089 on Build GRCh38
4569542 c.1889T>A p.V630D Chr.12: 57474128-57474128 on Build GRCh38
4662968 c.1872G>T p.W624C Chr.12: 57474145-57474145 on Build GRCh38
6537725 c.1852G>C p.D618H Chr.12: 57474165-57474165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117248179 Chr.12: 57473841-57473841 on Build GRCh38
rs747924330 Chr.12: 57474171-57474171 on Build GRCh38
rs2280140 Chr.12: 57474006-57474006 on Build GRCh38
rs911087292 Chr.12: 57473877-57473877 on Build GRCh38
rs774556128 Chr.12: 57474192-57474192 on Build GRCh38
rs1014774161 Chr.12: 57474205-57474205 on Build GRCh38
rs932621733 Chr.12: 57473965-57473965 on Build GRCh38
rs145518718 Chr.12: 57473819-57473819 on Build GRCh38
rs141789438 Chr.12: 57474053-57474053 on Build GRCh38
rs754710366 Chr.12: 57474066-57474066 on Build GRCh38
rs952056554 Chr.12: 57474158-57474158 on Build GRCh38
rs145106882 Chr.12: 57474349-57474349 on Build GRCh38
rs368092491 Chr.12: 57474170-57474170 on Build GRCh38
rs926265111 Chr.12: 57474324-57474324 on Build GRCh38
rs963977597 Chr.12: 57473880-57473880 on Build GRCh38
rs182354376 Chr.12: 57473845-57473845 on Build GRCh38
rs61758883 Chr.12: 57474102-57474102 on Build GRCh38
rs774691709 Chr.12: 57474087-57474087 on Build GRCh38
rs563318993 Chr.12: 57474207-57474207 on Build GRCh38
rs767768063 Chr.12: 57474107-57474107 on Build GRCh38
rs771132829 Chr.12: 57473975-57473975 on Build GRCh38
rs1013964948 Chr.12: 57474080-57474080 on Build GRCh38
rs745931260 Chr.12: 57474193-57474193 on Build GRCh38
rs190747515 Chr.12: 57473820-57473820 on Build GRCh38
rs754376059 Chr.12: 57474130-57474130 on Build GRCh38
rs532993 Chr.12: 57473943-57473943 on Build GRCh38
rs1010261059 Chr.12: 57473812-57473812 on Build GRCh38
rs926412804 Chr.12: 57474341-57474341 on Build GRCh38
rs753131180 Chr.12: 57474003-57474003 on Build GRCh38
rs754731556 Chr.12: 57474169-57474169 on Build GRCh38
rs61754174 Chr.12: 57474124-57474124 on Build GRCh38
rs138468362 Chr.12: 57474322-57474322 on Build GRCh38
rs976852816 Chr.12: 57474249-57474249 on Build GRCh38
rs113759971 Chr.12: 57474331-57474331 on Build GRCh38
rs970091922 Chr.12: 57473825-57473825 on Build GRCh38
rs764537790 Chr.12: 57474129-57474129 on Build GRCh38
rs1037854769 Chr.12: 57473977-57473977 on Build GRCh38
rs534874949 Chr.12: 57474079-57474079 on Build GRCh38
rs537397304 Chr.12: 57473954-57473954 on Build GRCh38
rs777819380 Chr.12: 57474182-57474182 on Build GRCh38
rs143453105 Chr.12: 57474270-57474270 on Build GRCh38
rs577394658 Chr.12: 57473979-57473979 on Build GRCh38
rs764590740 Chr.12: 57474031-57474031 on Build GRCh38
rs767972366 Chr.12: 57473998-57473998 on Build GRCh38
rs747896422 Chr.12: 57474074-57474074 on Build GRCh38
rs936216135 Chr.12: 57474032-57474032 on Build GRCh38
rs554505159 Chr.12: 57473889-57473889 on Build GRCh38
rs771119706 Chr.12: 57474184-57474184 on Build GRCh38
rs960001130 Chr.12: 57473810-57473810 on Build GRCh38
rs149363709 Chr.12: 57474148-57474148 on Build GRCh38
rs746718619 Chr.12: 57474055-57474055 on Build GRCh38
rs148815869 Chr.12: 57473974-57473974 on Build GRCh38
rs1046720407 Chr.12: 57473938-57473938 on Build GRCh38
rs202095270 Chr.12: 57474028-57474028 on Build GRCh38
rs781103708 Chr.12: 57474073-57474073 on Build GRCh38
rs751859447 Chr.12: 57473997-57473997 on Build GRCh38
rs922539866 Chr.12: 57473914-57473914 on Build GRCh38
rs929604130 Chr.12: 57473937-57473937 on Build GRCh38
rs941106064 Chr.12: 57474327-57474327 on Build GRCh38
rs1001597227 Chr.12: 57474131-57474131 on Build GRCh38
rs937467423 Chr.12: 57474344-57474344 on Build GRCh38
rs374309877 Chr.12: 57474308-57474308 on Build GRCh38
rs751161941 Chr.12: 57474155-57474155 on Build GRCh38
rs769793763 Chr.12: 57474076-57474076 on Build GRCh38
rs983277758 Chr.12: 57473862-57473862 on Build GRCh38
rs976555823 Chr.12: 57474266-57474266 on Build GRCh38
rs749879144 Chr.12: 57474046-57474046 on Build GRCh38
rs968830368 Chr.12: 57474127-57474127 on Build GRCh38
rs751591604 Chr.12: 57474274-57474274 on Build GRCh38
rs775552240 Chr.12: 57474214-57474214 on Build GRCh38
rs375878853 Chr.12: 57474123-57474123 on Build GRCh38
rs771172745 Chr.12: 57474082-57474082 on Build GRCh38
rs896074672 Chr.12: 57474101-57474101 on Build GRCh38
rs773301833 Chr.12: 57474081-57474081 on Build GRCh38
rs1023926908 Chr.12: 57473821-57473821 on Build GRCh38
rs982410662 Chr.12: 57474320-57474320 on Build GRCh38
rs757876238 Chr.12: 57474132-57474132 on Build GRCh38
rs968798279 Chr.12: 57473833-57473833 on Build GRCh38
rs756001973 Chr.12: 57474173-57474173 on Build GRCh38
rs749285896 Chr.12: 57474183-57474183 on Build GRCh38
rs779643644 Chr.12: 57474054-57474054 on Build GRCh38

View Full Product Details