Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33008964-33009471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK2
Entrez Gene ID
204
Gene Name
adenylate kinase 2
Gene Aliases
ADK2
UniGene
Hs.470907
Gene Chromosome Location
Chr.1: 33007940-33036911 on Build GRCh38
Transcripts
NM_001199199.1 NM_001319139.1 NM_001319141.1 NM_001319142.1 NM_013411.4 NR_134976.1

SNP Detail

SNP ID Chromosome Location
rs1054811159 Chr.1: 33009054-33009054 on Build GRCh38
rs895830494 Chr.1: 33009267-33009267 on Build GRCh38
rs970300836 Chr.1: 33009359-33009359 on Build GRCh38
rs1029143604 Chr.1: 33009259-33009259 on Build GRCh38
rs774553995 Chr.1: 33009225-33009225 on Build GRCh38
rs55711954 Chr.1: 33009417-33009417 on Build GRCh38
rs972358412 Chr.1: 33009400-33009400 on Build GRCh38
rs1015575620 Chr.1: 33009207-33009207 on Build GRCh38
rs757950461 Chr.1: 33009285-33009285 on Build GRCh38
rs778698293 Chr.1: 33009151-33009151 on Build GRCh38
rs34574865 Chr.1: 33009348-33009349 on Build GRCh38
rs759518797 Chr.1: 33009276-33009276 on Build GRCh38
rs569532390 Chr.1: 33009360-33009360 on Build GRCh38
rs1058771 Chr.1: 33009096-33009096 on Build GRCh38
rs116880635 Chr.1: 33009385-33009385 on Build GRCh38
rs189506091 Chr.1: 33009001-33009001 on Build GRCh38
rs11509 Chr.1: 33009027-33009027 on Build GRCh38
rs767828057 Chr.1: 33009356-33009356 on Build GRCh38
rs971134112 Chr.1: 33009407-33009407 on Build GRCh38
rs770999463 Chr.1: 33009207-33009207 on Build GRCh38
rs888511278 Chr.1: 33009164-33009164 on Build GRCh38
rs778075147 Chr.1: 33009447-33009447 on Build GRCh38
rs115654177 Chr.1: 33009178-33009178 on Build GRCh38
rs901925288 Chr.1: 33009180-33009180 on Build GRCh38
rs930441921 Chr.1: 33009135-33009135 on Build GRCh38
rs548205481 Chr.1: 33009113-33009113 on Build GRCh38
rs776576885 Chr.1: 33009064-33009064 on Build GRCh38
rs749995037 Chr.1: 33009188-33009188 on Build GRCh38
rs900343296 Chr.1: 33009102-33009102 on Build GRCh38
rs1006401457 Chr.1: 33009341-33009341 on Build GRCh38
rs779669632 Chr.1: 33009296-33009296 on Build GRCh38
rs760562051 Chr.1: 33009023-33009024 on Build GRCh38
rs757282655 Chr.1: 33008990-33008990 on Build GRCh38
rs1038962620 Chr.1: 33009090-33009090 on Build GRCh38
rs181009703 Chr.1: 33009049-33009049 on Build GRCh38
rs916953265 Chr.1: 33009405-33009405 on Build GRCh38
rs1033190259 Chr.1: 33009369-33009369 on Build GRCh38
rs78186437 Chr.1: 33009036-33009036 on Build GRCh38
rs1014499049 Chr.1: 33009277-33009277 on Build GRCh38
rs533711871 Chr.1: 33009370-33009370 on Build GRCh38
rs996252960 Chr.1: 33009254-33009254 on Build GRCh38
rs1010152219 Chr.1: 33009169-33009169 on Build GRCh38
rs905733146 Chr.1: 33009048-33009048 on Build GRCh38
rs891726308 Chr.1: 33009097-33009097 on Build GRCh38

View Full Product Details