Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202778438-202778835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
UniGene
Hs.516629
Gene Chromosome Location
Chr.2: 202773150-202871985 on Build GRCh38
Transcripts
NM_001288622.1 NM_001288623.1 NM_138468.5

SNP Detail

SNP ID Chromosome Location
rs951771872 Chr.2: 202778784-202778784 on Build GRCh38
rs775387079 Chr.2: 202778802-202778803 on Build GRCh38
rs926554028 Chr.2: 202778554-202778554 on Build GRCh38
rs78051637 Chr.2: 202778716-202778716 on Build GRCh38
rs867463072 Chr.2: 202778638-202778638 on Build GRCh38
rs954282908 Chr.2: 202778622-202778622 on Build GRCh38
rs1048941172 Chr.2: 202778554-202778558 on Build GRCh38
rs148892370 Chr.2: 202778478-202778478 on Build GRCh38
rs971950473 Chr.2: 202778653-202778653 on Build GRCh38
rs72932717 Chr.2: 202778587-202778587 on Build GRCh38
rs1036882718 Chr.2: 202778560-202778560 on Build GRCh38
rs115831835 Chr.2: 202778616-202778616 on Build GRCh38
rs903829952 Chr.2: 202778592-202778592 on Build GRCh38
rs959145987 Chr.2: 202778593-202778593 on Build GRCh38
rs748764072 Chr.2: 202778640-202778640 on Build GRCh38
rs773815412 Chr.2: 202778756-202778756 on Build GRCh38
rs918902478 Chr.2: 202778508-202778508 on Build GRCh38
rs898177232 Chr.2: 202778564-202778564 on Build GRCh38
rs919030444 Chr.2: 202778719-202778719 on Build GRCh38
rs555974270 Chr.2: 202778646-202778646 on Build GRCh38
rs1020191926 Chr.2: 202778690-202778690 on Build GRCh38
rs751385953 Chr.2: 202778462-202778462 on Build GRCh38
rs114466346 Chr.2: 202778717-202778717 on Build GRCh38
rs748065269 Chr.2: 202778573-202778573 on Build GRCh38
rs1010053111 Chr.2: 202778576-202778576 on Build GRCh38
rs777028717 Chr.2: 202778553-202778554 on Build GRCh38
rs558556203 Chr.2: 202778565-202778565 on Build GRCh38
rs1000882504 Chr.2: 202778605-202778605 on Build GRCh38
rs60785646 Chr.2: 202778559-202778559 on Build GRCh38
rs577630668 Chr.2: 202778526-202778526 on Build GRCh38
rs757674171 Chr.2: 202778563-202778563 on Build GRCh38
rs369062785 Chr.2: 202778618-202778618 on Build GRCh38
rs567123558 Chr.2: 202778497-202778497 on Build GRCh38

View Full Product Details