Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46660235-46660693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5548185 c.2212-10delT p.? Chr.16: 46660661-46660661 on Build GRCh38
3794854 c.2363G>A p.G788E Chr.16: 46660500-46660500 on Build GRCh38
6820570 c.2315A>G p.N772S Chr.16: 46660548-46660548 on Build GRCh38
2920745 c.2279G>T p.S760I Chr.16: 46660584-46660584 on Build GRCh38
417001 c.2358C>T p.S786S Chr.16: 46660505-46660505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765392455 Chr.16: 46660429-46660429 on Build GRCh38
rs752414695 Chr.16: 46660547-46660547 on Build GRCh38
rs970135927 Chr.16: 46660426-46660426 on Build GRCh38
rs1048064410 Chr.16: 46660522-46660522 on Build GRCh38
rs192419029 Chr.16: 46660543-46660543 on Build GRCh38
rs747505018 Chr.16: 46660629-46660629 on Build GRCh38
rs776023975 Chr.16: 46660481-46660481 on Build GRCh38
rs773175370 Chr.16: 46660516-46660516 on Build GRCh38
rs948546458 Chr.16: 46660506-46660506 on Build GRCh38
rs185420432 Chr.16: 46660416-46660416 on Build GRCh38
rs762118625 Chr.16: 46660505-46660505 on Build GRCh38
rs751335821 Chr.16: 46660445-46660445 on Build GRCh38
rs1031732732 Chr.16: 46660400-46660400 on Build GRCh38
rs763043556 Chr.16: 46660519-46660519 on Build GRCh38
rs893726279 Chr.16: 46660631-46660631 on Build GRCh38
rs774726845 Chr.16: 46660324-46660324 on Build GRCh38
rs76693948 Chr.16: 46660600-46660600 on Build GRCh38
rs563219537 Chr.16: 46660305-46660305 on Build GRCh38
rs755770851 Chr.16: 46660552-46660552 on Build GRCh38
rs370966099 Chr.16: 46660576-46660576 on Build GRCh38
rs530305374 Chr.16: 46660395-46660395 on Build GRCh38
rs769206002 Chr.16: 46660660-46660660 on Build GRCh38
rs999417037 Chr.16: 46660287-46660287 on Build GRCh38
rs777050595 Chr.16: 46660504-46660504 on Build GRCh38
rs779980787 Chr.16: 46660438-46660438 on Build GRCh38
rs527800239 Chr.16: 46660418-46660418 on Build GRCh38
rs184968916 Chr.16: 46660502-46660502 on Build GRCh38
rs1011657508 Chr.16: 46660398-46660398 on Build GRCh38
rs910938026 Chr.16: 46660524-46660524 on Build GRCh38
rs112747048 Chr.16: 46660399-46660399 on Build GRCh38
rs80303950 Chr.16: 46660599-46660599 on Build GRCh38
rs931014095 Chr.16: 46660508-46660508 on Build GRCh38
rs548502981 Chr.16: 46660415-46660415 on Build GRCh38
rs747417009 Chr.16: 46660487-46660487 on Build GRCh38
rs183923278 Chr.16: 46660465-46660465 on Build GRCh38
rs1022087928 Chr.16: 46660409-46660409 on Build GRCh38
rs547805228 Chr.16: 46660641-46660641 on Build GRCh38
rs780867284 Chr.16: 46660454-46660454 on Build GRCh38
rs188654478 Chr.16: 46660464-46660464 on Build GRCh38
rs758270070 Chr.16: 46660436-46660436 on Build GRCh38
rs11550461 Chr.16: 46660293-46660293 on Build GRCh38
rs968423274 Chr.16: 46660392-46660392 on Build GRCh38
rs777427799 Chr.16: 46660557-46660557 on Build GRCh38
rs570492580 Chr.16: 46660468-46660468 on Build GRCh38
rs977203062 Chr.16: 46660393-46660393 on Build GRCh38
rs754730255 Chr.16: 46660451-46660451 on Build GRCh38
rs751488260 Chr.16: 46660531-46660531 on Build GRCh38
rs754820224 Chr.16: 46660538-46660538 on Build GRCh38
rs189948577 Chr.16: 46660461-46660461 on Build GRCh38
rs181292654 Chr.16: 46660517-46660517 on Build GRCh38
rs759445626 Chr.16: 46660290-46660290 on Build GRCh38
rs140543776 Chr.16: 46660622-46660622 on Build GRCh38
rs750436964 Chr.16: 46660431-46660431 on Build GRCh38
rs532641584 Chr.16: 46660530-46660530 on Build GRCh38
rs769119901 Chr.16: 46660499-46660499 on Build GRCh38
rs765487058 Chr.16: 46660509-46660509 on Build GRCh38
rs200195623 Chr.16: 46660470-46660470 on Build GRCh38
rs759782658 Chr.16: 46660325-46660325 on Build GRCh38
rs113746010 Chr.16: 46660594-46660594 on Build GRCh38
rs768025111 Chr.16: 46660525-46660525 on Build GRCh38

View Full Product Details