Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80684360-80684933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL2
Entrez Gene ID
124359
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
PCCP1
UniGene
Hs.373908
Gene Chromosome Location
Chr.16: 80597899-80805043 on Build GRCh38
Transcripts
NM_152342.3 XM_011522866.1 XM_011522867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3402492 c.449A>G p.K150R Chr.16: 80684705-80684705 on Build GRCh38
4772560 c.254C>T p.S85L Chr.16: 80684900-80684900 on Build GRCh38
4895255 c.286C>T p.P96S Chr.16: 80684868-80684868 on Build GRCh38
3370568 c.541T>C p.L181L Chr.16: 80684613-80684613 on Build GRCh38
7179249 c.501G>A p.E167E Chr.16: 80684653-80684653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027674208 Chr.16: 80684775-80684775 on Build GRCh38
rs748321271 Chr.16: 80684897-80684897 on Build GRCh38
rs758179207 Chr.16: 80684609-80684609 on Build GRCh38
rs182620709 Chr.16: 80684545-80684545 on Build GRCh38
rs779344473 Chr.16: 80684518-80684518 on Build GRCh38
rs763182906 Chr.16: 80684719-80684719 on Build GRCh38
rs908827011 Chr.16: 80684389-80684389 on Build GRCh38
rs756433507 Chr.16: 80684736-80684736 on Build GRCh38
rs769293029 Chr.16: 80684606-80684606 on Build GRCh38
rs760360953 Chr.16: 80684711-80684711 on Build GRCh38
rs922231130 Chr.16: 80684847-80684847 on Build GRCh38
rs138924106 Chr.16: 80684552-80684552 on Build GRCh38
rs557233221 Chr.16: 80684563-80684563 on Build GRCh38
rs915694588 Chr.16: 80684647-80684647 on Build GRCh38
rs765324115 Chr.16: 80684643-80684643 on Build GRCh38
rs563881113 Chr.16: 80684385-80684385 on Build GRCh38
rs750840122 Chr.16: 80684732-80684732 on Build GRCh38
rs538917700 Chr.16: 80684879-80684879 on Build GRCh38
rs201777900 Chr.16: 80684822-80684822 on Build GRCh38
rs748118826 Chr.16: 80684691-80684691 on Build GRCh38
rs920049748 Chr.16: 80684427-80684427 on Build GRCh38
rs893033243 Chr.16: 80684700-80684700 on Build GRCh38
rs920183275 Chr.16: 80684615-80684615 on Build GRCh38
rs766737973 Chr.16: 80684832-80684832 on Build GRCh38
rs1023081357 Chr.16: 80684795-80684795 on Build GRCh38
rs145820855 Chr.16: 80684538-80684538 on Build GRCh38
rs757634954 Chr.16: 80684601-80684601 on Build GRCh38
rs959637712 Chr.16: 80684799-80684799 on Build GRCh38
rs144219171 Chr.16: 80684673-80684673 on Build GRCh38
rs370354110 Chr.16: 80684472-80684472 on Build GRCh38
rs780764060 Chr.16: 80684602-80684602 on Build GRCh38
rs778308961 Chr.16: 80684539-80684539 on Build GRCh38
rs761199293 Chr.16: 80684904-80684904 on Build GRCh38
rs373621157 Chr.16: 80684666-80684666 on Build GRCh38
rs751651545 Chr.16: 80684724-80684724 on Build GRCh38
rs758991617 Chr.16: 80684588-80684588 on Build GRCh38
rs952667894 Chr.16: 80684430-80684430 on Build GRCh38
rs140729164 Chr.16: 80684692-80684692 on Build GRCh38
rs377242985 Chr.16: 80684593-80684593 on Build GRCh38
rs147220027 Chr.16: 80684725-80684725 on Build GRCh38
rs928734436 Chr.16: 80684478-80684478 on Build GRCh38
rs377184768 Chr.16: 80684838-80684838 on Build GRCh38
rs761574264 Chr.16: 80684622-80684622 on Build GRCh38
rs772060772 Chr.16: 80684899-80684899 on Build GRCh38
rs775849515 Chr.16: 80684717-80684717 on Build GRCh38
rs768952459 Chr.16: 80684784-80684784 on Build GRCh38
rs974099405 Chr.16: 80684772-80684772 on Build GRCh38
rs561424671 Chr.16: 80684459-80684459 on Build GRCh38
rs753908069 Chr.16: 80684512-80684512 on Build GRCh38
rs145169210 Chr.16: 80684551-80684551 on Build GRCh38
rs768144891 Chr.16: 80684680-80684680 on Build GRCh38
rs562488396 Chr.16: 80684755-80684755 on Build GRCh38
rs746932115 Chr.16: 80684867-80684867 on Build GRCh38
rs368116348 Chr.16: 80684556-80684556 on Build GRCh38
rs150427251 Chr.16: 80684608-80684608 on Build GRCh38
rs149909917 Chr.16: 80684905-80684905 on Build GRCh38
rs539874749 Chr.16: 80684436-80684436 on Build GRCh38
rs200977842 Chr.16: 80684567-80684567 on Build GRCh38
rs745434982 Chr.16: 80684554-80684554 on Build GRCh38
rs778511407 Chr.16: 80684843-80684843 on Build GRCh38
rs773079432 Chr.16: 80684702-80684702 on Build GRCh38
rs200502636 Chr.16: 80684853-80684853 on Build GRCh38
rs751723900 Chr.16: 80684426-80684426 on Build GRCh38
rs767889120 Chr.16: 80684730-80684730 on Build GRCh38
rs202178136 Chr.16: 80684852-80684852 on Build GRCh38
rs370242036 Chr.16: 80684688-80684688 on Build GRCh38
rs149557557 Chr.16: 80684671-80684671 on Build GRCh38
rs772092026 Chr.16: 80684753-80684753 on Build GRCh38
rs139107743 Chr.16: 80684900-80684900 on Build GRCh38
rs1045413907 Chr.16: 80684516-80684516 on Build GRCh38
rs774539541 Chr.16: 80684786-80684786 on Build GRCh38
rs766579362 Chr.16: 80684569-80684569 on Build GRCh38
rs1043342205 Chr.16: 80684497-80684497 on Build GRCh38
rs528976454 Chr.16: 80684460-80684460 on Build GRCh38
rs997114920 Chr.16: 80684418-80684418 on Build GRCh38
rs140264385 Chr.16: 80684429-80684429 on Build GRCh38
rs151165278 Chr.16: 80684831-80684831 on Build GRCh38
rs768623859 Chr.16: 80684611-80684611 on Build GRCh38
rs138401474 Chr.16: 80684571-80684571 on Build GRCh38
rs149647231 Chr.16: 80684572-80684572 on Build GRCh38
rs75914279 Chr.16: 80684722-80684722 on Build GRCh38
rs761768744 Chr.16: 80684804-80684804 on Build GRCh38
rs376037805 Chr.16: 80684495-80684495 on Build GRCh38
rs750885476 Chr.16: 80684813-80684813 on Build GRCh38
rs768140025 Chr.16: 80684808-80684808 on Build GRCh38
rs755084138 Chr.16: 80684513-80684513 on Build GRCh38
rs187745628 Chr.16: 80684656-80684656 on Build GRCh38
rs977037619 Chr.16: 80684844-80684844 on Build GRCh38
rs370582000 Chr.16: 80684632-80684632 on Build GRCh38
rs373304541 Chr.16: 80684605-80684605 on Build GRCh38
rs202190020 Chr.16: 80684508-80684508 on Build GRCh38
rs759063822 Chr.16: 80684487-80684487 on Build GRCh38
rs762910339 Chr.16: 80684562-80684562 on Build GRCh38
rs1016182583 Chr.16: 80684437-80684437 on Build GRCh38
rs972098643 Chr.16: 80684464-80684464 on Build GRCh38
rs940248015 Chr.16: 80684404-80684404 on Build GRCh38
rs144386832 Chr.16: 80684752-80684752 on Build GRCh38
rs779742591 Chr.16: 80684737-80684737 on Build GRCh38
rs550467700 Chr.16: 80684474-80684474 on Build GRCh38
rs765125916 Chr.16: 80684488-80684488 on Build GRCh38
rs77004691 Chr.16: 80684520-80684520 on Build GRCh38
rs112747723 Chr.16: 80684770-80684770 on Build GRCh38
rs1013394913 Chr.16: 80684785-80684785 on Build GRCh38
rs900953571 Chr.16: 80684550-80684550 on Build GRCh38
rs752873125 Chr.16: 80684651-80684651 on Build GRCh38
rs113340497 Chr.16: 80684492-80684492 on Build GRCh38
rs770181158 Chr.16: 80684712-80684712 on Build GRCh38
rs763883018 Chr.16: 80684494-80684494 on Build GRCh38
rs575798444 Chr.16: 80684566-80684566 on Build GRCh38
rs372742877 Chr.16: 80684782-80684782 on Build GRCh38
rs759830866 Chr.16: 80684641-80684641 on Build GRCh38
rs138238685 Chr.16: 80684658-80684658 on Build GRCh38
rs1015557073 Chr.16: 80684762-80684762 on Build GRCh38
rs780304676 Chr.16: 80684502-80684502 on Build GRCh38
rs745637948 Chr.16: 80684880-80684880 on Build GRCh38
rs986776587 Chr.16: 80684576-80684576 on Build GRCh38
rs769040263 Chr.16: 80684883-80684883 on Build GRCh38
rs201069122 Chr.16: 80684846-80684846 on Build GRCh38
rs747417366 Chr.16: 80684600-80684600 on Build GRCh38
rs770835171 Chr.16: 80684766-80684766 on Build GRCh38
rs765359697 Chr.16: 80684584-80684584 on Build GRCh38
rs764763104 Chr.16: 80684653-80684653 on Build GRCh38
rs1055281965 Chr.16: 80684638-80684638 on Build GRCh38
rs1038521227 Chr.16: 80684411-80684411 on Build GRCh38
rs778339782 Chr.16: 80684690-80684690 on Build GRCh38
rs370784290 Chr.16: 80684740-80684740 on Build GRCh38
rs201425276 Chr.16: 80684670-80684670 on Build GRCh38
rs754556263 Chr.16: 80684743-80684743 on Build GRCh38
rs774270019 Chr.16: 80684614-80684614 on Build GRCh38
rs199772000 Chr.16: 80684586-80684586 on Build GRCh38
rs755893807 Chr.16: 80684726-80684726 on Build GRCh38
rs373182913 Chr.16: 80684889-80684889 on Build GRCh38
rs900990135 Chr.16: 80684407-80684407 on Build GRCh38
rs1036863414 Chr.16: 80684902-80684902 on Build GRCh38
rs998579239 Chr.16: 80684384-80684384 on Build GRCh38
rs924856145 Chr.16: 80684819-80684819 on Build GRCh38
rs756356016 Chr.16: 80684499-80684499 on Build GRCh38
rs746532742 Chr.16: 80684674-80684674 on Build GRCh38
rs372394862 Chr.16: 80684558-80684558 on Build GRCh38
rs757192320 Chr.16: 80684872-80684872 on Build GRCh38
rs752287768 Chr.16: 80684849-80684849 on Build GRCh38
rs754159227 Chr.16: 80684837-80684837 on Build GRCh38
rs62049936 Chr.16: 80684393-80684393 on Build GRCh38
rs369415596 Chr.16: 80684756-80684756 on Build GRCh38
rs376407812 Chr.16: 80684903-80684903 on Build GRCh38

View Full Product Details