Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155923966-155924564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
UniGene
Hs.75262
Gene Chromosome Location
Chr.4: 155924118-155953896 on Build GRCh38
Transcripts
NM_001334.2

SNP Detail

SNP ID Chromosome Location
rs1021099478 Chr.4: 155924522-155924522 on Build GRCh38
rs535288634 Chr.4: 155924116-155924116 on Build GRCh38
rs946439470 Chr.4: 155924526-155924526 on Build GRCh38
rs749948674 Chr.4: 155924079-155924079 on Build GRCh38
rs901428402 Chr.4: 155924141-155924141 on Build GRCh38
rs573239779 Chr.4: 155924167-155924167 on Build GRCh38
rs527418317 Chr.4: 155924537-155924537 on Build GRCh38
rs1025607115 Chr.4: 155924062-155924062 on Build GRCh38
rs997078143 Chr.4: 155924148-155924148 on Build GRCh38
rs982308044 Chr.4: 155924218-155924218 on Build GRCh38
rs991449873 Chr.4: 155924070-155924070 on Build GRCh38
rs922737282 Chr.4: 155924493-155924493 on Build GRCh38
rs184146663 Chr.4: 155924101-155924101 on Build GRCh38
rs549624445 Chr.4: 155924023-155924023 on Build GRCh38
rs777855091 Chr.4: 155924201-155924201 on Build GRCh38
rs964010468 Chr.4: 155924094-155924094 on Build GRCh38
rs1019508895 Chr.4: 155924327-155924327 on Build GRCh38
rs1037823683 Chr.4: 155924261-155924261 on Build GRCh38
rs563175144 Chr.4: 155924416-155924416 on Build GRCh38
rs575019301 Chr.4: 155924436-155924436 on Build GRCh38
rs755959464 Chr.4: 155924282-155924282 on Build GRCh38
rs952765472 Chr.4: 155924398-155924398 on Build GRCh38
rs1038154741 Chr.4: 155924015-155924015 on Build GRCh38
rs149113091 Chr.4: 155924262-155924263 on Build GRCh38
rs1010972503 Chr.4: 155924252-155924252 on Build GRCh38
rs1041200768 Chr.4: 155924135-155924135 on Build GRCh38
rs940049014 Chr.4: 155924474-155924474 on Build GRCh38
rs893503788 Chr.4: 155924242-155924242 on Build GRCh38
rs879528182 Chr.4: 155924113-155924113 on Build GRCh38
rs369308908 Chr.4: 155924223-155924223 on Build GRCh38
rs150512126 Chr.4: 155924404-155924404 on Build GRCh38
rs1032113997 Chr.4: 155924423-155924423 on Build GRCh38
rs78242193 Chr.4: 155923992-155923992 on Build GRCh38
rs990722577 Chr.4: 155924353-155924353 on Build GRCh38
rs1032497453 Chr.4: 155924114-155924114 on Build GRCh38
rs754844011 Chr.4: 155924183-155924183 on Build GRCh38
rs559011189 Chr.4: 155924255-155924255 on Build GRCh38
rs1046918712 Chr.4: 155924285-155924285 on Build GRCh38
rs560222709 Chr.4: 155924524-155924524 on Build GRCh38
rs752554842 Chr.4: 155924088-155924088 on Build GRCh38
rs898335585 Chr.4: 155924017-155924017 on Build GRCh38
rs555085063 Chr.4: 155924152-155924152 on Build GRCh38
rs937977945 Chr.4: 155924224-155924224 on Build GRCh38
rs1027972308 Chr.4: 155924373-155924373 on Build GRCh38
rs886916820 Chr.4: 155924286-155924286 on Build GRCh38
rs982340569 Chr.4: 155924528-155924528 on Build GRCh38
rs762809737 Chr.4: 155924080-155924080 on Build GRCh38
rs187325681 Chr.4: 155924284-155924284 on Build GRCh38
rs908633620 Chr.4: 155924422-155924422 on Build GRCh38
rs754147646 Chr.4: 155924304-155924304 on Build GRCh38
rs970832992 Chr.4: 155924527-155924527 on Build GRCh38
rs766332979 Chr.4: 155924081-155924081 on Build GRCh38
rs17033795 Chr.4: 155924488-155924488 on Build GRCh38
rs764782343 Chr.4: 155924074-155924074 on Build GRCh38
rs894047980 Chr.4: 155924525-155924526 on Build GRCh38
rs899531021 Chr.4: 155924348-155924348 on Build GRCh38
rs899243745 Chr.4: 155924274-155924274 on Build GRCh38
rs932795592 Chr.4: 155924505-155924505 on Build GRCh38
rs1008149993 Chr.4: 155924300-155924300 on Build GRCh38
rs914182214 Chr.4: 155924194-155924194 on Build GRCh38
rs879379129 Chr.4: 155924461-155924461 on Build GRCh38

View Full Product Details