Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66641533-66642112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

SNP Detail

SNP ID Chromosome Location
rs965300125 Chr.7: 66641852-66641852 on Build GRCh38
rs1010117463 Chr.7: 66641859-66641859 on Build GRCh38
rs566006114 Chr.7: 66641564-66641564 on Build GRCh38
rs767540235 Chr.7: 66641800-66641801 on Build GRCh38
rs974603287 Chr.7: 66641955-66641955 on Build GRCh38
rs926998002 Chr.7: 66641559-66641559 on Build GRCh38
rs971321154 Chr.7: 66641850-66641850 on Build GRCh38
rs1048465447 Chr.7: 66641746-66641746 on Build GRCh38
rs188529052 Chr.7: 66641762-66641762 on Build GRCh38
rs1267818 Chr.7: 66642037-66642037 on Build GRCh38
rs1051313975 Chr.7: 66641600-66641600 on Build GRCh38
rs1037624066 Chr.7: 66641683-66641683 on Build GRCh38
rs545306248 Chr.7: 66641929-66641929 on Build GRCh38
rs956087608 Chr.7: 66641737-66641737 on Build GRCh38
rs889828618 Chr.7: 66641804-66641804 on Build GRCh38
rs529429431 Chr.7: 66641558-66641558 on Build GRCh38
rs775131916 Chr.7: 66641917-66641917 on Build GRCh38
rs554416139 Chr.7: 66641562-66641562 on Build GRCh38
rs553224420 Chr.7: 66641890-66641890 on Build GRCh38
rs999927500 Chr.7: 66642081-66642081 on Build GRCh38
rs769243904 Chr.7: 66641842-66641842 on Build GRCh38
rs573138370 Chr.7: 66641924-66641924 on Build GRCh38
rs149443387 Chr.7: 66642084-66642084 on Build GRCh38
rs556769827 Chr.7: 66641805-66641805 on Build GRCh38
rs4565357 Chr.7: 66641894-66641894 on Build GRCh38
rs767325788 Chr.7: 66641936-66641936 on Build GRCh38
rs927029021 Chr.7: 66641973-66641973 on Build GRCh38
rs886062423 Chr.7: 66641554-66641554 on Build GRCh38
rs890109261 Chr.7: 66641606-66641606 on Build GRCh38
rs573688509 Chr.7: 66641885-66641885 on Build GRCh38
rs930241499 Chr.7: 66641578-66641578 on Build GRCh38
rs558064744 Chr.7: 66641630-66641630 on Build GRCh38
rs533167133 Chr.7: 66642054-66642054 on Build GRCh38
rs1037252162 Chr.7: 66642046-66642046 on Build GRCh38
rs763411190 Chr.7: 66642080-66642080 on Build GRCh38
rs181035235 Chr.7: 66641940-66641940 on Build GRCh38
rs992467137 Chr.7: 66642016-66642016 on Build GRCh38
rs574218835 Chr.7: 66641643-66641643 on Build GRCh38
rs927287789 Chr.7: 66641922-66641922 on Build GRCh38
rs891356953 Chr.7: 66641983-66641983 on Build GRCh38
rs770080043 Chr.7: 66642033-66642033 on Build GRCh38
rs949634464 Chr.7: 66641590-66641590 on Build GRCh38
rs900204057 Chr.7: 66642038-66642038 on Build GRCh38
rs896729875 Chr.7: 66641567-66641567 on Build GRCh38
rs978502767 Chr.7: 66641554-66641554 on Build GRCh38
rs886062425 Chr.7: 66642038-66642038 on Build GRCh38
rs968575664 Chr.7: 66641902-66641902 on Build GRCh38
rs1019569546 Chr.7: 66641848-66641848 on Build GRCh38
rs958395650 Chr.7: 66641978-66641978 on Build GRCh38
rs745356728 Chr.7: 66641731-66641731 on Build GRCh38
rs1008490972 Chr.7: 66641650-66641650 on Build GRCh38
rs886062424 Chr.7: 66641978-66641978 on Build GRCh38
rs1860469 Chr.7: 66641888-66641888 on Build GRCh38
rs1017244646 Chr.7: 66641682-66641682 on Build GRCh38
rs571134300 Chr.7: 66641701-66641701 on Build GRCh38
rs530330836 Chr.7: 66641566-66641566 on Build GRCh38

View Full Product Details