Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86573976-86574557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

SNP Detail

SNP ID Chromosome Location
rs914165933 Chr.8: 86574328-86574328 on Build GRCh38
rs1050171414 Chr.8: 86574152-86574152 on Build GRCh38
rs917913957 Chr.8: 86574206-86574206 on Build GRCh38
rs796676165 Chr.8: 86574150-86574150 on Build GRCh38
rs982329960 Chr.8: 86574099-86574099 on Build GRCh38
rs560958599 Chr.8: 86574066-86574066 on Build GRCh38
rs1059337 Chr.8: 86574424-86574424 on Build GRCh38
rs970889903 Chr.8: 86574071-86574071 on Build GRCh38
rs898215471 Chr.8: 86574459-86574459 on Build GRCh38
rs770160518 Chr.8: 86574207-86574207 on Build GRCh38
rs199880215 Chr.8: 86573998-86573998 on Build GRCh38
rs1023735228 Chr.8: 86574272-86574272 on Build GRCh38
rs777986938 Chr.8: 86574148-86574148 on Build GRCh38
rs936874713 Chr.8: 86574267-86574267 on Build GRCh38
rs908139316 Chr.8: 86574106-86574106 on Build GRCh38
rs759866106 Chr.8: 86574187-86574187 on Build GRCh38
rs1000146601 Chr.8: 86574201-86574201 on Build GRCh38
rs992178757 Chr.8: 86574204-86574204 on Build GRCh38
rs886063154 Chr.8: 86574103-86574103 on Build GRCh38
rs1052717488 Chr.8: 86574055-86574055 on Build GRCh38
rs778952875 Chr.8: 86574197-86574197 on Build GRCh38
rs537374436 Chr.8: 86574344-86574344 on Build GRCh38
rs1041519693 Chr.8: 86574496-86574496 on Build GRCh38
rs920251208 Chr.8: 86574346-86574346 on Build GRCh38
rs868592775 Chr.8: 86574030-86574030 on Build GRCh38
rs538697794 Chr.8: 86574326-86574326 on Build GRCh38
rs970858872 Chr.8: 86574317-86574317 on Build GRCh38
rs780284102 Chr.8: 86574445-86574445 on Build GRCh38
rs1013504894 Chr.8: 86574520-86574520 on Build GRCh38
rs146323195 Chr.8: 86574391-86574391 on Build GRCh38
rs142508439 Chr.8: 86574150-86574151 on Build GRCh38
rs944484004 Chr.8: 86574478-86574478 on Build GRCh38
rs1059338 Chr.8: 86574236-86574236 on Build GRCh38
rs28471019 Chr.8: 86574166-86574166 on Build GRCh38
rs777073827 Chr.8: 86574174-86574174 on Build GRCh38
rs951333046 Chr.8: 86574167-86574167 on Build GRCh38
rs192543896 Chr.8: 86574345-86574345 on Build GRCh38
rs17683284 Chr.8: 86574501-86574501 on Build GRCh38
rs1041587682 Chr.8: 86574182-86574182 on Build GRCh38
rs1059336 Chr.8: 86574426-86574426 on Build GRCh38
rs545070786 Chr.8: 86574529-86574529 on Build GRCh38
rs1012299376 Chr.8: 86574259-86574259 on Build GRCh38
rs568808064 Chr.8: 86574334-86574334 on Build GRCh38
rs886063155 Chr.8: 86574436-86574436 on Build GRCh38
rs757397657 Chr.8: 86574170-86574170 on Build GRCh38
rs1037753526 Chr.8: 86574133-86574133 on Build GRCh38
rs574196021 Chr.8: 86574387-86574387 on Build GRCh38
rs942480886 Chr.8: 86574355-86574355 on Build GRCh38
rs990192 Chr.8: 86574165-86574165 on Build GRCh38
rs980840656 Chr.8: 86574239-86574239 on Build GRCh38
rs931715032 Chr.8: 86574151-86574151 on Build GRCh38
rs563975295 Chr.8: 86574356-86574356 on Build GRCh38
rs1000519937 Chr.8: 86574526-86574526 on Build GRCh38
rs1009908794 Chr.8: 86574103-86574103 on Build GRCh38
rs372688369 Chr.8: 86574433-86574433 on Build GRCh38
rs916748731 Chr.8: 86574070-86574070 on Build GRCh38
rs936949954 Chr.8: 86574043-86574043 on Build GRCh38
rs958105324 Chr.8: 86574238-86574238 on Build GRCh38
rs529889744 Chr.8: 86574123-86574123 on Build GRCh38
rs911655351 Chr.8: 86574474-86574474 on Build GRCh38
rs149173480 Chr.8: 86574327-86574327 on Build GRCh38
rs71510444 Chr.8: 86574165-86574166 on Build GRCh38
rs1050100790 Chr.8: 86574457-86574457 on Build GRCh38

View Full Product Details