Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47681809-47682223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
UniGene
Hs.147377
Gene Chromosome Location
Chr.11: 47659168-47715397 on Build GRCh38
Transcripts
NM_024783.3 XM_005253138.4 XM_005253139.4 XM_005253140.4 XM_005253141.4 XM_006718327.3 XM_006718328.3 XM_011520372.2 XM_011520373.2 XM_011520374.2 XM_011520375.2 XM_011520376.2 XM_011520377.2 XM_011520378.2 XM_011520379.2 XM_011520381.2 XM_017018331.1 XM_017018332.1 XM_017018333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686652 c.1915+2T>G p.? Chr.11: 47681967-47681967 on Build GRCh38
687949 c.1865G>T p.G622V Chr.11: 47682019-47682019 on Build GRCh38
1981909 c.1875C>A p.N625K Chr.11: 47682009-47682009 on Build GRCh38
5886701 c.1895C>T p.T632I Chr.11: 47681989-47681989 on Build GRCh38
5360406 c.1824delA p.K608fs*17 Chr.11: 47682060-47682060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976828376 Chr.11: 47682043-47682043 on Build GRCh38
rs761295692 Chr.11: 47682135-47682135 on Build GRCh38
rs924663116 Chr.11: 47682182-47682182 on Build GRCh38
rs757228673 Chr.11: 47681989-47681989 on Build GRCh38
rs778795890 Chr.11: 47681991-47681991 on Build GRCh38
rs540679723 Chr.11: 47682201-47682201 on Build GRCh38
rs1023437001 Chr.11: 47681872-47681872 on Build GRCh38
rs905686936 Chr.11: 47681852-47681852 on Build GRCh38
rs377200932 Chr.11: 47681959-47681959 on Build GRCh38
rs1003314013 Chr.11: 47681914-47681914 on Build GRCh38
rs763548477 Chr.11: 47681974-47681974 on Build GRCh38
rs146693632 Chr.11: 47681967-47681967 on Build GRCh38
rs1032910330 Chr.11: 47681886-47681886 on Build GRCh38
rs767268698 Chr.11: 47681966-47681966 on Build GRCh38
rs954226188 Chr.11: 47682198-47682198 on Build GRCh38
rs773867938 Chr.11: 47681928-47681928 on Build GRCh38
rs763504221 Chr.11: 47681933-47681933 on Build GRCh38
rs776170923 Chr.11: 47682130-47682130 on Build GRCh38
rs745802329 Chr.11: 47681926-47681926 on Build GRCh38
rs964360244 Chr.11: 47682040-47682040 on Build GRCh38
rs140305206 Chr.11: 47682025-47682025 on Build GRCh38
rs777960187 Chr.11: 47682046-47682046 on Build GRCh38
rs866138159 Chr.11: 47682133-47682133 on Build GRCh38
rs749347656 Chr.11: 47682052-47682052 on Build GRCh38
rs150240657 Chr.11: 47682026-47682026 on Build GRCh38
rs775234437 Chr.11: 47682099-47682099 on Build GRCh38
rs750285686 Chr.11: 47681992-47681992 on Build GRCh38
rs983749613 Chr.11: 47682002-47682002 on Build GRCh38
rs893766390 Chr.11: 47681844-47681844 on Build GRCh38
rs868667107 Chr.11: 47682047-47682047 on Build GRCh38
rs772269136 Chr.11: 47682096-47682096 on Build GRCh38
rs370788989 Chr.11: 47682041-47682041 on Build GRCh38
rs760207684 Chr.11: 47682102-47682102 on Build GRCh38
rs1053624302 Chr.11: 47681834-47681834 on Build GRCh38
rs558577344 Chr.11: 47681981-47681981 on Build GRCh38
rs758120834 Chr.11: 47682008-47682008 on Build GRCh38
rs746250579 Chr.11: 47682084-47682084 on Build GRCh38
rs141369982 Chr.11: 47681969-47681969 on Build GRCh38
rs1041796508 Chr.11: 47682113-47682113 on Build GRCh38
rs778714918 Chr.11: 47681849-47681849 on Build GRCh38
rs554998224 Chr.11: 47682191-47682191 on Build GRCh38
rs763709502 Chr.11: 47682107-47682107 on Build GRCh38
rs372919703 Chr.11: 47681841-47681841 on Build GRCh38
rs771452694 Chr.11: 47681938-47681938 on Build GRCh38
rs774994505 Chr.11: 47682079-47682079 on Build GRCh38
rs367772382 Chr.11: 47682196-47682196 on Build GRCh38
rs1000464813 Chr.11: 47681881-47681881 on Build GRCh38
rs771624429 Chr.11: 47682053-47682053 on Build GRCh38
rs772010154 Chr.11: 47681946-47681946 on Build GRCh38
rs756992833 Chr.11: 47681845-47681845 on Build GRCh38
rs955917540 Chr.11: 47681909-47681909 on Build GRCh38

View Full Product Details