Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78216249-78216782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB2
Entrez Gene ID
9846
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
UniGene
Hs.429434
Gene Chromosome Location
Chr.11: 78215290-78417822 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
UniGene
Hs.531249
Gene Chromosome Location
Chr.11: 78188812-78233651 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

SNP Detail

SNP ID Chromosome Location
rs950637996 Chr.11: 78216703-78216703 on Build GRCh38
rs535784008 Chr.11: 78216670-78216670 on Build GRCh38
rs756554948 Chr.11: 78216311-78216311 on Build GRCh38
rs1029755862 Chr.11: 78216455-78216455 on Build GRCh38
rs975364921 Chr.11: 78216464-78216464 on Build GRCh38
rs763120300 Chr.11: 78216668-78216668 on Build GRCh38
rs907940980 Chr.11: 78216299-78216299 on Build GRCh38
rs535719100 Chr.11: 78216378-78216378 on Build GRCh38
rs971481267 Chr.11: 78216677-78216677 on Build GRCh38
rs973765005 Chr.11: 78216490-78216490 on Build GRCh38
rs991022720 Chr.11: 78216742-78216742 on Build GRCh38
rs113346695 Chr.11: 78216286-78216286 on Build GRCh38
rs1010067225 Chr.11: 78216384-78216384 on Build GRCh38
rs775408357 Chr.11: 78216511-78216511 on Build GRCh38
rs542100466 Chr.11: 78216557-78216557 on Build GRCh38
rs190949951 Chr.11: 78216467-78216467 on Build GRCh38
rs371998752 Chr.11: 78216376-78216377 on Build GRCh38
rs902786899 Chr.11: 78216574-78216574 on Build GRCh38
rs140676461 Chr.11: 78216329-78216329 on Build GRCh38
rs1021205421 Chr.11: 78216411-78216411 on Build GRCh38
rs914485566 Chr.11: 78216763-78216763 on Build GRCh38
rs1054305382 Chr.11: 78216345-78216345 on Build GRCh38
rs1046221431 Chr.11: 78216347-78216347 on Build GRCh38
rs941964443 Chr.11: 78216304-78216304 on Build GRCh38
rs1007892100 Chr.11: 78216644-78216644 on Build GRCh38
rs905799244 Chr.11: 78216328-78216328 on Build GRCh38
rs909860220 Chr.11: 78216297-78216297 on Build GRCh38
rs576289901 Chr.11: 78216377-78216377 on Build GRCh38
rs778144807 Chr.11: 78216318-78216318 on Build GRCh38
rs962733262 Chr.11: 78216472-78216472 on Build GRCh38
rs929171458 Chr.11: 78216507-78216507 on Build GRCh38
rs955109675 Chr.11: 78216427-78216427 on Build GRCh38
rs1029808217 Chr.11: 78216621-78216622 on Build GRCh38
rs191425629 Chr.11: 78216705-78216705 on Build GRCh38
rs1020474002 Chr.11: 78216425-78216425 on Build GRCh38
rs570535764 Chr.11: 78216618-78216618 on Build GRCh38
rs927357915 Chr.11: 78216718-78216718 on Build GRCh38
rs772056387 Chr.11: 78216445-78216445 on Build GRCh38
rs75865244 Chr.11: 78216353-78216353 on Build GRCh38
rs762449370 Chr.11: 78216730-78216731 on Build GRCh38
rs777911252 Chr.11: 78216540-78216541 on Build GRCh38
rs890894421 Chr.11: 78216606-78216606 on Build GRCh38
rs555845275 Chr.11: 78216339-78216339 on Build GRCh38
rs555618206 Chr.11: 78216380-78216380 on Build GRCh38
rs531896579 Chr.11: 78216556-78216556 on Build GRCh38
rs990151471 Chr.11: 78216737-78216737 on Build GRCh38
rs963077166 Chr.11: 78216653-78216653 on Build GRCh38
rs566762087 Chr.11: 78216667-78216667 on Build GRCh38
rs776446224 Chr.11: 78216663-78216663 on Build GRCh38
rs188727651 Chr.11: 78216506-78216506 on Build GRCh38
rs964902740 Chr.11: 78216279-78216279 on Build GRCh38
rs2852483 Chr.11: 78216492-78216492 on Build GRCh38
rs937361029 Chr.11: 78216729-78216729 on Build GRCh38
rs768357140 Chr.11: 78216527-78216527 on Build GRCh38
rs964089659 Chr.11: 78216652-78216652 on Build GRCh38
rs561969906 Chr.11: 78216558-78216558 on Build GRCh38
rs933778080 Chr.11: 78216471-78216471 on Build GRCh38
rs112724014 Chr.11: 78216285-78216285 on Build GRCh38
rs117013478 Chr.11: 78216520-78216520 on Build GRCh38
rs998369181 Chr.11: 78216584-78216584 on Build GRCh38
rs983625479 Chr.11: 78216491-78216491 on Build GRCh38
rs995166449 Chr.11: 78216658-78216658 on Build GRCh38
rs144366979 Chr.11: 78216563-78216563 on Build GRCh38
rs183285595 Chr.11: 78216477-78216477 on Build GRCh38
rs139688873 Chr.11: 78216281-78216281 on Build GRCh38
rs1007861179 Chr.11: 78216440-78216440 on Build GRCh38
rs753408377 Chr.11: 78216456-78216456 on Build GRCh38
rs111624673 Chr.11: 78216287-78216287 on Build GRCh38
rs1054818620 Chr.11: 78216528-78216528 on Build GRCh38
rs555679764 Chr.11: 78216678-78216678 on Build GRCh38
rs548431238 Chr.11: 78216280-78216280 on Build GRCh38
rs902725941 Chr.11: 78216433-78216433 on Build GRCh38
rs115057958 Chr.11: 78216457-78216457 on Build GRCh38
rs1048319202 Chr.11: 78216290-78216290 on Build GRCh38
rs201967496 Chr.11: 78216380-78216380 on Build GRCh38
rs547215409 Chr.11: 78216608-78216608 on Build GRCh38
rs532840313 Chr.11: 78216642-78216642 on Build GRCh38
rs548269863 Chr.11: 78216521-78216521 on Build GRCh38
rs549770907 Chr.11: 78216651-78216651 on Build GRCh38
rs766598059 Chr.11: 78216723-78216723 on Build GRCh38
rs927818469 Chr.11: 78216722-78216722 on Build GRCh38
rs927766301 Chr.11: 78216518-78216518 on Build GRCh38
rs937723173 Chr.11: 78216375-78216376 on Build GRCh38
rs1041501954 Chr.11: 78216577-78216577 on Build GRCh38
rs1042412345 Chr.11: 78216569-78216570 on Build GRCh38
rs952337357 Chr.11: 78216470-78216470 on Build GRCh38
rs892991887 Chr.11: 78216365-78216365 on Build GRCh38
rs927246797 Chr.11: 78216522-78216522 on Build GRCh38
rs748919116 Chr.11: 78216323-78216323 on Build GRCh38

View Full Product Details