Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134259306-134259800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD8
Entrez Gene ID
27034
Gene Name
acyl-CoA dehydrogenase family member 8
Gene Aliases
ACAD-8, ARC42
UniGene
Hs.14791
Gene Chromosome Location
Chr.11: 134253532-134265858 on Build GRCh38
Transcripts
NM_014384.2 XM_005271505.3 XM_011542750.2 XM_017017542.1 XM_017017543.1 XM_017017544.1 XM_017017545.1 XM_017017546.1 XM_017017547.1 XM_017017548.1 XM_017017549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269813 c.578G>T p.S193I Chr.11: 134259618-134259618 on Build GRCh38
7353872 c.346A>T p.S116C Chr.11: 134259617-134259617 on Build GRCh38
4794539 c.274G>A p.A92T Chr.11: 134259608-134259608 on Build GRCh38
7353871 c.577A>T p.S193C Chr.11: 134259617-134259617 on Build GRCh38
7353873 c.283A>T p.S95C Chr.11: 134259617-134259617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113750526 Chr.11: 134259545-134259545 on Build GRCh38
rs905586035 Chr.11: 134259357-134259357 on Build GRCh38
rs369445365 Chr.11: 134259656-134259656 on Build GRCh38
rs760347118 Chr.11: 134259779-134259779 on Build GRCh38
rs760295777 Chr.11: 134259663-134259663 on Build GRCh38
rs1037256588 Chr.11: 134259406-134259406 on Build GRCh38
rs779974265 Chr.11: 134259708-134259708 on Build GRCh38
rs533215955 Chr.11: 134259336-134259336 on Build GRCh38
rs768767939 Chr.11: 134259564-134259564 on Build GRCh38
rs374360285 Chr.11: 134259588-134259588 on Build GRCh38
rs908083700 Chr.11: 134259635-134259635 on Build GRCh38
rs1030747079 Chr.11: 134259554-134259554 on Build GRCh38
rs953276903 Chr.11: 134259382-134259382 on Build GRCh38
rs774987397 Chr.11: 134259734-134259734 on Build GRCh38
rs879213183 Chr.11: 134259424-134259424 on Build GRCh38
rs777736629 Chr.11: 134259725-134259725 on Build GRCh38
rs773395894 Chr.11: 134259605-134259605 on Build GRCh38
rs777062847 Chr.11: 134259585-134259585 on Build GRCh38
rs978483459 Chr.11: 134259331-134259331 on Build GRCh38
rs868257353 Chr.11: 134259366-134259366 on Build GRCh38
rs1048436767 Chr.11: 134259491-134259491 on Build GRCh38
rs886043698 Chr.11: 134259645-134259645 on Build GRCh38
rs918549995 Chr.11: 134259439-134259439 on Build GRCh38
rs1020124747 Chr.11: 134259425-134259425 on Build GRCh38
rs374148675 Chr.11: 134259657-134259657 on Build GRCh38
rs927511531 Chr.11: 134259748-134259748 on Build GRCh38
rs750326667 Chr.11: 134259685-134259685 on Build GRCh38
rs749331373 Chr.11: 134259727-134259727 on Build GRCh38
rs775796762 Chr.11: 134259757-134259757 on Build GRCh38
rs954766586 Chr.11: 134259556-134259556 on Build GRCh38
rs968609274 Chr.11: 134259461-134259461 on Build GRCh38
rs267602788 Chr.11: 134259675-134259675 on Build GRCh38
rs999962848 Chr.11: 134259485-134259485 on Build GRCh38
rs1010563191 Chr.11: 134259401-134259401 on Build GRCh38
rs151125742 Chr.11: 134259683-134259683 on Build GRCh38
rs761246201 Chr.11: 134259599-134259599 on Build GRCh38
rs968344615 Chr.11: 134259678-134259678 on Build GRCh38
rs142958309 Chr.11: 134259395-134259395 on Build GRCh38
rs562699173 Chr.11: 134259756-134259756 on Build GRCh38
rs1010118612 Chr.11: 134259565-134259565 on Build GRCh38
rs377263662 Chr.11: 134259614-134259614 on Build GRCh38
rs200275294 Chr.11: 134259655-134259655 on Build GRCh38
rs193038282 Chr.11: 134259553-134259553 on Build GRCh38
rs763629898 Chr.11: 134259673-134259673 on Build GRCh38
rs757158583 Chr.11: 134259682-134259682 on Build GRCh38
rs748422726 Chr.11: 134259710-134259710 on Build GRCh38
rs748369707 Chr.11: 134259586-134259586 on Build GRCh38
rs973537605 Chr.11: 134259665-134259665 on Build GRCh38
rs944284147 Chr.11: 134259402-134259402 on Build GRCh38
rs562943735 Chr.11: 134259400-134259400 on Build GRCh38
rs942139810 Chr.11: 134259329-134259329 on Build GRCh38
rs568277085 Chr.11: 134259350-134259350 on Build GRCh38
rs774591276 Chr.11: 134259624-134259624 on Build GRCh38
rs796865354 Chr.11: 134259616-134259616 on Build GRCh38
rs199544611 Chr.11: 134259753-134259753 on Build GRCh38
rs202028201 Chr.11: 134259761-134259761 on Build GRCh38
rs757552131 Chr.11: 134259550-134259550 on Build GRCh38
rs759877257 Chr.11: 134259647-134259647 on Build GRCh38
rs776214300 Chr.11: 134259590-134259590 on Build GRCh38
rs530467407 Chr.11: 134259513-134259513 on Build GRCh38
rs758328658 Chr.11: 134259692-134259692 on Build GRCh38
rs17849561 Chr.11: 134259669-134259669 on Build GRCh38
rs771419550 Chr.11: 134259618-134259618 on Build GRCh38
rs771345382 Chr.11: 134259730-134259730 on Build GRCh38
rs770101596 Chr.11: 134259719-134259719 on Build GRCh38

View Full Product Details