Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154589545-154590124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400065 c.2635G>C p.E879Q Chr.1: 154589790-154589790 on Build GRCh38
4022839 c.2662G>A p.A888T Chr.1: 154589892-154589892 on Build GRCh38
4790483 c.2742C>T p.A914A Chr.1: 154589812-154589812 on Build GRCh38
4872792 c.2760C>T p.D920D Chr.1: 154589794-154589794 on Build GRCh38
7281270 c.2611G>A p.A871T Chr.1: 154589814-154589814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149012931 Chr.1: 154589920-154589920 on Build GRCh38
rs200910534 Chr.1: 154589730-154589730 on Build GRCh38
rs776679441 Chr.1: 154589709-154589709 on Build GRCh38
rs776928411 Chr.1: 154589749-154589749 on Build GRCh38
rs374897040 Chr.1: 154589711-154589711 on Build GRCh38
rs398122893 Chr.1: 154589817-154589817 on Build GRCh38
rs1015474864 Chr.1: 154589911-154589911 on Build GRCh38
rs762568334 Chr.1: 154589877-154589877 on Build GRCh38
rs534131989 Chr.1: 154589926-154589926 on Build GRCh38
rs113249520 Chr.1: 154589943-154589943 on Build GRCh38
rs145115780 Chr.1: 154589732-154589732 on Build GRCh38
rs1027165880 Chr.1: 154589597-154589597 on Build GRCh38
rs749444156 Chr.1: 154589944-154589944 on Build GRCh38
rs978150708 Chr.1: 154590036-154590036 on Build GRCh38
rs752148228 Chr.1: 154590101-154590101 on Build GRCh38
rs761544164 Chr.1: 154589783-154589783 on Build GRCh38
rs1055028275 Chr.1: 154590051-154590051 on Build GRCh38
rs775500671 Chr.1: 154589832-154589832 on Build GRCh38
rs780635627 Chr.1: 154589797-154589797 on Build GRCh38
rs760529708 Chr.1: 154589775-154589775 on Build GRCh38
rs764889694 Chr.1: 154589784-154589784 on Build GRCh38
rs115805812 Chr.1: 154589737-154589737 on Build GRCh38
rs749923012 Chr.1: 154589788-154589788 on Build GRCh38
rs575429853 Chr.1: 154590100-154590100 on Build GRCh38
rs575538000 Chr.1: 154589655-154589655 on Build GRCh38
rs769985255 Chr.1: 154589814-154589814 on Build GRCh38
rs953964268 Chr.1: 154589990-154589990 on Build GRCh38
rs767361741 Chr.1: 154589720-154589720 on Build GRCh38
rs190881240 Chr.1: 154589751-154589751 on Build GRCh38
rs773272107 Chr.1: 154589764-154589764 on Build GRCh38
rs371802266 Chr.1: 154589733-154589733 on Build GRCh38
rs552601351 Chr.1: 154589949-154589949 on Build GRCh38
rs920319323 Chr.1: 154589932-154589932 on Build GRCh38
rs763826690 Chr.1: 154589728-154589728 on Build GRCh38
rs748254914 Chr.1: 154589750-154589750 on Build GRCh38
rs907698628 Chr.1: 154589626-154589626 on Build GRCh38
rs990982544 Chr.1: 154590061-154590061 on Build GRCh38
rs774279898 Chr.1: 154589713-154589713 on Build GRCh38
rs989910480 Chr.1: 154589996-154589996 on Build GRCh38
rs398122897 Chr.1: 154589810-154589810 on Build GRCh38
rs944933560 Chr.1: 154589712-154589712 on Build GRCh38
rs879097270 Chr.1: 154589690-154589690 on Build GRCh38
rs1015424543 Chr.1: 154589637-154589637 on Build GRCh38
rs754465071 Chr.1: 154589780-154589780 on Build GRCh38
rs755430740 Chr.1: 154589923-154589923 on Build GRCh38
rs765742773 Chr.1: 154589878-154589878 on Build GRCh38
rs752179215 Chr.1: 154589898-154589898 on Build GRCh38
rs747015655 Chr.1: 154589953-154589953 on Build GRCh38
rs777316853 Chr.1: 154589598-154589598 on Build GRCh38
rs1028671184 Chr.1: 154589970-154589970 on Build GRCh38
rs371394611 Chr.1: 154590004-154590004 on Build GRCh38
rs753087385 Chr.1: 154589934-154589934 on Build GRCh38
rs1004073895 Chr.1: 154589600-154589600 on Build GRCh38
rs138707431 Chr.1: 154589833-154589833 on Build GRCh38
rs757942430 Chr.1: 154589794-154589794 on Build GRCh38
rs751955732 Chr.1: 154589801-154589801 on Build GRCh38
rs770937947 Chr.1: 154589828-154589828 on Build GRCh38
rs758870630 Chr.1: 154589587-154589587 on Build GRCh38
rs370898628 Chr.1: 154589867-154589867 on Build GRCh38
rs750048114 Chr.1: 154589876-154589876 on Build GRCh38
rs11545928 Chr.1: 154589785-154589785 on Build GRCh38
rs200989942 Chr.1: 154589776-154589776 on Build GRCh38
rs768855853 Chr.1: 154589746-154589746 on Build GRCh38
rs138927668 Chr.1: 154589872-154589872 on Build GRCh38
rs747979489 Chr.1: 154589972-154589972 on Build GRCh38
rs931634512 Chr.1: 154589666-154589666 on Build GRCh38
rs768446958 Chr.1: 154589956-154589956 on Build GRCh38
rs944377552 Chr.1: 154590013-154590013 on Build GRCh38
rs755823017 Chr.1: 154589725-154589725 on Build GRCh38
rs577926927 Chr.1: 154589723-154589734 on Build GRCh38
rs756417649 Chr.1: 154589936-154589936 on Build GRCh38
rs776592160 Chr.1: 154589961-154589961 on Build GRCh38
rs369415788 Chr.1: 154589773-154589773 on Build GRCh38
rs781409066 Chr.1: 154589811-154589811 on Build GRCh38
rs939528347 Chr.1: 154590053-154590053 on Build GRCh38
rs965320940 Chr.1: 154589653-154589653 on Build GRCh38
rs371843017 Chr.1: 154589721-154589721 on Build GRCh38
rs748430309 Chr.1: 154589813-154589813 on Build GRCh38
rs200312914 Chr.1: 154589790-154589790 on Build GRCh38
rs542445699 Chr.1: 154589678-154589678 on Build GRCh38
rs182183195 Chr.1: 154590050-154590050 on Build GRCh38
rs776268194 Chr.1: 154589841-154589841 on Build GRCh38
rs375311508 Chr.1: 154589742-154589742 on Build GRCh38
rs115161953 Chr.1: 154590019-154590019 on Build GRCh38
rs759505132 Chr.1: 154589766-154589766 on Build GRCh38
rs982028597 Chr.1: 154589603-154589609 on Build GRCh38
rs796402433 Chr.1: 154589676-154589676 on Build GRCh38
rs777928382 Chr.1: 154589815-154589815 on Build GRCh38
rs973151713 Chr.1: 154589639-154589639 on Build GRCh38
rs367861888 Chr.1: 154589719-154589719 on Build GRCh38
rs377696698 Chr.1: 154589977-154589977 on Build GRCh38
rs72999489 Chr.1: 154589710-154589710 on Build GRCh38
rs771106880 Chr.1: 154589945-154589945 on Build GRCh38
rs753592604 Chr.1: 154589781-154589781 on Build GRCh38
rs772025392 Chr.1: 154589772-154589772 on Build GRCh38
rs749389266 Chr.1: 154589827-154589827 on Build GRCh38

View Full Product Details