Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154582322-154582921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

SNP Detail

SNP ID Chromosome Location
rs544606177 Chr.1: 154582511-154582511 on Build GRCh38
rs201027063 Chr.1: 154582491-154582491 on Build GRCh38
rs893912785 Chr.1: 154582877-154582877 on Build GRCh38
rs200489041 Chr.1: 154582469-154582475 on Build GRCh38
rs538988470 Chr.1: 154582869-154582869 on Build GRCh38
rs559413890 Chr.1: 154582429-154582429 on Build GRCh38
rs781205612 Chr.1: 154582765-154582765 on Build GRCh38
rs1009750742 Chr.1: 154582531-154582531 on Build GRCh38
rs879619331 Chr.1: 154582472-154582472 on Build GRCh38
rs943987611 Chr.1: 154582814-154582814 on Build GRCh38
rs1802729 Chr.1: 154582403-154582403 on Build GRCh38
rs928322326 Chr.1: 154582630-154582630 on Build GRCh38
rs546744933 Chr.1: 154582714-154582714 on Build GRCh38
rs931042981 Chr.1: 154582774-154582774 on Build GRCh38
rs913386562 Chr.1: 154582657-154582657 on Build GRCh38
rs1043520108 Chr.1: 154582749-154582749 on Build GRCh38
rs751348427 Chr.1: 154582576-154582576 on Build GRCh38
rs987619376 Chr.1: 154582634-154582634 on Build GRCh38
rs373226106 Chr.1: 154582897-154582897 on Build GRCh38
rs113937928 Chr.1: 154582382-154582382 on Build GRCh38
rs569555134 Chr.1: 154582895-154582895 on Build GRCh38
rs367964862 Chr.1: 154582489-154582489 on Build GRCh38
rs949281548 Chr.1: 154582805-154582805 on Build GRCh38
rs140535856 Chr.1: 154582607-154582607 on Build GRCh38
rs1053757519 Chr.1: 154582863-154582863 on Build GRCh38
rs1021097695 Chr.1: 154582571-154582571 on Build GRCh38
rs113734083 Chr.1: 154582605-154582605 on Build GRCh38
rs757149237 Chr.1: 154582703-154582703 on Build GRCh38
rs1040965111 Chr.1: 154582821-154582821 on Build GRCh38
rs895029420 Chr.1: 154582803-154582804 on Build GRCh38
rs996857778 Chr.1: 154582847-154582847 on Build GRCh38
rs991083622 Chr.1: 154582713-154582713 on Build GRCh38
rs1046344874 Chr.1: 154582832-154582832 on Build GRCh38
rs189183388 Chr.1: 154582591-154582591 on Build GRCh38
rs1055459466 Chr.1: 154582387-154582387 on Build GRCh38
rs1005032670 Chr.1: 154582551-154582551 on Build GRCh38
rs1004743346 Chr.1: 154582869-154582869 on Build GRCh38
rs899827266 Chr.1: 154582510-154582510 on Build GRCh38
rs530757718 Chr.1: 154582530-154582530 on Build GRCh38
rs982371041 Chr.1: 154582617-154582617 on Build GRCh38
rs904512754 Chr.1: 154582499-154582499 on Build GRCh38
rs1032429944 Chr.1: 154582516-154582516 on Build GRCh38
rs946268074 Chr.1: 154582695-154582695 on Build GRCh38
rs557438551 Chr.1: 154582879-154582879 on Build GRCh38
rs11545926 Chr.1: 154582454-154582454 on Build GRCh38
rs571724294 Chr.1: 154582773-154582773 on Build GRCh38
rs899711388 Chr.1: 154582825-154582825 on Build GRCh38
rs954968298 Chr.1: 154582616-154582616 on Build GRCh38
rs147198769 Chr.1: 154582515-154582515 on Build GRCh38
rs887044899 Chr.1: 154582460-154582460 on Build GRCh38
rs922161967 Chr.1: 154582615-154582615 on Build GRCh38
rs200952110 Chr.1: 154582475-154582475 on Build GRCh38
rs113414804 Chr.1: 154582475-154582476 on Build GRCh38
rs1046867712 Chr.1: 154582451-154582451 on Build GRCh38
rs908359434 Chr.1: 154582799-154582799 on Build GRCh38
rs545120940 Chr.1: 154582579-154582579 on Build GRCh38
rs540739768 Chr.1: 154582385-154582385 on Build GRCh38
rs926050380 Chr.1: 154582771-154582771 on Build GRCh38
rs14002 Chr.1: 154582474-154582474 on Build GRCh38
rs898213219 Chr.1: 154582425-154582425 on Build GRCh38
rs1047362053 Chr.1: 154582777-154582777 on Build GRCh38
rs532724567 Chr.1: 154582450-154582450 on Build GRCh38
rs753888408 Chr.1: 154582762-154582762 on Build GRCh38
rs1008129045 Chr.1: 154582470-154582470 on Build GRCh38
rs200367762 Chr.1: 154582490-154582490 on Build GRCh38
rs958359847 Chr.1: 154582692-154582692 on Build GRCh38

View Full Product Details