Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122780915-122781378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC62
Entrez Gene ID
84660
Gene Name
coiled-coil domain containing 62
Gene Aliases
CT109, ERAP75, TSP-NY
UniGene
Hs.592008
Gene Chromosome Location
Chr.12: 122773621-122827385 on Build GRCh38
Transcripts
NM_201435.4 NR_027918.2 XM_006719643.1 XM_006719644.1 XM_011538856.1 XM_011538857.1 XM_017020033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359664 c.245G>T p.R82I Chr.12: 122781179-122781179 on Build GRCh38
4543870 c.345G>A p.Q115Q Chr.12: 122781279-122781279 on Build GRCh38
4487797 c.322C>T p.Q108* Chr.12: 122781256-122781256 on Build GRCh38
5645624 c.314C>T p.T105I Chr.12: 122781248-122781248 on Build GRCh38
6894770 c.327G>T p.K109N Chr.12: 122781261-122781261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774139244 Chr.12: 122781217-122781217 on Build GRCh38
rs1039432269 Chr.12: 122780976-122780976 on Build GRCh38
rs376000949 Chr.12: 122781246-122781246 on Build GRCh38
rs899578894 Chr.12: 122781033-122781033 on Build GRCh38
rs982015460 Chr.12: 122781061-122781061 on Build GRCh38
rs754981333 Chr.12: 122781258-122781258 on Build GRCh38
rs370074875 Chr.12: 122781345-122781345 on Build GRCh38
rs778982134 Chr.12: 122781176-122781176 on Build GRCh38
rs761119386 Chr.12: 122781278-122781278 on Build GRCh38
rs540999747 Chr.12: 122781001-122781001 on Build GRCh38
rs376082452 Chr.12: 122781203-122781203 on Build GRCh38
rs952602452 Chr.12: 122781341-122781341 on Build GRCh38
rs190725601 Chr.12: 122780998-122780998 on Build GRCh38
rs926533245 Chr.12: 122781094-122781094 on Build GRCh38
rs570227180 Chr.12: 122780934-122780934 on Build GRCh38
rs755108973 Chr.12: 122781174-122781174 on Build GRCh38
rs746449167 Chr.12: 122781350-122781350 on Build GRCh38
rs571581485 Chr.12: 122781060-122781060 on Build GRCh38
rs745492770 Chr.12: 122781213-122781215 on Build GRCh38
rs149063838 Chr.12: 122780956-122780956 on Build GRCh38
rs767486801 Chr.12: 122781115-122781115 on Build GRCh38
rs761615898 Chr.12: 122781234-122781234 on Build GRCh38
rs908426873 Chr.12: 122781212-122781213 on Build GRCh38
rs867260317 Chr.12: 122780935-122780935 on Build GRCh38
rs753824077 Chr.12: 122781173-122781173 on Build GRCh38
rs747285720 Chr.12: 122781333-122781333 on Build GRCh38
rs186017832 Chr.12: 122780944-122780944 on Build GRCh38
rs933788432 Chr.12: 122781034-122781034 on Build GRCh38
rs1010883985 Chr.12: 122781099-122781099 on Build GRCh38
rs141722651 Chr.12: 122781162-122781162 on Build GRCh38
rs758561527 Chr.12: 122781326-122781326 on Build GRCh38
rs372007109 Chr.12: 122781138-122781138 on Build GRCh38
rs781517166 Chr.12: 122781347-122781347 on Build GRCh38
rs145920110 Chr.12: 122781300-122781300 on Build GRCh38
rs1051186847 Chr.12: 122781053-122781053 on Build GRCh38
rs748722684 Chr.12: 122781204-122781204 on Build GRCh38
rs546971318 Chr.12: 122781045-122781045 on Build GRCh38
rs908207442 Chr.12: 122780982-122780982 on Build GRCh38
rs764579807 Chr.12: 122781330-122781330 on Build GRCh38
rs372877789 Chr.12: 122781206-122781206 on Build GRCh38
rs952474825 Chr.12: 122780961-122780961 on Build GRCh38
rs771873623 Chr.12: 122781303-122781305 on Build GRCh38
rs773072646 Chr.12: 122781248-122781248 on Build GRCh38
rs778172327 Chr.12: 122781198-122781198 on Build GRCh38
rs149651954 Chr.12: 122781327-122781327 on Build GRCh38
rs12312837 Chr.12: 122781141-122781141 on Build GRCh38
rs567971468 Chr.12: 122780995-122780995 on Build GRCh38
rs765372582 Chr.12: 122781299-122781299 on Build GRCh38
rs982449999 Chr.12: 122780968-122780969 on Build GRCh38
rs201788426 Chr.12: 122781179-122781179 on Build GRCh38
rs951426691 Chr.12: 122781315-122781315 on Build GRCh38
rs139405893 Chr.12: 122781324-122781324 on Build GRCh38
rs753864344 Chr.12: 122781255-122781255 on Build GRCh38
rs369606370 Chr.12: 122781187-122781187 on Build GRCh38
rs766180037 Chr.12: 122781251-122781251 on Build GRCh38
rs147096647 Chr.12: 122781249-122781249 on Build GRCh38

View Full Product Details