Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11294647-11295191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELX
Entrez Gene ID
265
Gene Name
amelogenin, X-linked
Gene Aliases
AI1E, AIH1, ALGN, AMG, AMGL, AMGX
UniGene
Hs.654436
Gene Chromosome Location
Chr.X: 11293413-11300761 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 XM_017029404.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

Gene Symbol
ARHGAP6
Entrez Gene ID
395
Gene Name
Rho GTPase activating protein 6
Gene Aliases
RHOGAP6, RHOGAPX-1
UniGene
Hs.435291
Gene Chromosome Location
Chr.X: 11137543-11665701 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 XM_017029404.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

SNP Detail

SNP ID Chromosome Location
rs12850389 Chr.X: 11295136-11295136 on Build GRCh38
rs946252 Chr.X: 11294907-11294907 on Build GRCh38
rs780336512 Chr.X: 11294818-11294818 on Build GRCh38
rs1008213552 Chr.X: 11294943-11294943 on Build GRCh38
rs949997666 Chr.X: 11295146-11295146 on Build GRCh38
rs946251 Chr.X: 11295020-11295020 on Build GRCh38
rs387906488 Chr.X: 11294802-11294810 on Build GRCh38
rs953677617 Chr.X: 11295084-11295084 on Build GRCh38
rs748483718 Chr.X: 11294855-11294855 on Build GRCh38
rs139502132 Chr.X: 11295021-11295021 on Build GRCh38
rs142795990 Chr.X: 11295045-11295045 on Build GRCh38
rs184371797 Chr.X: 11294743-11294743 on Build GRCh38
rs760464147 Chr.X: 11294769-11294769 on Build GRCh38
rs104894738 Chr.X: 11294799-11294799 on Build GRCh38
rs772844760 Chr.X: 11294937-11294937 on Build GRCh38
rs907794530 Chr.X: 11294674-11294674 on Build GRCh38
rs775768556 Chr.X: 11294851-11294851 on Build GRCh38
rs756431625 Chr.X: 11294862-11294862 on Build GRCh38
rs752404330 Chr.X: 11295128-11295128 on Build GRCh38
rs764153884 Chr.X: 11294782-11294782 on Build GRCh38
rs964053448 Chr.X: 11294990-11294990 on Build GRCh38
rs1045414805 Chr.X: 11294703-11294703 on Build GRCh38
rs753863554 Chr.X: 11294796-11294796 on Build GRCh38
rs1034070836 Chr.X: 11294951-11294951 on Build GRCh38
rs751652555 Chr.X: 11294823-11294823 on Build GRCh38
rs1010558343 Chr.X: 11294723-11294723 on Build GRCh38
rs949745077 Chr.X: 11294681-11294681 on Build GRCh38
rs749844204 Chr.X: 11294878-11294878 on Build GRCh38
rs104894737 Chr.X: 11294790-11294790 on Build GRCh38
rs755158864 Chr.X: 11294838-11294838 on Build GRCh38
rs766401250 Chr.X: 11294974-11294974 on Build GRCh38
rs951791307 Chr.X: 11294958-11294958 on Build GRCh38
rs961706717 Chr.X: 11295022-11295022 on Build GRCh38
rs879079881 Chr.X: 11295058-11295058 on Build GRCh38
rs757235287 Chr.X: 11294806-11294806 on Build GRCh38
rs1001209376 Chr.X: 11294731-11294731 on Build GRCh38
rs905603443 Chr.X: 11294721-11294721 on Build GRCh38
rs767197248 Chr.X: 11294755-11294755 on Build GRCh38
rs760714398 Chr.X: 11294970-11294970 on Build GRCh38
rs983340182 Chr.X: 11295054-11295054 on Build GRCh38
rs1020946655 Chr.X: 11294822-11294822 on Build GRCh38
rs1045624931 Chr.X: 11294734-11294734 on Build GRCh38
rs771907722 Chr.X: 11294931-11294931 on Build GRCh38
rs759170832 Chr.X: 11294749-11294749 on Build GRCh38
rs752442424 Chr.X: 11294756-11294756 on Build GRCh38
rs750679679 Chr.X: 11295055-11295055 on Build GRCh38
rs778080525 Chr.X: 11294867-11294867 on Build GRCh38

View Full Product Details