Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87253982-87254531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
UniGene
Hs.476930
Gene Chromosome Location
Chr.3: 87227263-87255548 on Build GRCh38
Transcripts
NM_001244644.1 NM_014043.3 XM_011533576.2

SNP Detail

SNP ID Chromosome Location
rs408793 Chr.3: 87254090-87254090 on Build GRCh38
rs390857 Chr.3: 87254311-87254311 on Build GRCh38
rs948704485 Chr.3: 87254333-87254333 on Build GRCh38
rs889386991 Chr.3: 87254028-87254028 on Build GRCh38
rs985821803 Chr.3: 87254104-87254104 on Build GRCh38
rs910304227 Chr.3: 87254183-87254183 on Build GRCh38
rs114730892 Chr.3: 87254265-87254265 on Build GRCh38
rs535402661 Chr.3: 87254239-87254239 on Build GRCh38
rs139070624 Chr.3: 87254479-87254479 on Build GRCh38
rs956689387 Chr.3: 87254051-87254051 on Build GRCh38
rs384094 Chr.3: 87254436-87254436 on Build GRCh38
rs550468260 Chr.3: 87254064-87254064 on Build GRCh38
rs886058908 Chr.3: 87254424-87254424 on Build GRCh38
rs904642784 Chr.3: 87254402-87254402 on Build GRCh38
rs1031726547 Chr.3: 87254206-87254206 on Build GRCh38
rs448316 Chr.3: 87254033-87254033 on Build GRCh38
rs17189270 Chr.3: 87254053-87254053 on Build GRCh38
rs909088119 Chr.3: 87254411-87254411 on Build GRCh38
rs936487490 Chr.3: 87254419-87254419 on Build GRCh38
rs954392543 Chr.3: 87254083-87254083 on Build GRCh38
rs1022953735 Chr.3: 87254262-87254262 on Build GRCh38
rs1044465877 Chr.3: 87254380-87254380 on Build GRCh38
rs971797498 Chr.3: 87254448-87254448 on Build GRCh38
rs969361327 Chr.3: 87254300-87254300 on Build GRCh38
rs981090210 Chr.3: 87254389-87254389 on Build GRCh38
rs1044512 Chr.3: 87254173-87254173 on Build GRCh38
rs575796569 Chr.3: 87254493-87254493 on Build GRCh38
rs189471374 Chr.3: 87254288-87254288 on Build GRCh38
rs568774746 Chr.3: 87254213-87254213 on Build GRCh38
rs992805770 Chr.3: 87254248-87254248 on Build GRCh38
rs969289188 Chr.3: 87254184-87254184 on Build GRCh38
rs764379791 Chr.3: 87254015-87254015 on Build GRCh38
rs960041852 Chr.3: 87254233-87254233 on Build GRCh38
rs541208359 Chr.3: 87254016-87254016 on Build GRCh38
rs748575704 Chr.3: 87254252-87254252 on Build GRCh38
rs1060230 Chr.3: 87254365-87254365 on Build GRCh38
rs1030029418 Chr.3: 87254076-87254076 on Build GRCh38
rs393439 Chr.3: 87254088-87254088 on Build GRCh38
rs371181321 Chr.3: 87254202-87254202 on Build GRCh38
rs181525689 Chr.3: 87254435-87254435 on Build GRCh38

View Full Product Details