Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61680919-61681253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3 XM_006717692.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932002 c.9C>A p.F3L Chr.10: 61681251-61681251 on Build GRCh38
6264521 c.2T>C p.M1T Chr.10: 61681244-61681244 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029300166 Chr.10: 61681075-61681075 on Build GRCh38
rs779731267 Chr.10: 61680968-61680968 on Build GRCh38
rs771504743 Chr.10: 61681220-61681220 on Build GRCh38
rs982725685 Chr.10: 61681113-61681113 on Build GRCh38
rs867787373 Chr.10: 61681049-61681049 on Build GRCh38
rs529363354 Chr.10: 61680990-61680990 on Build GRCh38
rs963665474 Chr.10: 61681000-61681000 on Build GRCh38
rs562151534 Chr.10: 61680959-61680959 on Build GRCh38
rs183434407 Chr.10: 61681055-61681055 on Build GRCh38
rs963465879 Chr.10: 61681069-61681069 on Build GRCh38
rs1007135582 Chr.10: 61680993-61680993 on Build GRCh38
rs778744307 Chr.10: 61681196-61681196 on Build GRCh38
rs962612386 Chr.10: 61681150-61681150 on Build GRCh38
rs745348268 Chr.10: 61681216-61681216 on Build GRCh38
rs879624102 Chr.10: 61681124-61681124 on Build GRCh38
rs192520061 Chr.10: 61680957-61680957 on Build GRCh38
rs544644319 Chr.10: 61681077-61681077 on Build GRCh38
rs71018992 Chr.10: 61681054-61681055 on Build GRCh38
rs1000346638 Chr.10: 61680989-61680989 on Build GRCh38
rs908362479 Chr.10: 61681121-61681121 on Build GRCh38
rs200926415 Chr.10: 61681194-61681194 on Build GRCh38
rs140407515 Chr.10: 61681049-61681050 on Build GRCh38
rs996498889 Chr.10: 61681064-61681064 on Build GRCh38
rs12357502 Chr.10: 61680974-61680974 on Build GRCh38
rs919275577 Chr.10: 61681149-61681149 on Build GRCh38

View Full Product Details