Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89627532-89628110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
UniGene
Hs.513134
Gene Chromosome Location
Chr.15: 89627970-89663086 on Build GRCh38
Transcripts
NM_198525.2 XM_011521531.2 NM_001308025.1 NM_152259.3

Gene Symbol
TICRR
Entrez Gene ID
90381
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C15orf42, SLD3, Treslin
UniGene
Hs.441708
Gene Chromosome Location
Chr.15: 89575587-89628023 on Build GRCh38
Transcripts
NM_198525.2 XM_011521531.2 NM_001308025.1 NM_152259.3

SNP Detail

SNP ID Chromosome Location
rs1038379116 Chr.15: 89627945-89627945 on Build GRCh38
rs1025757151 Chr.15: 89628012-89628012 on Build GRCh38
rs962817879 Chr.15: 89627779-89627779 on Build GRCh38
rs1036112703 Chr.15: 89627961-89627962 on Build GRCh38
rs969307512 Chr.15: 89627890-89627890 on Build GRCh38
rs3195377 Chr.15: 89627870-89627870 on Build GRCh38
rs964091354 Chr.15: 89627831-89627831 on Build GRCh38
rs906291999 Chr.15: 89627682-89627682 on Build GRCh38
rs998877657 Chr.15: 89627823-89627823 on Build GRCh38
rs564842729 Chr.15: 89627561-89627561 on Build GRCh38
rs532972088 Chr.15: 89627715-89627715 on Build GRCh38
rs987490682 Chr.15: 89627813-89627818 on Build GRCh38
rs539941455 Chr.15: 89627791-89627791 on Build GRCh38
rs754624089 Chr.15: 89627578-89627579 on Build GRCh38
rs979509188 Chr.15: 89628074-89628074 on Build GRCh38
rs774087431 Chr.15: 89628080-89628080 on Build GRCh38
rs947716639 Chr.15: 89627654-89627654 on Build GRCh38
rs939109009 Chr.15: 89628036-89628036 on Build GRCh38
rs927602791 Chr.15: 89628024-89628024 on Build GRCh38
rs1036563403 Chr.15: 89627699-89627699 on Build GRCh38
rs1018925195 Chr.15: 89627783-89627783 on Build GRCh38
rs566074793 Chr.15: 89628089-89628089 on Build GRCh38
rs1008479181 Chr.15: 89628042-89628042 on Build GRCh38
rs980857672 Chr.15: 89627929-89627929 on Build GRCh38
rs551376264 Chr.15: 89627775-89627775 on Build GRCh38
rs71763708 Chr.15: 89627878-89627879 on Build GRCh38
rs949317642 Chr.15: 89627881-89627881 on Build GRCh38
rs76530445 Chr.15: 89627689-89627689 on Build GRCh38
rs1017469220 Chr.15: 89627739-89627739 on Build GRCh38
rs986502397 Chr.15: 89627553-89627553 on Build GRCh38
rs759673742 Chr.15: 89628081-89628081 on Build GRCh38
rs746563228 Chr.15: 89627600-89627600 on Build GRCh38
rs567482406 Chr.15: 89628041-89628041 on Build GRCh38
rs535935295 Chr.15: 89628046-89628046 on Build GRCh38
rs145910246 Chr.15: 89627603-89627603 on Build GRCh38
rs767988951 Chr.15: 89627609-89627609 on Build GRCh38
rs113145397 Chr.15: 89627988-89627989 on Build GRCh38
rs182275540 Chr.15: 89627611-89627611 on Build GRCh38
rs1007976182 Chr.15: 89627782-89627782 on Build GRCh38
rs3743371 Chr.15: 89627702-89627702 on Build GRCh38
rs989292396 Chr.15: 89627857-89627857 on Build GRCh38
rs925029752 Chr.15: 89627836-89627836 on Build GRCh38
rs942454162 Chr.15: 89627585-89627585 on Build GRCh38
rs762684798 Chr.15: 89627936-89627936 on Build GRCh38
rs977475987 Chr.15: 89627782-89627794 on Build GRCh38
rs915144349 Chr.15: 89627882-89627882 on Build GRCh38
rs773037385 Chr.15: 89627758-89627758 on Build GRCh38
rs1043892119 Chr.15: 89627586-89627586 on Build GRCh38
rs1045410279 Chr.15: 89627883-89627883 on Build GRCh38
rs1049191221 Chr.15: 89628059-89628059 on Build GRCh38
rs963711107 Chr.15: 89628053-89628053 on Build GRCh38
rs536949336 Chr.15: 89627931-89627931 on Build GRCh38
rs955635933 Chr.15: 89627811-89627811 on Build GRCh38
rs1007795358 Chr.15: 89628077-89628077 on Build GRCh38
rs569892063 Chr.15: 89627832-89627832 on Build GRCh38
rs905860927 Chr.15: 89627918-89627918 on Build GRCh38
rs543957612 Chr.15: 89627607-89627607 on Build GRCh38
rs398057924 Chr.15: 89627882-89627883 on Build GRCh38
rs528928133 Chr.15: 89627612-89627612 on Build GRCh38
rs966062445 Chr.15: 89627587-89627587 on Build GRCh38
rs987919322 Chr.15: 89628079-89628079 on Build GRCh38
rs138875667 Chr.15: 89628038-89628038 on Build GRCh38
rs1052284677 Chr.15: 89627875-89627878 on Build GRCh38
rs889339665 Chr.15: 89627757-89627757 on Build GRCh38
rs1044694505 Chr.15: 89627672-89627672 on Build GRCh38
rs910873575 Chr.15: 89627829-89627829 on Build GRCh38
rs976567219 Chr.15: 89627592-89627592 on Build GRCh38
rs910935200 Chr.15: 89627572-89627573 on Build GRCh38
rs551310792 Chr.15: 89627855-89627855 on Build GRCh38
rs930806498 Chr.15: 89628048-89628048 on Build GRCh38
rs548897408 Chr.15: 89627799-89627800 on Build GRCh38
rs966127119 Chr.15: 89627786-89627786 on Build GRCh38
rs1028314330 Chr.15: 89627750-89627750 on Build GRCh38
rs995129581 Chr.15: 89627735-89627735 on Build GRCh38
rs1054435 Chr.15: 89628075-89628075 on Build GRCh38

View Full Product Details