Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7688076-7688672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_017024812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986721 c.1A>G p.M1V Chr.17: 7688649-7688649 on Build GRCh38
219982 c.7A>C p.T3P Chr.17: 7688655-7688655 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560833428 Chr.17: 7688349-7688349 on Build GRCh38
rs755130675 Chr.17: 7688629-7688629 on Build GRCh38
rs867179605 Chr.17: 7688649-7688649 on Build GRCh38
rs548795605 Chr.17: 7688518-7688518 on Build GRCh38
rs927417073 Chr.17: 7688273-7688273 on Build GRCh38
rs761454111 Chr.17: 7688609-7688609 on Build GRCh38
rs1040642619 Chr.17: 7688233-7688233 on Build GRCh38
rs986262623 Chr.17: 7688615-7688615 on Build GRCh38
rs34097279 Chr.17: 7688191-7688191 on Build GRCh38
rs17551150 Chr.17: 7688443-7688443 on Build GRCh38
rs564330662 Chr.17: 7688394-7688394 on Build GRCh38
rs564890743 Chr.17: 7688424-7688424 on Build GRCh38
rs913237048 Chr.17: 7688192-7688192 on Build GRCh38
rs960318586 Chr.17: 7688479-7688479 on Build GRCh38
rs759226877 Chr.17: 7688641-7688642 on Build GRCh38
rs896833793 Chr.17: 7688351-7688351 on Build GRCh38
rs758882247 Chr.17: 7688249-7688249 on Build GRCh38
rs1030693849 Chr.17: 7688204-7688204 on Build GRCh38
rs888460319 Chr.17: 7688323-7688323 on Build GRCh38
rs750849403 Chr.17: 7688513-7688513 on Build GRCh38
rs1052258786 Chr.17: 7688402-7688402 on Build GRCh38
rs17883670 Chr.17: 7688404-7688404 on Build GRCh38
rs967391051 Chr.17: 7688503-7688503 on Build GRCh38
rs1057252209 Chr.17: 7688148-7688148 on Build GRCh38
rs572121442 Chr.17: 7688252-7688252 on Build GRCh38
rs531543362 Chr.17: 7688357-7688357 on Build GRCh38
rs530208087 Chr.17: 7688516-7688516 on Build GRCh38
rs918768607 Chr.17: 7688519-7688519 on Build GRCh38
rs1050620062 Chr.17: 7688245-7688245 on Build GRCh38
rs1018487748 Chr.17: 7688196-7688196 on Build GRCh38
rs969094860 Chr.17: 7688467-7688467 on Build GRCh38
rs927378297 Chr.17: 7688161-7688161 on Build GRCh38
rs556234644 Chr.17: 7688218-7688218 on Build GRCh38
rs542836334 Chr.17: 7688294-7688294 on Build GRCh38
rs750344241 Chr.17: 7688626-7688626 on Build GRCh38
rs17882910 Chr.17: 7688237-7688237 on Build GRCh38
rs923542780 Chr.17: 7688514-7688514 on Build GRCh38
rs1039607781 Chr.17: 7688336-7688336 on Build GRCh38
rs11539199 Chr.17: 7688550-7688550 on Build GRCh38
rs35833877 Chr.17: 7688221-7688221 on Build GRCh38
rs372820855 Chr.17: 7688601-7688601 on Build GRCh38
rs866880761 Chr.17: 7688281-7688281 on Build GRCh38
rs901125160 Chr.17: 7688341-7688341 on Build GRCh38
rs997859612 Chr.17: 7688202-7688202 on Build GRCh38
rs1057516027 Chr.17: 7688335-7688335 on Build GRCh38
rs780391494 Chr.17: 7688260-7688260 on Build GRCh38
rs879058020 Chr.17: 7688557-7688557 on Build GRCh38
rs951508354 Chr.17: 7688563-7688563 on Build GRCh38
rs942586705 Chr.17: 7688333-7688333 on Build GRCh38
rs990211123 Chr.17: 7688449-7688449 on Build GRCh38
rs901028817 Chr.17: 7688199-7688199 on Build GRCh38
rs932258006 Chr.17: 7688559-7688559 on Build GRCh38
rs930112339 Chr.17: 7688320-7688320 on Build GRCh38
rs748265946 Chr.17: 7688501-7688501 on Build GRCh38
rs869221578 Chr.17: 7688201-7688202 on Build GRCh38
rs984243245 Chr.17: 7688593-7688593 on Build GRCh38
rs990076465 Chr.17: 7688181-7688181 on Build GRCh38
rs370181839 Chr.17: 7688410-7688410 on Build GRCh38
rs1421314 Chr.17: 7688222-7688222 on Build GRCh38
rs17883184 Chr.17: 7688236-7688236 on Build GRCh38
rs397856875 Chr.17: 7688200-7688201 on Build GRCh38
rs765268037 Chr.17: 7688623-7688623 on Build GRCh38
rs886053517 Chr.17: 7688563-7688563 on Build GRCh38
rs980366857 Chr.17: 7688102-7688102 on Build GRCh38
rs929949871 Chr.17: 7688195-7688196 on Build GRCh38
rs888355656 Chr.17: 7688195-7688195 on Build GRCh38
rs1024209548 Chr.17: 7688376-7688376 on Build GRCh38
rs17551157 Chr.17: 7688193-7688194 on Build GRCh38
rs747300674 Chr.17: 7688311-7688311 on Build GRCh38
rs938819561 Chr.17: 7688285-7688285 on Build GRCh38
rs570354139 Chr.17: 7688642-7688642 on Build GRCh38
rs186185308 Chr.17: 7688162-7688162 on Build GRCh38
rs1048842870 Chr.17: 7688321-7688321 on Build GRCh38
rs79199718 Chr.17: 7688511-7688511 on Build GRCh38
rs199831002 Chr.17: 7688196-7688197 on Build GRCh38
rs879619965 Chr.17: 7688236-7688237 on Build GRCh38
rs781470763 Chr.17: 7688640-7688640 on Build GRCh38
rs386795047 Chr.17: 7688236-7688237 on Build GRCh38
rs547538493 Chr.17: 7688472-7688472 on Build GRCh38
rs889530811 Chr.17: 7688253-7688253 on Build GRCh38

View Full Product Details