Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7005394-7005993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12
Entrez Gene ID
239
Gene Name
arachidonate 12-lipoxygenase, 12S type
Gene Aliases
12-LOX, 12S-LOX, LOG12
UniGene
Hs.654431
Gene Chromosome Location
Chr.17: 6993791-7010736 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Gene Symbol
ALOX12-AS1
Entrez Gene ID
100506713
Gene Name
ALOX12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.422967
Gene Chromosome Location
Chr.17: 6985123-7012334 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069106 c.1288C>A p.R430S Chr.17: 7005897-7005897 on Build GRCh38
7144562 c.1375C>G p.L459V Chr.17: 7005984-7005984 on Build GRCh38
6598642 c.1379A>G p.Y460C Chr.17: 7005988-7005988 on Build GRCh38
6147426 c.1348_1349CG>AA p.R450K Chr.17: 7005957-7005958 on Build GRCh38
6894444 c.1377C>G p.L459L Chr.17: 7005986-7005986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954727193 Chr.17: 7005814-7005814 on Build GRCh38
rs902036653 Chr.17: 7005970-7005970 on Build GRCh38
rs1028993342 Chr.17: 7005806-7005806 on Build GRCh38
rs755559457 Chr.17: 7005851-7005851 on Build GRCh38
rs772988081 Chr.17: 7005901-7005901 on Build GRCh38
rs141140949 Chr.17: 7005763-7005763 on Build GRCh38
rs980537275 Chr.17: 7005858-7005858 on Build GRCh38
rs772153398 Chr.17: 7005897-7005897 on Build GRCh38
rs749924865 Chr.17: 7005474-7005475 on Build GRCh38
rs754684647 Chr.17: 7005897-7005898 on Build GRCh38
rs553679162 Chr.17: 7005958-7005958 on Build GRCh38
rs575566127 Chr.17: 7005478-7005478 on Build GRCh38
rs12453632 Chr.17: 7005599-7005599 on Build GRCh38
rs745960734 Chr.17: 7005894-7005894 on Build GRCh38
rs200546604 Chr.17: 7005900-7005900 on Build GRCh38
rs946336605 Chr.17: 7005878-7005878 on Build GRCh38
rs530265926 Chr.17: 7005606-7005606 on Build GRCh38
rs765981671 Chr.17: 7005833-7005833 on Build GRCh38
rs749577929 Chr.17: 7005883-7005883 on Build GRCh38
rs917710796 Chr.17: 7005520-7005520 on Build GRCh38
rs763428921 Chr.17: 7005475-7005475 on Build GRCh38
rs577783457 Chr.17: 7005625-7005625 on Build GRCh38
rs772868538 Chr.17: 7005820-7005820 on Build GRCh38
rs987708916 Chr.17: 7005831-7005831 on Build GRCh38
rs768159638 Chr.17: 7005853-7005853 on Build GRCh38
rs754117733 Chr.17: 7005969-7005969 on Build GRCh38
rs895489472 Chr.17: 7005616-7005616 on Build GRCh38
rs937568540 Chr.17: 7005563-7005563 on Build GRCh38
rs1662682 Chr.17: 7005538-7005538 on Build GRCh38
rs762709740 Chr.17: 7005904-7005904 on Build GRCh38
rs757414782 Chr.17: 7005884-7005884 on Build GRCh38
rs886935442 Chr.17: 7005635-7005635 on Build GRCh38
rs144428758 Chr.17: 7005472-7005472 on Build GRCh38
rs974825328 Chr.17: 7005725-7005725 on Build GRCh38
rs970439869 Chr.17: 7005788-7005788 on Build GRCh38
rs926324122 Chr.17: 7005550-7005550 on Build GRCh38
rs913391967 Chr.17: 7005453-7005453 on Build GRCh38
rs753212708 Chr.17: 7005859-7005859 on Build GRCh38
rs764615926 Chr.17: 7005966-7005966 on Build GRCh38
rs961910830 Chr.17: 7005444-7005444 on Build GRCh38
rs1024719235 Chr.17: 7005717-7005717 on Build GRCh38
rs534787064 Chr.17: 7005566-7005566 on Build GRCh38
rs569242292 Chr.17: 7005872-7005872 on Build GRCh38
rs532478875 Chr.17: 7005733-7005733 on Build GRCh38
rs889123706 Chr.17: 7005653-7005653 on Build GRCh38
rs971797120 Chr.17: 7005497-7005497 on Build GRCh38
rs756551061 Chr.17: 7005870-7005870 on Build GRCh38
rs371454972 Chr.17: 7005885-7005885 on Build GRCh38
rs767069577 Chr.17: 7005957-7005957 on Build GRCh38
rs762500426 Chr.17: 7005821-7005821 on Build GRCh38
rs778247211 Chr.17: 7005913-7005922 on Build GRCh38
rs185197646 Chr.17: 7005605-7005605 on Build GRCh38
rs868017585 Chr.17: 7005671-7005671 on Build GRCh38
rs751004020 Chr.17: 7005844-7005844 on Build GRCh38
rs201728791 Chr.17: 7005474-7005474 on Build GRCh38
rs544928028 Chr.17: 7005626-7005626 on Build GRCh38
rs201453162 Chr.17: 7005963-7005963 on Build GRCh38
rs149233598 Chr.17: 7005905-7005905 on Build GRCh38
rs560683777 Chr.17: 7005630-7005630 on Build GRCh38
rs35201122 Chr.17: 7005498-7005499 on Build GRCh38
rs980465310 Chr.17: 7005542-7005542 on Build GRCh38
rs746308885 Chr.17: 7005608-7005608 on Build GRCh38
rs114312186 Chr.17: 7005914-7005914 on Build GRCh38
rs1016214258 Chr.17: 7005681-7005681 on Build GRCh38
rs1006514787 Chr.17: 7005667-7005667 on Build GRCh38
rs946205187 Chr.17: 7005944-7005944 on Build GRCh38
rs987759465 Chr.17: 7005435-7005435 on Build GRCh38
rs979311996 Chr.17: 7005891-7005891 on Build GRCh38
rs751221343 Chr.17: 7005823-7005823 on Build GRCh38
rs574231912 Chr.17: 7005432-7005432 on Build GRCh38
rs11078661 Chr.17: 7005584-7005584 on Build GRCh38
rs11571342 Chr.17: 7005898-7005898 on Build GRCh38
rs369898939 Chr.17: 7005567-7005567 on Build GRCh38
rs962180791 Chr.17: 7005684-7005684 on Build GRCh38
rs1025552878 Chr.17: 7005634-7005634 on Build GRCh38
rs557231605 Chr.17: 7005700-7005700 on Build GRCh38
rs758997226 Chr.17: 7005951-7005951 on Build GRCh38
rs949017350 Chr.17: 7005541-7005541 on Build GRCh38
rs893261340 Chr.17: 7005579-7005579 on Build GRCh38

View Full Product Details