Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35166197-35166692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD5
Entrez Gene ID
53827
Gene Name
FXYD domain containing ion transport regulator 5
Gene Aliases
DYSAD, HSPC113, IWU1, KCT1, OIT2, PRO6241, RIC
UniGene
Hs.333418
Gene Chromosome Location
Chr.19: 35154722-35169885 on Build GRCh38
Transcripts
NM_001164605.1 NM_001320912.1 NM_001320913.1 NM_014164.5 NM_144779.2 NR_028406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166345 c.220C>T p.R74W Chr.19: 35166271-35166271 on Build GRCh38
5202395 c.470_471insCAT p.I160_L161insI Chr.19: 35166308-35166309 on Build GRCh38
3197450 c.200A>T p.D67V Chr.19: 35166251-35166251 on Build GRCh38
1241196 c.438G>A p.G146G Chr.19: 35166276-35166276 on Build GRCh38
5202396 c.470_471insCAT p.I160_L161insI Chr.19: 35166308-35166309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944441728 Chr.19: 35166241-35166241 on Build GRCh38
rs890242900 Chr.19: 35166217-35166217 on Build GRCh38
rs774069675 Chr.19: 35166464-35166464 on Build GRCh38
rs773413800 Chr.19: 35166355-35166355 on Build GRCh38
rs749245946 Chr.19: 35166449-35166449 on Build GRCh38
rs369833565 Chr.19: 35166380-35166380 on Build GRCh38
rs776686477 Chr.19: 35166252-35166252 on Build GRCh38
rs201399482 Chr.19: 35166258-35166258 on Build GRCh38
rs755441989 Chr.19: 35166269-35166269 on Build GRCh38
rs762679900 Chr.19: 35166259-35166259 on Build GRCh38
rs748493009 Chr.19: 35166272-35166272 on Build GRCh38
rs768439280 Chr.19: 35166453-35166453 on Build GRCh38
rs769724741 Chr.19: 35166310-35166310 on Build GRCh38
rs780916646 Chr.19: 35166292-35166292 on Build GRCh38
rs145467834 Chr.19: 35166297-35166297 on Build GRCh38
rs1004360163 Chr.19: 35166644-35166644 on Build GRCh38
rs753853880 Chr.19: 35166373-35166373 on Build GRCh38
rs150419745 Chr.19: 35166435-35166435 on Build GRCh38
rs920514209 Chr.19: 35166390-35166390 on Build GRCh38
rs779297539 Chr.19: 35166271-35166271 on Build GRCh38
rs1017626398 Chr.19: 35166606-35166606 on Build GRCh38
rs1057061409 Chr.19: 35166600-35166600 on Build GRCh38
rs1052157367 Chr.19: 35166450-35166450 on Build GRCh38
rs372529207 Chr.19: 35166266-35166266 on Build GRCh38
rs759605413 Chr.19: 35166255-35166255 on Build GRCh38
rs570205448 Chr.19: 35166626-35166626 on Build GRCh38
rs185457352 Chr.19: 35166459-35166459 on Build GRCh38
rs887141592 Chr.19: 35166512-35166512 on Build GRCh38
rs200190054 Chr.19: 35166256-35166256 on Build GRCh38
rs775323352 Chr.19: 35166330-35166330 on Build GRCh38
rs567697320 Chr.19: 35166597-35166597 on Build GRCh38
rs991943700 Chr.19: 35166667-35166667 on Build GRCh38
rs1015658279 Chr.19: 35166376-35166376 on Build GRCh38
rs758717714 Chr.19: 35166279-35166279 on Build GRCh38
rs773958211 Chr.19: 35166513-35166513 on Build GRCh38
rs777534958 Chr.19: 35166280-35166280 on Build GRCh38
rs115016713 Chr.19: 35166591-35166591 on Build GRCh38
rs1051093365 Chr.19: 35166468-35166468 on Build GRCh38
rs746706137 Chr.19: 35166285-35166285 on Build GRCh38
rs200435409 Chr.19: 35166265-35166265 on Build GRCh38
rs73044024 Chr.19: 35166431-35166431 on Build GRCh38
rs781014916 Chr.19: 35166440-35166440 on Build GRCh38
rs748919394 Chr.19: 35166444-35166444 on Build GRCh38
rs75807286 Chr.19: 35166381-35166381 on Build GRCh38
rs531039278 Chr.19: 35166529-35166529 on Build GRCh38
rs748252488 Chr.19: 35166309-35166309 on Build GRCh38
rs767740800 Chr.19: 35166260-35166260 on Build GRCh38
rs758865132 Chr.19: 35166314-35166314 on Build GRCh38
rs745343854 Chr.19: 35166443-35166443 on Build GRCh38
rs1015689284 Chr.19: 35166660-35166660 on Build GRCh38
rs551038509 Chr.19: 35166551-35166551 on Build GRCh38
rs750686218 Chr.19: 35166262-35166262 on Build GRCh38
rs551621866 Chr.19: 35166400-35166400 on Build GRCh38
rs199648617 Chr.19: 35166286-35166286 on Build GRCh38
rs771089322 Chr.19: 35166470-35166470 on Build GRCh38
rs960802824 Chr.19: 35166342-35166342 on Build GRCh38
rs755329112 Chr.19: 35166447-35166447 on Build GRCh38
rs551699128 Chr.19: 35166539-35166539 on Build GRCh38
rs553418796 Chr.19: 35166429-35166429 on Build GRCh38
rs770604373 Chr.19: 35166234-35166234 on Build GRCh38
rs530105083 Chr.19: 35166665-35166665 on Build GRCh38
rs8106303 Chr.19: 35166448-35166448 on Build GRCh38
rs755229821 Chr.19: 35166284-35166284 on Build GRCh38
rs979383623 Chr.19: 35166424-35166424 on Build GRCh38
rs766588841 Chr.19: 35166362-35166362 on Build GRCh38
rs768492251 Chr.19: 35166354-35166354 on Build GRCh38
rs762781344 Chr.19: 35166350-35166350 on Build GRCh38

View Full Product Details