Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132891830-132892406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
UniGene
Hs.370854
Gene Chromosome Location
Chr.9: 132891348-132945269 on Build GRCh38
Transcripts
NM_000368.4 NM_001162426.1 NM_001162427.1 XM_005272211.1 XM_006717271.1 XM_011518979.2 XM_017015096.1 XM_017015097.1 XM_017015098.1 XM_017015099.1 XM_017015100.1 XM_017015101.1

SNP Detail

SNP ID Chromosome Location
rs955358269 Chr.9: 132892331-132892331 on Build GRCh38
rs766856579 Chr.9: 132892099-132892099 on Build GRCh38
rs558930752 Chr.9: 132892380-132892380 on Build GRCh38
rs907357659 Chr.9: 132892151-132892151 on Build GRCh38
rs966448764 Chr.9: 132892317-132892317 on Build GRCh38
rs139801034 Chr.9: 132891981-132891981 on Build GRCh38
rs986559590 Chr.9: 132892026-132892026 on Build GRCh38
rs1016277383 Chr.9: 132892061-132892061 on Build GRCh38
rs1021974016 Chr.9: 132892306-132892306 on Build GRCh38
rs567187061 Chr.9: 132891948-132891948 on Build GRCh38
rs796893029 Chr.9: 132892097-132892097 on Build GRCh38
rs886063567 Chr.9: 132891987-132891987 on Build GRCh38
rs751581381 Chr.9: 132892322-132892322 on Build GRCh38
rs910930441 Chr.9: 132892074-132892074 on Build GRCh38
rs114454155 Chr.9: 132892283-132892283 on Build GRCh38
rs959051780 Chr.9: 132892116-132892116 on Build GRCh38
rs937769454 Chr.9: 132891921-132891922 on Build GRCh38
rs886063571 Chr.9: 132892076-132892076 on Build GRCh38
rs766234756 Chr.9: 132891959-132891959 on Build GRCh38
rs886063574 Chr.9: 132892248-132892248 on Build GRCh38
rs1034791140 Chr.9: 132891893-132891893 on Build GRCh38
rs765057102 Chr.9: 132892374-132892374 on Build GRCh38
rs1006238490 Chr.9: 132892342-132892342 on Build GRCh38
rs564238927 Chr.9: 132891876-132891876 on Build GRCh38
rs918403035 Chr.9: 132891994-132891994 on Build GRCh38
rs886063569 Chr.9: 132892042-132892042 on Build GRCh38
rs547579970 Chr.9: 132892168-132892168 on Build GRCh38
rs182163633 Chr.9: 132892214-132892214 on Build GRCh38
rs922441092 Chr.9: 132892321-132892321 on Build GRCh38
rs886063575 Chr.9: 132892311-132892311 on Build GRCh38
rs111543364 Chr.9: 132892215-132892215 on Build GRCh38
rs990734980 Chr.9: 132891921-132891921 on Build GRCh38
rs901058761 Chr.9: 132891993-132891993 on Build GRCh38
rs1026493450 Chr.9: 132891853-132891853 on Build GRCh38
rs926319710 Chr.9: 132891885-132891885 on Build GRCh38
rs886063573 Chr.9: 132892163-132892163 on Build GRCh38
rs555515069 Chr.9: 132892239-132892239 on Build GRCh38
rs1010544692 Chr.9: 132892297-132892297 on Build GRCh38
rs563835484 Chr.9: 132892247-132892247 on Build GRCh38
rs17149898 Chr.9: 132892178-132892178 on Build GRCh38
rs143740161 Chr.9: 132892356-132892356 on Build GRCh38
rs754744080 Chr.9: 132892279-132892280 on Build GRCh38
rs915059914 Chr.9: 132891955-132891955 on Build GRCh38
rs972989516 Chr.9: 132891987-132891987 on Build GRCh38
rs1041962834 Chr.9: 132892264-132892264 on Build GRCh38
rs914224368 Chr.9: 132892244-132892244 on Build GRCh38
rs886063568 Chr.9: 132892002-132892002 on Build GRCh38
rs879566125 Chr.9: 132892228-132892228 on Build GRCh38
rs958688346 Chr.9: 132891911-132891911 on Build GRCh38
rs886063566 Chr.9: 132891879-132891879 on Build GRCh38
rs397514787 Chr.9: 132892300-132892300 on Build GRCh38
rs397514853 Chr.9: 132892299-132892299 on Build GRCh38
rs946297778 Chr.9: 132892129-132892129 on Build GRCh38
rs1025927147 Chr.9: 132892218-132892218 on Build GRCh38
rs1055854260 Chr.9: 132892298-132892298 on Build GRCh38
rs962619192 Chr.9: 132892188-132892188 on Build GRCh38
rs758417405 Chr.9: 132892236-132892236 on Build GRCh38
rs910983032 Chr.9: 132892232-132892232 on Build GRCh38
rs899216407 Chr.9: 132892233-132892233 on Build GRCh38
rs34809769 Chr.9: 132891923-132891924 on Build GRCh38
rs1042304858 Chr.9: 132892134-132892134 on Build GRCh38
rs1006184571 Chr.9: 132892174-132892174 on Build GRCh38
rs576689814 Chr.9: 132892227-132892227 on Build GRCh38
rs971000148 Chr.9: 132892088-132892088 on Build GRCh38
rs886063572 Chr.9: 132892107-132892107 on Build GRCh38
rs148982924 Chr.9: 132892238-132892238 on Build GRCh38
rs879610910 Chr.9: 132891875-132891875 on Build GRCh38
rs909804097 Chr.9: 132891960-132891960 on Build GRCh38
rs989164525 Chr.9: 132892119-132892119 on Build GRCh38
rs1007578552 Chr.9: 132892060-132892060 on Build GRCh38
rs911093700 Chr.9: 132892368-132892368 on Build GRCh38
rs565885149 Chr.9: 132892172-132892172 on Build GRCh38
rs933909739 Chr.9: 132892179-132892179 on Build GRCh38
rs550488189 Chr.9: 132892023-132892023 on Build GRCh38
rs1003401389 Chr.9: 132892162-132892162 on Build GRCh38
rs886063570 Chr.9: 132892068-132892068 on Build GRCh38
rs993591266 Chr.9: 132892080-132892080 on Build GRCh38
rs982804845 Chr.9: 132892091-132892091 on Build GRCh38
rs933906882 Chr.9: 132892009-132892009 on Build GRCh38
rs568131325 Chr.9: 132892086-132892086 on Build GRCh38
rs907401792 Chr.9: 132892266-132892266 on Build GRCh38
rs889143799 Chr.9: 132892051-132892051 on Build GRCh38
rs899266919 Chr.9: 132892078-132892078 on Build GRCh38
rs149697824 Chr.9: 132892114-132892114 on Build GRCh38
rs752925196 Chr.9: 132892226-132892226 on Build GRCh38

View Full Product Details