Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2937785-2938318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSE
Entrez Gene ID
415
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Gene Aliases
ASE, CDPX, CDPX1, CDPXR
UniGene
Hs.386975
Gene Chromosome Location
Chr.X: 2934632-2968310 on Build GRCh38
Transcripts
NM_000047.2 NM_001282628.1 NM_001282631.1 XM_005274518.2 XM_005274519.4 XM_005274521.4 XM_011545521.1 XM_017029525.1 XM_017029526.1

SNP Detail

SNP ID Chromosome Location
rs758398497 Chr.X: 2938119-2938119 on Build GRCh38
rs763508487 Chr.X: 2937900-2937900 on Build GRCh38
rs764849893 Chr.X: 2938247-2938247 on Build GRCh38
rs758104330 Chr.X: 2938166-2938166 on Build GRCh38
rs182684120 Chr.X: 2938039-2938039 on Build GRCh38
rs35143646 Chr.X: 2938114-2938114 on Build GRCh38
rs137882432 Chr.X: 2938125-2938125 on Build GRCh38
rs746787848 Chr.X: 2938272-2938272 on Build GRCh38
rs756846895 Chr.X: 2938139-2938139 on Build GRCh38
rs751548925 Chr.X: 2938132-2938132 on Build GRCh38
rs779865953 Chr.X: 2938270-2938270 on Build GRCh38
rs944277473 Chr.X: 2938118-2938118 on Build GRCh38
rs919969346 Chr.X: 2938066-2938066 on Build GRCh38
rs775534769 Chr.X: 2938074-2938074 on Build GRCh38
rs759757131 Chr.X: 2938211-2938211 on Build GRCh38
rs759543650 Chr.X: 2937993-2937993 on Build GRCh38
rs916397684 Chr.X: 2937939-2937939 on Build GRCh38
rs768735666 Chr.X: 2938108-2938108 on Build GRCh38
rs999293531 Chr.X: 2938197-2938197 on Build GRCh38
rs762658659 Chr.X: 2938040-2938040 on Build GRCh38
rs771967408 Chr.X: 2938054-2938054 on Build GRCh38
rs773359335 Chr.X: 2938111-2938111 on Build GRCh38
rs758029161 Chr.X: 2938267-2938267 on Build GRCh38
rs978121452 Chr.X: 2937868-2937868 on Build GRCh38
rs201424543 Chr.X: 2938195-2938195 on Build GRCh38
rs369695084 Chr.X: 2938157-2938157 on Build GRCh38
rs377070602 Chr.X: 2938112-2938112 on Build GRCh38
rs149169623 Chr.X: 2937825-2937825 on Build GRCh38
rs375386476 Chr.X: 2938086-2938086 on Build GRCh38
rs767667671 Chr.X: 2938213-2938213 on Build GRCh38
rs780592542 Chr.X: 2938225-2938225 on Build GRCh38
rs192265966 Chr.X: 2937877-2937877 on Build GRCh38
rs774418329 Chr.X: 2938203-2938203 on Build GRCh38
rs145964787 Chr.X: 2938220-2938220 on Build GRCh38
rs771481894 Chr.X: 2937925-2937925 on Build GRCh38
rs745573863 Chr.X: 2938150-2938150 on Build GRCh38
rs947917298 Chr.X: 2937987-2937987 on Build GRCh38
rs1053029369 Chr.X: 2937853-2937853 on Build GRCh38
rs767613131 Chr.X: 2938002-2938002 on Build GRCh38
rs749325947 Chr.X: 2938192-2938192 on Build GRCh38
rs774849746 Chr.X: 2937983-2937983 on Build GRCh38
rs754500316 Chr.X: 2938221-2938221 on Build GRCh38
rs967832525 Chr.X: 2937852-2937852 on Build GRCh38
rs142382411 Chr.X: 2938145-2938145 on Build GRCh38
rs776768053 Chr.X: 2938189-2938189 on Build GRCh38
rs372359634 Chr.X: 2938185-2938185 on Build GRCh38
rs759065417 Chr.X: 2937930-2937930 on Build GRCh38
rs35557443 Chr.X: 2937844-2937845 on Build GRCh38
rs749570001 Chr.X: 2938276-2938279 on Build GRCh38
rs776164692 Chr.X: 2937887-2937887 on Build GRCh38
rs749922782 Chr.X: 2938261-2938261 on Build GRCh38
rs778253985 Chr.X: 2938126-2938126 on Build GRCh38
rs775761812 Chr.X: 2938214-2938214 on Build GRCh38
rs779945606 Chr.X: 2938180-2938180 on Build GRCh38
rs746830733 Chr.X: 2938184-2938184 on Build GRCh38
rs201122295 Chr.X: 2938115-2938115 on Build GRCh38
rs769304724 Chr.X: 2937967-2937967 on Build GRCh38

View Full Product Details