Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138116451-138117048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1D1
Entrez Gene ID
6718
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
3o5bred, CBAS2, SRD5B1
UniGene
Hs.201667
Gene Chromosome Location
Chr.7: 138076432-138118304 on Build GRCh38
Transcripts
NM_001190906.1 NM_001190907.1 NM_005989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6875607 c.946G>A p.D316N Chr.7: 138116627-138116627 on Build GRCh38
1448349 c.944G>A p.R315H Chr.7: 138116625-138116625 on Build GRCh38
3633844 c.952C>T p.P318S Chr.7: 138116633-138116633 on Build GRCh38
3633843 c.939-1G>A p.? Chr.7: 138116619-138116619 on Build GRCh38
2859776 c.945C>T p.R315R Chr.7: 138116626-138116626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145469170 Chr.7: 138116583-138116583 on Build GRCh38
rs886062006 Chr.7: 138116960-138116960 on Build GRCh38
rs146935906 Chr.7: 138116625-138116625 on Build GRCh38
rs974162855 Chr.7: 138116865-138116865 on Build GRCh38
rs764480001 Chr.7: 138116699-138116699 on Build GRCh38
rs771893098 Chr.7: 138116668-138116668 on Build GRCh38
rs375747867 Chr.7: 138116608-138116608 on Build GRCh38
rs3735023 Chr.7: 138117024-138117024 on Build GRCh38
rs1023101741 Chr.7: 138116924-138116924 on Build GRCh38
rs577575310 Chr.7: 138116728-138116728 on Build GRCh38
rs758411386 Chr.7: 138116632-138116632 on Build GRCh38
rs911142128 Chr.7: 138116790-138116790 on Build GRCh38
rs1872930 Chr.7: 138116698-138116698 on Build GRCh38
rs1049554532 Chr.7: 138116779-138116779 on Build GRCh38
rs147259015 Chr.7: 138116714-138116714 on Build GRCh38
rs556066002 Chr.7: 138117018-138117018 on Build GRCh38
rs914648217 Chr.7: 138116727-138116727 on Build GRCh38
rs201088819 Chr.7: 138116710-138116710 on Build GRCh38
rs777849387 Chr.7: 138116641-138116641 on Build GRCh38
rs1048632390 Chr.7: 138116901-138116901 on Build GRCh38
rs943902027 Chr.7: 138116791-138116791 on Build GRCh38
rs1872929 Chr.7: 138116667-138116667 on Build GRCh38
rs574043390 Chr.7: 138116950-138116950 on Build GRCh38
rs570358233 Chr.7: 138116933-138116933 on Build GRCh38
rs772294953 Chr.7: 138116996-138116996 on Build GRCh38
rs112849023 Chr.7: 138116624-138116624 on Build GRCh38
rs1043415767 Chr.7: 138116724-138116724 on Build GRCh38
rs202168720 Chr.7: 138116645-138116645 on Build GRCh38
rs553896043 Chr.7: 138116847-138116847 on Build GRCh38
rs768526432 Chr.7: 138116577-138116577 on Build GRCh38
rs760637408 Chr.7: 138116590-138116591 on Build GRCh38
rs879096512 Chr.7: 138116557-138116557 on Build GRCh38
rs369415317 Chr.7: 138116567-138116567 on Build GRCh38
rs11763625 Chr.7: 138116732-138116732 on Build GRCh38
rs368713869 Chr.7: 138116711-138116711 on Build GRCh38
rs766113984 Chr.7: 138116679-138116679 on Build GRCh38
rs567246144 Chr.7: 138116590-138116590 on Build GRCh38
rs746014627 Chr.7: 138116859-138116859 on Build GRCh38
rs997056133 Chr.7: 138116558-138116558 on Build GRCh38
rs955672007 Chr.7: 138116793-138116793 on Build GRCh38
rs781542563 Chr.7: 138116649-138116649 on Build GRCh38
rs781573316 Chr.7: 138116712-138116712 on Build GRCh38
rs142012948 Chr.7: 138116627-138116627 on Build GRCh38
rs916033085 Chr.7: 138116888-138116888 on Build GRCh38
rs1040974355 Chr.7: 138116802-138116802 on Build GRCh38
rs774296707 Chr.7: 138116579-138116579 on Build GRCh38
rs955805152 Chr.7: 138116600-138116600 on Build GRCh38
rs759924214 Chr.7: 138116692-138116692 on Build GRCh38
rs768462545 Chr.7: 138116659-138116659 on Build GRCh38
rs369864342 Chr.7: 138116833-138116833 on Build GRCh38
rs534032657 Chr.7: 138116777-138116777 on Build GRCh38
rs773069793 Chr.7: 138116591-138116591 on Build GRCh38
rs939248912 Chr.7: 138116701-138116701 on Build GRCh38
rs1007914212 Chr.7: 138116510-138116510 on Build GRCh38
rs1019252099 Chr.7: 138116551-138116551 on Build GRCh38
rs138517404 Chr.7: 138116626-138116626 on Build GRCh38
rs779264026 Chr.7: 138116763-138116763 on Build GRCh38
rs201918644 Chr.7: 138116617-138116617 on Build GRCh38
rs776212810 Chr.7: 138116686-138116686 on Build GRCh38
rs376092126 Chr.7: 138116673-138116673 on Build GRCh38
rs1006803631 Chr.7: 138116920-138116920 on Build GRCh38
rs772774403 Chr.7: 138116669-138116669 on Build GRCh38
rs913573472 Chr.7: 138117014-138117016 on Build GRCh38
rs749118657 Chr.7: 138116654-138116654 on Build GRCh38
rs753156011 Chr.7: 138116697-138116697 on Build GRCh38
rs80147006 Chr.7: 138116720-138116720 on Build GRCh38
rs974130794 Chr.7: 138116639-138116639 on Build GRCh38
rs888180498 Chr.7: 138116905-138116905 on Build GRCh38
rs766988795 Chr.7: 138116587-138116587 on Build GRCh38
rs1004858298 Chr.7: 138116805-138116805 on Build GRCh38
rs766009281 Chr.7: 138116616-138116616 on Build GRCh38
rs546917734 Chr.7: 138116997-138116997 on Build GRCh38
rs766066468 Chr.7: 138116682-138116682 on Build GRCh38
rs886062005 Chr.7: 138116956-138116956 on Build GRCh38
rs757594806 Chr.7: 138116648-138116648 on Build GRCh38
rs376735598 Chr.7: 138116734-138116734 on Build GRCh38
rs752643782 Chr.7: 138116709-138116709 on Build GRCh38
rs373883809 Chr.7: 138116498-138116498 on Build GRCh38

View Full Product Details