Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 61501578-61502110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
UniGene
Hs.332422
Gene Chromosome Location
Chr.8: 61500556-61714640 on Build GRCh38
Transcripts
NM_001164750.1 NM_004318.3 XM_005251235.2 XM_005251236.2 XM_005251238.2 XM_005251239.2 XM_005251240.1 XM_005251242.2 XM_005251243.2 XM_005251244.1 XM_005251246.2 XM_005251247.2 XM_005251248.1 XM_005251250.2 XM_017013419.1 XM_017013420.1 XM_017013421.1 XM_017013422.1 XM_017013423.1 XM_017013424.1 XM_017013425.1 XM_017013426.1 XM_017013427.1 XM_017013428.1 XM_017013429.1 XM_017013430.1 XM_017013431.1 XM_017013432.1 XM_017013433.1 XM_017013434.1 XM_017013435.1 XM_017013436.1 XM_017013437.1 XM_017013438.1 XM_017013439.1 XM_017013440.1 XM_017013441.1 XM_017013442.1 XM_017013443.1 XM_017013444.1 NM_173519.2 XM_017013141.1 XM_017013142.1

Gene Symbol
CLVS1
Entrez Gene ID
157807
Gene Name
clavesin 1
Gene Aliases
CRALBPL, RLBP1L1
UniGene
Hs.591874
Gene Chromosome Location
Chr.8: 61057158-61501645 on Build GRCh38
Transcripts
NM_001164750.1 NM_004318.3 XM_005251235.2 XM_005251236.2 XM_005251238.2 XM_005251239.2 XM_005251240.1 XM_005251242.2 XM_005251243.2 XM_005251244.1 XM_005251246.2 XM_005251247.2 XM_005251248.1 XM_005251250.2 XM_017013419.1 XM_017013420.1 XM_017013421.1 XM_017013422.1 XM_017013423.1 XM_017013424.1 XM_017013425.1 XM_017013426.1 XM_017013427.1 XM_017013428.1 XM_017013429.1 XM_017013430.1 XM_017013431.1 XM_017013432.1 XM_017013433.1 XM_017013434.1 XM_017013435.1 XM_017013436.1 XM_017013437.1 XM_017013438.1 XM_017013439.1 XM_017013440.1 XM_017013441.1 XM_017013442.1 XM_017013443.1 XM_017013444.1 NM_173519.2 XM_017013141.1 XM_017013142.1

SNP Detail

SNP ID Chromosome Location
rs767353398 Chr.8: 61501853-61501853 on Build GRCh38
rs931144450 Chr.8: 61501829-61501829 on Build GRCh38
rs1023437004 Chr.8: 61501724-61501724 on Build GRCh38
rs945761699 Chr.8: 61501936-61501936 on Build GRCh38
rs538636340 Chr.8: 61502032-61502032 on Build GRCh38
rs72657080 Chr.8: 61502042-61502042 on Build GRCh38
rs11539105 Chr.8: 61501815-61501815 on Build GRCh38
rs763253460 Chr.8: 61502084-61502084 on Build GRCh38
rs34898205 Chr.8: 61501994-61501995 on Build GRCh38
rs116819712 Chr.8: 61501834-61501834 on Build GRCh38
rs1033858256 Chr.8: 61502047-61502047 on Build GRCh38
rs1023978169 Chr.8: 61502057-61502057 on Build GRCh38
rs985293411 Chr.8: 61501832-61501832 on Build GRCh38
rs942944160 Chr.8: 61501855-61501858 on Build GRCh38
rs994567903 Chr.8: 61501650-61501650 on Build GRCh38
rs886217915 Chr.8: 61501992-61501993 on Build GRCh38
rs781568408 Chr.8: 61501964-61501964 on Build GRCh38
rs753025743 Chr.8: 61501839-61501840 on Build GRCh38
rs577998542 Chr.8: 61502022-61502022 on Build GRCh38
rs182998141 Chr.8: 61501859-61501859 on Build GRCh38
rs886103583 Chr.8: 61501680-61501680 on Build GRCh38
rs78410879 Chr.8: 61502004-61502004 on Build GRCh38
rs896152474 Chr.8: 61502024-61502024 on Build GRCh38
rs117301546 Chr.8: 61501786-61501786 on Build GRCh38
rs931519321 Chr.8: 61501947-61501949 on Build GRCh38
rs754677049 Chr.8: 61501746-61501746 on Build GRCh38
rs1042809888 Chr.8: 61501963-61501963 on Build GRCh38
rs150177803 Chr.8: 61501847-61501852 on Build GRCh38
rs960679136 Chr.8: 61501708-61501708 on Build GRCh38
rs898678182 Chr.8: 61501606-61501606 on Build GRCh38
rs891469085 Chr.8: 61501933-61501933 on Build GRCh38
rs777208207 Chr.8: 61501657-61501657 on Build GRCh38
rs749891075 Chr.8: 61501665-61501665 on Build GRCh38
rs973213056 Chr.8: 61501814-61501814 on Build GRCh38
rs753461719 Chr.8: 61501705-61501705 on Build GRCh38
rs1056107256 Chr.8: 61502006-61502006 on Build GRCh38
rs370995098 Chr.8: 61501709-61501709 on Build GRCh38
rs1048789769 Chr.8: 61501976-61501976 on Build GRCh38
rs551487162 Chr.8: 61501783-61501783 on Build GRCh38
rs1025400417 Chr.8: 61501763-61501763 on Build GRCh38
rs190471538 Chr.8: 61501701-61501701 on Build GRCh38
rs769729745 Chr.8: 61502037-61502037 on Build GRCh38
rs538491219 Chr.8: 61501960-61501960 on Build GRCh38
rs879182685 Chr.8: 61502062-61502062 on Build GRCh38

View Full Product Details