Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243500773-243501370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2

SNP Detail

SNP ID Chromosome Location
rs1006542047 Chr.1: 243501068-243501068 on Build GRCh38
rs938622698 Chr.1: 243500803-243500803 on Build GRCh38
rs979374963 Chr.1: 243501214-243501214 on Build GRCh38
rs138661915 Chr.1: 243501244-243501244 on Build GRCh38
rs889493917 Chr.1: 243501064-243501064 on Build GRCh38
rs776940854 Chr.1: 243501093-243501094 on Build GRCh38
rs762363178 Chr.1: 243501102-243501102 on Build GRCh38
rs933500303 Chr.1: 243501254-243501254 on Build GRCh38
rs113544916 Chr.1: 243501312-243501312 on Build GRCh38
rs925527371 Chr.1: 243500897-243500897 on Build GRCh38
rs946060746 Chr.1: 243501108-243501108 on Build GRCh38
rs925558493 Chr.1: 243501235-243501235 on Build GRCh38
rs994033975 Chr.1: 243501094-243501094 on Build GRCh38
rs900263514 Chr.1: 243500908-243500909 on Build GRCh38
rs998307958 Chr.1: 243501026-243501026 on Build GRCh38
rs1038780959 Chr.1: 243500907-243500907 on Build GRCh38
rs560069415 Chr.1: 243500967-243500967 on Build GRCh38
rs532373474 Chr.1: 243501059-243501059 on Build GRCh38
rs979174570 Chr.1: 243501311-243501311 on Build GRCh38
rs917813953 Chr.1: 243500851-243500851 on Build GRCh38
rs1025859862 Chr.1: 243501101-243501101 on Build GRCh38
rs936479379 Chr.1: 243501125-243501125 on Build GRCh38
rs1050855162 Chr.1: 243501032-243501032 on Build GRCh38
rs149318674 Chr.1: 243501261-243501261 on Build GRCh38
rs183108225 Chr.1: 243501106-243501106 on Build GRCh38
rs936536092 Chr.1: 243500872-243500872 on Build GRCh38
rs562287245 Chr.1: 243501148-243501148 on Build GRCh38
rs933052466 Chr.1: 243501183-243501183 on Build GRCh38
rs1058305 Chr.1: 243501340-243501340 on Build GRCh38
rs961052137 Chr.1: 243500797-243500797 on Build GRCh38
rs1028556325 Chr.1: 243501078-243501078 on Build GRCh38
rs531160211 Chr.1: 243501153-243501153 on Build GRCh38
rs1016961195 Chr.1: 243500988-243500988 on Build GRCh38
rs529724119 Chr.1: 243501099-243501100 on Build GRCh38
rs763016492 Chr.1: 243501311-243501312 on Build GRCh38
rs992692036 Chr.1: 243501132-243501132 on Build GRCh38
rs1057165113 Chr.1: 243500850-243500852 on Build GRCh38
rs923465631 Chr.1: 243501253-243501253 on Build GRCh38
rs997110393 Chr.1: 243500910-243500910 on Build GRCh38
rs1051486828 Chr.1: 243501207-243501207 on Build GRCh38
rs960127861 Chr.1: 243501089-243501089 on Build GRCh38
rs568378973 Chr.1: 243501234-243501234 on Build GRCh38
rs925231629 Chr.1: 243501121-243501121 on Build GRCh38
rs980677697 Chr.1: 243501009-243501010 on Build GRCh38
rs750973612 Chr.1: 243501308-243501308 on Build GRCh38
rs969389953 Chr.1: 243501206-243501206 on Build GRCh38
rs562536728 Chr.1: 243500864-243500865 on Build GRCh38
rs916646835 Chr.1: 243501146-243501146 on Build GRCh38
rs935516711 Chr.1: 243500969-243500969 on Build GRCh38
rs988332662 Chr.1: 243501245-243501245 on Build GRCh38
rs575280622 Chr.1: 243500858-243500858 on Build GRCh38
rs534852128 Chr.1: 243501300-243501300 on Build GRCh38
rs775991264 Chr.1: 243500975-243500975 on Build GRCh38
rs1020416150 Chr.1: 243501306-243501306 on Build GRCh38
rs962590503 Chr.1: 243501085-243501085 on Build GRCh38
rs60418621 Chr.1: 243500862-243500862 on Build GRCh38
rs992180505 Chr.1: 243500806-243500806 on Build GRCh38
rs550016430 Chr.1: 243501283-243501283 on Build GRCh38
rs994526099 Chr.1: 243501314-243501314 on Build GRCh38
rs1053012192 Chr.1: 243500971-243500971 on Build GRCh38
rs963573860 Chr.1: 243501001-243501001 on Build GRCh38
rs1016889684 Chr.1: 243501083-243501083 on Build GRCh38
rs1029971897 Chr.1: 243500952-243500952 on Build GRCh38
rs548162696 Chr.1: 243501238-243501239 on Build GRCh38
rs531561178 Chr.1: 243501260-243501260 on Build GRCh38
rs72759895 Chr.1: 243500796-243500796 on Build GRCh38
rs549248513 Chr.1: 243500859-243500859 on Build GRCh38
rs766074349 Chr.1: 243501288-243501288 on Build GRCh38
rs547790258 Chr.1: 243501219-243501219 on Build GRCh38
rs978027490 Chr.1: 243501119-243501119 on Build GRCh38
rs1034401690 Chr.1: 243501088-243501088 on Build GRCh38
rs769095830 Chr.1: 243500793-243500793 on Build GRCh38
rs916615487 Chr.1: 243500814-243500814 on Build GRCh38

View Full Product Details