Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65777331-65777928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5B1
Entrez Gene ID
91056
Gene Name
adaptor related protein complex 5 beta 1 subunit
Gene Aliases
AP-5, PP1030
UniGene
Hs.591957
Gene Chromosome Location
Chr.11: 65773898-65780591 on Build GRCh38
Transcripts
NM_138368.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475759 c.2573_2576delCCGA p.T858fs*>20 Chr.11: 65777917-65777920 on Build GRCh38
6519248 c.2601G>T p.A867A Chr.11: 65777892-65777892 on Build GRCh38
4482321 c.2595C>T p.P865P Chr.11: 65777898-65777898 on Build GRCh38
5956801 c.2583G>C p.W861C Chr.11: 65777910-65777910 on Build GRCh38
3782664 c.2576A>G p.D859G Chr.11: 65777917-65777917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536039638 Chr.11: 65777851-65777851 on Build GRCh38
rs762416294 Chr.11: 65777468-65777484 on Build GRCh38
rs772601445 Chr.11: 65777375-65777375 on Build GRCh38
rs374297007 Chr.11: 65777361-65777361 on Build GRCh38
rs183505536 Chr.11: 65777674-65777674 on Build GRCh38
rs959428301 Chr.11: 65777474-65777474 on Build GRCh38
rs963209669 Chr.11: 65777480-65777480 on Build GRCh38
rs995819334 Chr.11: 65777442-65777443 on Build GRCh38
rs531361569 Chr.11: 65777735-65777735 on Build GRCh38
rs943146631 Chr.11: 65777385-65777385 on Build GRCh38
rs944845732 Chr.11: 65777638-65777638 on Build GRCh38
rs1043039844 Chr.11: 65777440-65777440 on Build GRCh38
rs577605653 Chr.11: 65777897-65777897 on Build GRCh38
rs943117161 Chr.11: 65777530-65777530 on Build GRCh38
rs139297906 Chr.11: 65777658-65777658 on Build GRCh38
rs370641343 Chr.11: 65777781-65777781 on Build GRCh38
rs1016100100 Chr.11: 65777479-65777479 on Build GRCh38
rs775281874 Chr.11: 65777429-65777429 on Build GRCh38
rs190099029 Chr.11: 65777868-65777868 on Build GRCh38
rs376274062 Chr.11: 65777731-65777731 on Build GRCh38
rs906758247 Chr.11: 65777657-65777657 on Build GRCh38
rs187520343 Chr.11: 65777749-65777749 on Build GRCh38
rs1042619474 Chr.11: 65777633-65777633 on Build GRCh38
rs1009935466 Chr.11: 65777719-65777719 on Build GRCh38
rs555761772 Chr.11: 65777448-65777448 on Build GRCh38
rs149598961 Chr.11: 65777767-65777767 on Build GRCh38
rs1026561656 Chr.11: 65777899-65777899 on Build GRCh38
rs559962156 Chr.11: 65777547-65777547 on Build GRCh38
rs919182767 Chr.11: 65777490-65777490 on Build GRCh38
rs144545623 Chr.11: 65777776-65777776 on Build GRCh38
rs760346334 Chr.11: 65777851-65777851 on Build GRCh38
rs895186664 Chr.11: 65777729-65777729 on Build GRCh38
rs989160357 Chr.11: 65777800-65777800 on Build GRCh38
rs987441934 Chr.11: 65777774-65777774 on Build GRCh38
rs769126554 Chr.11: 65777655-65777655 on Build GRCh38
rs529032160 Chr.11: 65777623-65777623 on Build GRCh38
rs898970374 Chr.11: 65777756-65777756 on Build GRCh38
rs375436485 Chr.11: 65777893-65777893 on Build GRCh38
rs1055527603 Chr.11: 65777728-65777728 on Build GRCh38
rs945548208 Chr.11: 65777553-65777553 on Build GRCh38
rs939595438 Chr.11: 65777850-65777850 on Build GRCh38
rs763642177 Chr.11: 65777498-65777498 on Build GRCh38
rs932991071 Chr.11: 65777358-65777358 on Build GRCh38
rs1033308320 Chr.11: 65777690-65777690 on Build GRCh38
rs373036807 Chr.11: 65777636-65777636 on Build GRCh38
rs377444504 Chr.11: 65777400-65777400 on Build GRCh38
rs930682413 Chr.11: 65777620-65777620 on Build GRCh38
rs544596955 Chr.11: 65777879-65777879 on Build GRCh38
rs370026402 Chr.11: 65777541-65777541 on Build GRCh38
rs564625613 Chr.11: 65777481-65777481 on Build GRCh38
rs906505817 Chr.11: 65777451-65777451 on Build GRCh38
rs1047835917 Chr.11: 65777394-65777394 on Build GRCh38
rs899207578 Chr.11: 65777367-65777367 on Build GRCh38
rs575626286 Chr.11: 65777461-65777461 on Build GRCh38
rs563090606 Chr.11: 65777493-65777493 on Build GRCh38
rs993756634 Chr.11: 65777765-65777765 on Build GRCh38
rs1020017524 Chr.11: 65777720-65777720 on Build GRCh38
rs1047783603 Chr.11: 65777631-65777631 on Build GRCh38
rs575995291 Chr.11: 65777632-65777632 on Build GRCh38
rs920583134 Chr.11: 65777519-65777519 on Build GRCh38
rs964351218 Chr.11: 65777784-65777784 on Build GRCh38
rs763235486 Chr.11: 65777812-65777812 on Build GRCh38
rs919865071 Chr.11: 65777814-65777816 on Build GRCh38
rs557825940 Chr.11: 65777892-65777892 on Build GRCh38
rs928188012 Chr.11: 65777840-65777840 on Build GRCh38
rs1053508773 Chr.11: 65777531-65777531 on Build GRCh38
rs996826350 Chr.11: 65777673-65777673 on Build GRCh38
rs200142638 Chr.11: 65777906-65777906 on Build GRCh38
rs200115064 Chr.11: 65777702-65777702 on Build GRCh38
rs116888179 Chr.11: 65777366-65777366 on Build GRCh38
rs575818427 Chr.11: 65777878-65777878 on Build GRCh38
rs767523038 Chr.11: 65777742-65777742 on Build GRCh38
rs7107453 Chr.11: 65777835-65777835 on Build GRCh38
rs918891775 Chr.11: 65777353-65777353 on Build GRCh38
rs567095164 Chr.11: 65777838-65777838 on Build GRCh38
rs548398375 Chr.11: 65777730-65777730 on Build GRCh38
rs1026170509 Chr.11: 65777766-65777766 on Build GRCh38
rs150832077 Chr.11: 65777402-65777402 on Build GRCh38
rs371791187 Chr.11: 65777907-65777907 on Build GRCh38
rs571865787 Chr.11: 65777514-65777514 on Build GRCh38
rs952032415 Chr.11: 65777834-65777834 on Build GRCh38
rs987525050 Chr.11: 65777503-65777503 on Build GRCh38
rs78903837 Chr.11: 65777417-65777417 on Build GRCh38
rs1003971801 Chr.11: 65777457-65777457 on Build GRCh38
rs544244726 Chr.11: 65777475-65777475 on Build GRCh38
rs1019178311 Chr.11: 65777410-65777410 on Build GRCh38
rs769016025 Chr.11: 65777605-65777605 on Build GRCh38
rs915009381 Chr.11: 65777808-65777808 on Build GRCh38

View Full Product Details