Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53307675-53308046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAAS
Entrez Gene ID
8086
Gene Name
aladin WD repeat nucleoporin
Gene Aliases
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
UniGene
Hs.369144
Gene Chromosome Location
Chr.12: 53307456-53321628 on Build GRCh38
Transcripts
NM_001173466.1 NM_015665.5 XM_011538778.1 XM_011538780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043060 c.1403C>T p.A468V Chr.12: 53307858-53307858 on Build GRCh38
4043059 c.1413T>C p.S471S Chr.12: 53307848-53307848 on Build GRCh38
5900781 c.1432C>T p.R478* Chr.12: 53307698-53307698 on Build GRCh38
5051720 c.1448C>T p.P483L Chr.12: 53307682-53307682 on Build GRCh38
1606317 c.1398delA p.A468fs*>79 Chr.12: 53307863-53307863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562815320 Chr.12: 53307716-53307716 on Build GRCh38
rs1005325746 Chr.12: 53307791-53307791 on Build GRCh38
rs748004231 Chr.12: 53307727-53307727 on Build GRCh38
rs779233030 Chr.12: 53307877-53307877 on Build GRCh38
rs759148907 Chr.12: 53307914-53307914 on Build GRCh38
rs771513237 Chr.12: 53307992-53307995 on Build GRCh38
rs201437624 Chr.12: 53307886-53307886 on Build GRCh38
rs753880528 Chr.12: 53307977-53307977 on Build GRCh38
rs777653495 Chr.12: 53308021-53308021 on Build GRCh38
rs746875610 Chr.12: 53307904-53307904 on Build GRCh38
rs370325323 Chr.12: 53307837-53307837 on Build GRCh38
rs137930169 Chr.12: 53307702-53307702 on Build GRCh38
rs377397437 Chr.12: 53307830-53307830 on Build GRCh38
rs368813962 Chr.12: 53307694-53307694 on Build GRCh38
rs755150001 Chr.12: 53307876-53307876 on Build GRCh38
rs761702188 Chr.12: 53307955-53307955 on Build GRCh38
rs886973415 Chr.12: 53307766-53307766 on Build GRCh38
rs768875274 Chr.12: 53307707-53307707 on Build GRCh38
rs368550681 Chr.12: 53307942-53307942 on Build GRCh38
rs778988213 Chr.12: 53308005-53308005 on Build GRCh38
rs910652294 Chr.12: 53307966-53307966 on Build GRCh38
rs758552525 Chr.12: 53308013-53308013 on Build GRCh38
rs770631161 Chr.12: 53307751-53307751 on Build GRCh38
rs777566590 Chr.12: 53307898-53307898 on Build GRCh38
rs999279472 Chr.12: 53307989-53307989 on Build GRCh38
rs775259180 Chr.12: 53307927-53307927 on Build GRCh38
rs750990011 Chr.12: 53307838-53307838 on Build GRCh38
rs767701527 Chr.12: 53307895-53307895 on Build GRCh38
rs771005134 Chr.12: 53307911-53307911 on Build GRCh38
rs781738941 Chr.12: 53307697-53307697 on Build GRCh38
rs759207129 Chr.12: 53307824-53307824 on Build GRCh38
rs756184724 Chr.12: 53307850-53307850 on Build GRCh38
rs188658003 Chr.12: 53307995-53307995 on Build GRCh38
rs933861044 Chr.12: 53307718-53307718 on Build GRCh38
rs776232138 Chr.12: 53307913-53307913 on Build GRCh38
rs764966443 Chr.12: 53307829-53307829 on Build GRCh38
rs943399751 Chr.12: 53307988-53307988 on Build GRCh38
rs746822504 Chr.12: 53308025-53308025 on Build GRCh38
rs370634096 Chr.12: 53307836-53307836 on Build GRCh38
rs773249176 Chr.12: 53307749-53307749 on Build GRCh38
rs201394699 Chr.12: 53307798-53307798 on Build GRCh38
rs375015186 Chr.12: 53308002-53308002 on Build GRCh38
rs370258728 Chr.12: 53307956-53307956 on Build GRCh38
rs747870235 Chr.12: 53307881-53307881 on Build GRCh38
rs372555408 Chr.12: 53307738-53307738 on Build GRCh38
rs753933300 Chr.12: 53307873-53307873 on Build GRCh38
rs762073396 Chr.12: 53307933-53307933 on Build GRCh38
rs121918548 Chr.12: 53307698-53307698 on Build GRCh38
rs543727788 Chr.12: 53307713-53307713 on Build GRCh38
rs142399878 Chr.12: 53307844-53307844 on Build GRCh38
rs766471192 Chr.12: 53308009-53308009 on Build GRCh38
rs1046970581 Chr.12: 53307730-53307730 on Build GRCh38
rs971086098 Chr.12: 53307859-53307859 on Build GRCh38
rs780045347 Chr.12: 53307869-53307869 on Build GRCh38
rs749197881 Chr.12: 53307774-53307774 on Build GRCh38
rs750794667 Chr.12: 53307947-53307947 on Build GRCh38
rs762799690 Chr.12: 53307699-53307699 on Build GRCh38
rs769301347 Chr.12: 53307925-53307925 on Build GRCh38
rs370929275 Chr.12: 53307750-53307750 on Build GRCh38
rs894202271 Chr.12: 53307817-53307818 on Build GRCh38

View Full Product Details