Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112450186-112450768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7409926 c.317A>C p.D106A Chr.12: 112450497-112450497 on Build GRCh38
13019 c.218C>T p.T73I Chr.12: 112450398-112450398 on Build GRCh38
6006458 c.208A>T p.K70* Chr.12: 112450388-112450388 on Build GRCh38
13025 c.227A>T p.E76V Chr.12: 112450407-112450407 on Build GRCh38
935274 c.165G>T p.K55N Chr.12: 112450345-112450345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936474187 Chr.12: 112450468-112450468 on Build GRCh38
rs761994563 Chr.12: 112450311-112450311 on Build GRCh38
rs746313728 Chr.12: 112450269-112450269 on Build GRCh38
rs121918461 Chr.12: 112450362-112450362 on Build GRCh38
rs764070941 Chr.12: 112450287-112450287 on Build GRCh38
rs397516804 Chr.12: 112450495-112450495 on Build GRCh38
rs762605422 Chr.12: 112450667-112450667 on Build GRCh38
rs397507506 Chr.12: 112450354-112450354 on Build GRCh38
rs777529056 Chr.12: 112450396-112450396 on Build GRCh38
rs1037065620 Chr.12: 112450606-112450606 on Build GRCh38
rs1039250039 Chr.12: 112450507-112450507 on Build GRCh38
rs397507515 Chr.12: 112450424-112450424 on Build GRCh38
rs80338836 Chr.12: 112450359-112450361 on Build GRCh38
rs121918460 Chr.12: 112450364-112450364 on Build GRCh38
rs747470140 Chr.12: 112450481-112450481 on Build GRCh38
rs1013419211 Chr.12: 112450365-112450365 on Build GRCh38
rs397507505 Chr.12: 112450352-112450352 on Build GRCh38
rs397507516 Chr.12: 112450469-112450469 on Build GRCh38
rs397507513 Chr.12: 112450397-112450397 on Build GRCh38
rs936748485 Chr.12: 112450702-112450702 on Build GRCh38
rs886043790 Chr.12: 112450355-112450355 on Build GRCh38
rs397516802 Chr.12: 112450397-112450398 on Build GRCh38
rs397507504 Chr.12: 112450346-112450346 on Build GRCh38
rs551445031 Chr.12: 112450240-112450240 on Build GRCh38
rs751437780 Chr.12: 112450353-112450353 on Build GRCh38
rs776842434 Chr.12: 112450310-112450310 on Build GRCh38
rs769864470 Chr.12: 112450552-112450552 on Build GRCh38
rs121918465 Chr.12: 112450407-112450407 on Build GRCh38
rs182335523 Chr.12: 112450241-112450241 on Build GRCh38
rs1015260760 Chr.12: 112450530-112450530 on Build GRCh38
rs397507509 Chr.12: 112450359-112450359 on Build GRCh38
rs375553813 Chr.12: 112450515-112450515 on Build GRCh38
rs747300372 Chr.12: 112450490-112450490 on Build GRCh38
rs121918464 Chr.12: 112450406-112450406 on Build GRCh38
rs397507507 Chr.12: 112450358-112450358 on Build GRCh38
rs121918453 Chr.12: 112450394-112450394 on Build GRCh38
rs891211606 Chr.12: 112450224-112450224 on Build GRCh38
rs1026762762 Chr.12: 112450492-112450492 on Build GRCh38
rs147388185 Chr.12: 112450336-112450336 on Build GRCh38
rs397516801 Chr.12: 112450389-112450389 on Build GRCh38
rs143189981 Chr.12: 112450237-112450237 on Build GRCh38
rs1050511779 Chr.12: 112450244-112450244 on Build GRCh38
rs765806729 Chr.12: 112450701-112450701 on Build GRCh38
rs1057517917 Chr.12: 112450368-112450369 on Build GRCh38
rs748952554 Chr.12: 112450417-112450417 on Build GRCh38
rs397507503 Chr.12: 112450335-112450335 on Build GRCh38
rs61736914 Chr.12: 112450435-112450435 on Build GRCh38
rs186941126 Chr.12: 112450547-112450547 on Build GRCh38
rs121918462 Chr.12: 112450398-112450398 on Build GRCh38
rs121918466 Chr.12: 112450416-112450416 on Build GRCh38
rs397507510 Chr.12: 112450361-112450361 on Build GRCh38
rs1057517935 Chr.12: 112450360-112450361 on Build GRCh38
rs115658366 Chr.12: 112450529-112450529 on Build GRCh38
rs397507511 Chr.12: 112450385-112450385 on Build GRCh38
rs121918459 Chr.12: 112450368-112450368 on Build GRCh38
rs587778636 Chr.12: 112450328-112450328 on Build GRCh38
rs111512672 Chr.12: 112450651-112450651 on Build GRCh38
rs387907158 Chr.12: 112450475-112450475 on Build GRCh38
rs397507518 Chr.12: 112450508-112450508 on Build GRCh38
rs727503380 Chr.12: 112450386-112450386 on Build GRCh38
rs397507519 Chr.12: 112450509-112450509 on Build GRCh38
rs368301721 Chr.12: 112450580-112450580 on Build GRCh38
rs925129380 Chr.12: 112450436-112450436 on Build GRCh38
rs869312744 Chr.12: 112450452-112450452 on Build GRCh38
rs765495843 Chr.12: 112450323-112450323 on Build GRCh38
rs121918454 Chr.12: 112450395-112450395 on Build GRCh38
rs986693078 Chr.12: 112450672-112450672 on Build GRCh38
rs190208557 Chr.12: 112450668-112450668 on Build GRCh38
rs748350921 Chr.12: 112450285-112450285 on Build GRCh38
rs397507512 Chr.12: 112450391-112450391 on Build GRCh38
rs1051510206 Chr.12: 112450219-112450219 on Build GRCh38
rs397516803 Chr.12: 112450415-112450415 on Build GRCh38
rs1052392524 Chr.12: 112450718-112450718 on Build GRCh38
rs16942067 Chr.12: 112450238-112450238 on Build GRCh38
rs755709231 Chr.12: 112450392-112450392 on Build GRCh38
rs995709639 Chr.12: 112450677-112450677 on Build GRCh38
rs913863806 Chr.12: 112450742-112450742 on Build GRCh38
rs772834134 Chr.12: 112450559-112450559 on Build GRCh38
rs550671096 Chr.12: 112450228-112450228 on Build GRCh38
rs397507508 Chr.12: 112450359-112450362 on Build GRCh38
rs1052382672 Chr.12: 112450347-112450347 on Build GRCh38
rs959868124 Chr.12: 112450314-112450314 on Build GRCh38
rs753788277 Chr.12: 112450426-112450426 on Build GRCh38
rs397507514 Chr.12: 112450408-112450408 on Build GRCh38
rs923454305 Chr.12: 112450661-112450661 on Build GRCh38
rs372902758 Chr.12: 112450556-112450556 on Build GRCh38
rs891118259 Chr.12: 112450560-112450560 on Build GRCh38
rs976323952 Chr.12: 112450612-112450614 on Build GRCh38
rs772929931 Chr.12: 112450723-112450724 on Build GRCh38
rs869025574 Chr.12: 112450361-112450363 on Build GRCh38
rs766205831 Chr.12: 112450339-112450339 on Build GRCh38
rs925409442 Chr.12: 112450686-112450686 on Build GRCh38
rs940147849 Chr.12: 112450586-112450586 on Build GRCh38
rs397507517 Chr.12: 112450497-112450497 on Build GRCh38

View Full Product Details