Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119894841-119895428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
UniGene
Hs.283011
Gene Chromosome Location
Chr.1: 119893533-119896524 on Build GRCh38
Transcripts
NM_021794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894728 c.1164T>C p.C388C Chr.1: 119895173-119895173 on Build GRCh38
675007 c.1274G>A p.R425Q Chr.1: 119895063-119895063 on Build GRCh38
5508262 c.1059C>G p.C353W Chr.1: 119895278-119895278 on Build GRCh38
5451128 c.1056C>T p.Y352Y Chr.1: 119895281-119895281 on Build GRCh38
5415806 c.1087A>G p.M363V Chr.1: 119895250-119895250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764956077 Chr.1: 119895159-119895159 on Build GRCh38
rs778567520 Chr.1: 119895049-119895049 on Build GRCh38
rs368574621 Chr.1: 119894923-119894923 on Build GRCh38
rs1005591450 Chr.1: 119895070-119895070 on Build GRCh38
rs781411347 Chr.1: 119895192-119895192 on Build GRCh38
rs777017829 Chr.1: 119894985-119894985 on Build GRCh38
rs983558313 Chr.1: 119894874-119894874 on Build GRCh38
rs587701025 Chr.1: 119895212-119895212 on Build GRCh38
rs867694217 Chr.1: 119894922-119894922 on Build GRCh38
rs202246434 Chr.1: 119895354-119895354 on Build GRCh38
rs769317336 Chr.1: 119895361-119895361 on Build GRCh38
rs760941629 Chr.1: 119895403-119895403 on Build GRCh38
rs774183540 Chr.1: 119894882-119894882 on Build GRCh38
rs773509669 Chr.1: 119895203-119895203 on Build GRCh38
rs775944924 Chr.1: 119895237-119895237 on Build GRCh38
rs766145128 Chr.1: 119895375-119895375 on Build GRCh38
rs756845946 Chr.1: 119895158-119895158 on Build GRCh38
rs770650583 Chr.1: 119894961-119894961 on Build GRCh38
rs145317971 Chr.1: 119895063-119895063 on Build GRCh38
rs993084456 Chr.1: 119895275-119895275 on Build GRCh38
rs937654827 Chr.1: 119894986-119894986 on Build GRCh38
rs992094249 Chr.1: 119895266-119895266 on Build GRCh38
rs945077516 Chr.1: 119895333-119895333 on Build GRCh38
rs764425856 Chr.1: 119894916-119894916 on Build GRCh38
rs772666710 Chr.1: 119895364-119895364 on Build GRCh38
rs587601976 Chr.1: 119895135-119895135 on Build GRCh38
rs751005187 Chr.1: 119895408-119895408 on Build GRCh38
rs770832799 Chr.1: 119894871-119894871 on Build GRCh38
rs267597955 Chr.1: 119894884-119894884 on Build GRCh38
rs767305753 Chr.1: 119895390-119895390 on Build GRCh38
rs368258402 Chr.1: 119895064-119895064 on Build GRCh38
rs756973261 Chr.1: 119895040-119895040 on Build GRCh38
rs776087621 Chr.1: 119895349-119895349 on Build GRCh38
rs778815352 Chr.1: 119894964-119894964 on Build GRCh38
rs768086429 Chr.1: 119895258-119895259 on Build GRCh38
rs776893426 Chr.1: 119895075-119895075 on Build GRCh38
rs766919168 Chr.1: 119895015-119895015 on Build GRCh38
rs745759637 Chr.1: 119894971-119894971 on Build GRCh38
rs776647536 Chr.1: 119894992-119894992 on Build GRCh38
rs941346359 Chr.1: 119895363-119895363 on Build GRCh38
rs759610347 Chr.1: 119894885-119894885 on Build GRCh38
rs777403590 Chr.1: 119894931-119894931 on Build GRCh38
rs587607904 Chr.1: 119895030-119895030 on Build GRCh38
rs750069068 Chr.1: 119895249-119895249 on Build GRCh38
rs762302511 Chr.1: 119894921-119894921 on Build GRCh38
rs900803268 Chr.1: 119895127-119895127 on Build GRCh38
rs183935565 Chr.1: 119894926-119894926 on Build GRCh38
rs770115548 Chr.1: 119895101-119895101 on Build GRCh38
rs764329203 Chr.1: 119895142-119895142 on Build GRCh38
rs748278094 Chr.1: 119895195-119895195 on Build GRCh38
rs781198225 Chr.1: 119895263-119895263 on Build GRCh38
rs762145751 Chr.1: 119894991-119894991 on Build GRCh38
rs917390514 Chr.1: 119895291-119895291 on Build GRCh38
rs776040586 Chr.1: 119895271-119895271 on Build GRCh38
rs752265647 Chr.1: 119894927-119894927 on Build GRCh38
rs111925671 Chr.1: 119895114-119895114 on Build GRCh38
rs201810316 Chr.1: 119895133-119895133 on Build GRCh38
rs587640612 Chr.1: 119895032-119895032 on Build GRCh38
rs147643084 Chr.1: 119894980-119894980 on Build GRCh38
rs771208017 Chr.1: 119895303-119895303 on Build GRCh38
rs772921860 Chr.1: 119895086-119895092 on Build GRCh38
rs587663718 Chr.1: 119895121-119895121 on Build GRCh38
rs766274936 Chr.1: 119895253-119895253 on Build GRCh38
rs145141310 Chr.1: 119895065-119895065 on Build GRCh38
rs774735574 Chr.1: 119895213-119895213 on Build GRCh38
rs140489804 Chr.1: 119895055-119895055 on Build GRCh38
rs763562788 Chr.1: 119895134-119895134 on Build GRCh38
rs766828026 Chr.1: 119895119-119895119 on Build GRCh38
rs41276632 Chr.1: 119895380-119895380 on Build GRCh38
rs777761187 Chr.1: 119895286-119895286 on Build GRCh38
rs936839708 Chr.1: 119895372-119895372 on Build GRCh38
rs139123456 Chr.1: 119895200-119895200 on Build GRCh38
rs900680793 Chr.1: 119895406-119895406 on Build GRCh38
rs369903502 Chr.1: 119895234-119895234 on Build GRCh38
rs752675578 Chr.1: 119895265-119895265 on Build GRCh38
rs374260098 Chr.1: 119895251-119895251 on Build GRCh38
rs762684242 Chr.1: 119895369-119895369 on Build GRCh38
rs377705019 Chr.1: 119895281-119895281 on Build GRCh38
rs751870675 Chr.1: 119895177-119895177 on Build GRCh38
rs759439950 Chr.1: 119895386-119895386 on Build GRCh38
rs750145217 Chr.1: 119895176-119895176 on Build GRCh38
rs148460316 Chr.1: 119894929-119894929 on Build GRCh38
rs2793823 Chr.1: 119895095-119895095 on Build GRCh38
rs749563971 Chr.1: 119895210-119895210 on Build GRCh38
rs866185301 Chr.1: 119894950-119894950 on Build GRCh38
rs2641348 Chr.1: 119895261-119895261 on Build GRCh38
rs749492038 Chr.1: 119895294-119895294 on Build GRCh38
rs372067600 Chr.1: 119895000-119895000 on Build GRCh38
rs769611489 Chr.1: 119895005-119895016 on Build GRCh38
rs148513488 Chr.1: 119895335-119895335 on Build GRCh38
rs144613770 Chr.1: 119894981-119894981 on Build GRCh38
rs760030228 Chr.1: 119895232-119895232 on Build GRCh38
rs754909170 Chr.1: 119895262-119895262 on Build GRCh38
rs962518414 Chr.1: 119895225-119895225 on Build GRCh38
rs759719895 Chr.1: 119895210-119895211 on Build GRCh38
rs768835755 Chr.1: 119895069-119895069 on Build GRCh38
rs749522525 Chr.1: 119895096-119895099 on Build GRCh38
rs761124638 Chr.1: 119895305-119895306 on Build GRCh38
rs746244883 Chr.1: 119895334-119895334 on Build GRCh38
rs774505840 Chr.1: 119895192-119895193 on Build GRCh38
rs754189445 Chr.1: 119894917-119894917 on Build GRCh38
rs763377661 Chr.1: 119895112-119895112 on Build GRCh38
rs771336391 Chr.1: 119895147-119895148 on Build GRCh38
rs761418593 Chr.1: 119895238-119895238 on Build GRCh38
rs199841398 Chr.1: 119895104-119895104 on Build GRCh38
rs775697948 Chr.1: 119894972-119894972 on Build GRCh38
rs1037007087 Chr.1: 119895388-119895388 on Build GRCh38
rs587718011 Chr.1: 119895014-119895014 on Build GRCh38
rs752132571 Chr.1: 119895016-119895016 on Build GRCh38
rs775753022 Chr.1: 119894896-119894896 on Build GRCh38
rs756236574 Chr.1: 119895275-119895275 on Build GRCh38
rs867116861 Chr.1: 119895116-119895116 on Build GRCh38
rs749091259 Chr.1: 119894867-119894867 on Build GRCh38
rs755701771 Chr.1: 119894930-119894930 on Build GRCh38
rs144550343 Chr.1: 119895328-119895328 on Build GRCh38
rs750875369 Chr.1: 119894924-119894924 on Build GRCh38
rs374608819 Chr.1: 119894905-119894905 on Build GRCh38
rs748316220 Chr.1: 119895081-119895081 on Build GRCh38
rs749039340 Chr.1: 119894941-119894941 on Build GRCh38
rs141186333 Chr.1: 119895186-119895186 on Build GRCh38
rs141390897 Chr.1: 119895185-119895185 on Build GRCh38
rs755650418 Chr.1: 119895024-119895024 on Build GRCh38
rs764747494 Chr.1: 119895242-119895242 on Build GRCh38

View Full Product Details