Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55150442-55150924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

SNP Detail

SNP ID Chromosome Location
rs554924423 Chr.14: 55150582-55150582 on Build GRCh38
rs144201637 Chr.14: 55150609-55150609 on Build GRCh38
rs1010704494 Chr.14: 55150698-55150698 on Build GRCh38
rs552147265 Chr.14: 55150868-55150868 on Build GRCh38
rs375143367 Chr.14: 55150763-55150763 on Build GRCh38
rs771019551 Chr.14: 55150836-55150836 on Build GRCh38
rs368350454 Chr.14: 55150822-55150822 on Build GRCh38
rs983025582 Chr.14: 55150594-55150595 on Build GRCh38
rs367742199 Chr.14: 55150797-55150797 on Build GRCh38
rs955453508 Chr.14: 55150562-55150562 on Build GRCh38
rs540018459 Chr.14: 55150636-55150636 on Build GRCh38
rs769244934 Chr.14: 55150774-55150774 on Build GRCh38
rs1052390117 Chr.14: 55150685-55150685 on Build GRCh38
rs879095340 Chr.14: 55150766-55150766 on Build GRCh38
rs148719748 Chr.14: 55150696-55150696 on Build GRCh38
rs34261542 Chr.14: 55150467-55150468 on Build GRCh38
rs769411625 Chr.14: 55150814-55150814 on Build GRCh38
rs750819174 Chr.14: 55150892-55150892 on Build GRCh38
rs974114434 Chr.14: 55150585-55150585 on Build GRCh38
rs776955946 Chr.14: 55150824-55150825 on Build GRCh38
rs559677394 Chr.14: 55150644-55150644 on Build GRCh38
rs759095441 Chr.14: 55150860-55150860 on Build GRCh38
rs911588781 Chr.14: 55150663-55150663 on Build GRCh38
rs781261250 Chr.14: 55150808-55150808 on Build GRCh38
rs758894618 Chr.14: 55150860-55150861 on Build GRCh38
rs960914064 Chr.14: 55150593-55150593 on Build GRCh38
rs536120913 Chr.14: 55150516-55150516 on Build GRCh38
rs192235318 Chr.14: 55150605-55150605 on Build GRCh38
rs774466827 Chr.14: 55150856-55150856 on Build GRCh38
rs945594042 Chr.14: 55150664-55150664 on Build GRCh38
rs796166278 Chr.14: 55150867-55150868 on Build GRCh38
rs1057020277 Chr.14: 55150637-55150637 on Build GRCh38
rs962891133 Chr.14: 55150603-55150603 on Build GRCh38
rs1011404733 Chr.14: 55150560-55150560 on Build GRCh38
rs530334156 Chr.14: 55150654-55150654 on Build GRCh38
rs765705892 Chr.14: 55150855-55150857 on Build GRCh38
rs748472516 Chr.14: 55150810-55150810 on Build GRCh38
rs34206841 Chr.14: 55150495-55150496 on Build GRCh38
rs908513963 Chr.14: 55150583-55150583 on Build GRCh38
rs78556632 Chr.14: 55150598-55150598 on Build GRCh38
rs762360287 Chr.14: 55150831-55150832 on Build GRCh38
rs975645845 Chr.14: 55150508-55150508 on Build GRCh38
rs902819751 Chr.14: 55150721-55150721 on Build GRCh38
rs766850441 Chr.14: 55150895-55150895 on Build GRCh38
rs200491148 Chr.14: 55150777-55150777 on Build GRCh38
rs373150066 Chr.14: 55150828-55150828 on Build GRCh38
rs767111449 Chr.14: 55150862-55150862 on Build GRCh38
rs765470580 Chr.14: 55150873-55150873 on Build GRCh38
rs142439379 Chr.14: 55150702-55150702 on Build GRCh38
rs749185762 Chr.14: 55150834-55150834 on Build GRCh38
rs543453032 Chr.14: 55150595-55150595 on Build GRCh38
rs35448900 Chr.14: 55150861-55150861 on Build GRCh38
rs746695224 Chr.14: 55150483-55150483 on Build GRCh38
rs768292092 Chr.14: 55150569-55150569 on Build GRCh38
rs146474046 Chr.14: 55150676-55150679 on Build GRCh38
rs79582807 Chr.14: 55150869-55150869 on Build GRCh38
rs57750925 Chr.14: 55150894-55150894 on Build GRCh38
rs531145796 Chr.14: 55150773-55150773 on Build GRCh38
rs28405981 Chr.14: 55150584-55150584 on Build GRCh38
rs963952254 Chr.14: 55150488-55150488 on Build GRCh38

View Full Product Details