Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46659721-46660184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

SNP Detail

SNP ID Chromosome Location
rs1006640513 Chr.16: 46660127-46660127 on Build GRCh38
rs774565598 Chr.16: 46659971-46659971 on Build GRCh38
rs1012263955 Chr.16: 46659789-46659789 on Build GRCh38
rs968372667 Chr.16: 46660019-46660019 on Build GRCh38
rs985863143 Chr.16: 46659959-46659959 on Build GRCh38
rs894312500 Chr.16: 46659773-46659773 on Build GRCh38
rs776500632 Chr.16: 46660089-46660089 on Build GRCh38
rs779756007 Chr.16: 46659847-46659847 on Build GRCh38
rs1032102320 Chr.16: 46659767-46659767 on Build GRCh38
rs1045140300 Chr.16: 46659803-46659803 on Build GRCh38
rs1045123990 Chr.16: 46660110-46660110 on Build GRCh38
rs887896442 Chr.16: 46660125-46660126 on Build GRCh38
rs948904221 Chr.16: 46659864-46659864 on Build GRCh38
rs897842857 Chr.16: 46660138-46660139 on Build GRCh38
rs754785560 Chr.16: 46659885-46659885 on Build GRCh38
rs1021409720 Chr.16: 46660006-46660006 on Build GRCh38
rs749705519 Chr.16: 46659828-46659828 on Build GRCh38
rs953537707 Chr.16: 46659936-46659936 on Build GRCh38
rs188743379 Chr.16: 46660018-46660018 on Build GRCh38
rs564951006 Chr.16: 46660126-46660129 on Build GRCh38
rs938925484 Chr.16: 46659956-46659956 on Build GRCh38
rs181198454 Chr.16: 46660050-46660050 on Build GRCh38
rs542974709 Chr.16: 46659889-46659889 on Build GRCh38
rs887760536 Chr.16: 46659813-46659813 on Build GRCh38
rs746602580 Chr.16: 46659855-46659855 on Build GRCh38
rs768759859 Chr.16: 46659897-46659897 on Build GRCh38
rs998146101 Chr.16: 46660154-46660154 on Build GRCh38
rs992778344 Chr.16: 46660074-46660074 on Build GRCh38
rs543740178 Chr.16: 46660072-46660072 on Build GRCh38
rs780922363 Chr.16: 46659888-46659888 on Build GRCh38
rs977527968 Chr.16: 46660030-46660030 on Build GRCh38
rs931067259 Chr.16: 46660092-46660092 on Build GRCh38
rs762727734 Chr.16: 46659860-46659860 on Build GRCh38
rs545755034 Chr.16: 46660126-46660126 on Build GRCh38
rs886052004 Chr.16: 46660092-46660092 on Build GRCh38
rs948578513 Chr.16: 46660100-46660101 on Build GRCh38
rs553790361 Chr.16: 46660071-46660071 on Build GRCh38
rs11550460 Chr.16: 46659772-46659772 on Build GRCh38
rs572048744 Chr.16: 46660118-46660118 on Build GRCh38
rs928836241 Chr.16: 46659906-46659906 on Build GRCh38

View Full Product Details