Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60445603-60446157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPBP2
Entrez Gene ID
10513
Gene Name
amyloid beta precursor protein binding protein 2
Gene Aliases
APP-BP2, HS.84084, PAT1
UniGene
Hs.84084
Gene Chromosome Location
Chr.17: 60443149-60526240 on Build GRCh38
Transcripts
NM_001282476.1 NM_006380.3 XM_011524198.2 XM_011524199.2

SNP Detail

SNP ID Chromosome Location
rs770935920 Chr.17: 60446048-60446048 on Build GRCh38
rs906367731 Chr.17: 60445954-60445954 on Build GRCh38
rs1015294757 Chr.17: 60446004-60446004 on Build GRCh38
rs1055781743 Chr.17: 60445881-60445881 on Build GRCh38
rs936040863 Chr.17: 60445867-60445867 on Build GRCh38
rs893560410 Chr.17: 60445766-60445766 on Build GRCh38
rs34935707 Chr.17: 60445833-60445833 on Build GRCh38
rs563038620 Chr.17: 60445917-60445917 on Build GRCh38
rs952763040 Chr.17: 60445964-60445964 on Build GRCh38
rs911193546 Chr.17: 60445996-60445996 on Build GRCh38
rs935128735 Chr.17: 60446041-60446041 on Build GRCh38
rs540498458 Chr.17: 60445744-60445744 on Build GRCh38
rs912078292 Chr.17: 60445741-60445741 on Build GRCh38
rs1052101569 Chr.17: 60445763-60445763 on Build GRCh38
rs1014362260 Chr.17: 60445923-60445923 on Build GRCh38
rs1035957529 Chr.17: 60445907-60445907 on Build GRCh38
rs1004685094 Chr.17: 60445985-60445985 on Build GRCh38
rs758053122 Chr.17: 60446042-60446043 on Build GRCh38
rs748490012 Chr.17: 60445648-60445648 on Build GRCh38
rs560706609 Chr.17: 60445745-60445745 on Build GRCh38
rs911606076 Chr.17: 60446081-60446081 on Build GRCh38
rs953023817 Chr.17: 60446034-60446034 on Build GRCh38
rs546495477 Chr.17: 60446133-60446133 on Build GRCh38
rs942598182 Chr.17: 60445720-60445720 on Build GRCh38
rs922533349 Chr.17: 60446025-60446025 on Build GRCh38
rs946177464 Chr.17: 60445756-60445756 on Build GRCh38
rs188151871 Chr.17: 60446107-60446107 on Build GRCh38
rs530463634 Chr.17: 60445989-60445989 on Build GRCh38
rs1028670485 Chr.17: 60446023-60446023 on Build GRCh38
rs1024372848 Chr.17: 60445947-60445947 on Build GRCh38
rs987154541 Chr.17: 60445714-60445714 on Build GRCh38
rs972782916 Chr.17: 60445694-60445694 on Build GRCh38
rs550145660 Chr.17: 60446021-60446021 on Build GRCh38
rs1002169712 Chr.17: 60445866-60445866 on Build GRCh38
rs552790192 Chr.17: 60445872-60445872 on Build GRCh38
rs777848632 Chr.17: 60445836-60445836 on Build GRCh38
rs774606860 Chr.17: 60445880-60445880 on Build GRCh38
rs904767538 Chr.17: 60445864-60445864 on Build GRCh38
rs16944363 Chr.17: 60446097-60446097 on Build GRCh38
rs1044845225 Chr.17: 60446120-60446120 on Build GRCh38
rs1040987828 Chr.17: 60445731-60445731 on Build GRCh38
rs756539123 Chr.17: 60445653-60445653 on Build GRCh38
rs904886872 Chr.17: 60445866-60445866 on Build GRCh38
rs79424927 Chr.17: 60445785-60445785 on Build GRCh38
rs749581395 Chr.17: 60446017-60446017 on Build GRCh38
rs931262700 Chr.17: 60445700-60445700 on Build GRCh38
rs984249760 Chr.17: 60445995-60445995 on Build GRCh38
rs1023714924 Chr.17: 60445839-60445839 on Build GRCh38
rs376972404 Chr.17: 60445658-60445658 on Build GRCh38
rs762120457 Chr.17: 60446014-60446014 on Build GRCh38
rs369614186 Chr.17: 60445665-60445665 on Build GRCh38
rs918682053 Chr.17: 60445695-60445695 on Build GRCh38
rs984524249 Chr.17: 60445701-60445701 on Build GRCh38

View Full Product Details