Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22414530-22415083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE1
Entrez Gene ID
64693
Gene Name
cutaneous T-cell lymphoma-associated antigen 1
Gene Aliases
CT21.1, CT21.2, CTAGE, CTAGE-1, CTAGE-2
UniGene
Hs.406709
Gene Chromosome Location
Chr.18: 22413601-22417915 on Build GRCh38
Transcripts
NM_172241.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611104 c.57A>G p.L19L Chr.18: 22414798-22414798 on Build GRCh38
6457595 c.112C>T p.Q38* Chr.18: 22414743-22414743 on Build GRCh38
5882982 c.221G>T p.R74I Chr.18: 22414634-22414634 on Build GRCh38
5013279 c.57A>C p.L19F Chr.18: 22414798-22414798 on Build GRCh38
3773371 c.45T>C p.F15F Chr.18: 22414810-22414810 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762373758 Chr.18: 22414845-22414845 on Build GRCh38
rs971803035 Chr.18: 22414901-22414901 on Build GRCh38
rs906968302 Chr.18: 22414717-22414717 on Build GRCh38
rs150287394 Chr.18: 22414879-22414879 on Build GRCh38
rs750734038 Chr.18: 22414903-22414903 on Build GRCh38
rs986032479 Chr.18: 22414936-22414936 on Build GRCh38
rs923139697 Chr.18: 22414923-22414923 on Build GRCh38
rs755908040 Chr.18: 22414956-22414956 on Build GRCh38
rs955965881 Chr.18: 22414551-22414551 on Build GRCh38
rs151156268 Chr.18: 22414723-22414723 on Build GRCh38
rs1013214578 Chr.18: 22414853-22414853 on Build GRCh38
rs908942014 Chr.18: 22414650-22414650 on Build GRCh38
rs962104071 Chr.18: 22414891-22414891 on Build GRCh38
rs555498746 Chr.18: 22414976-22414976 on Build GRCh38
rs763532215 Chr.18: 22414849-22414849 on Build GRCh38
rs750218487 Chr.18: 22414889-22414889 on Build GRCh38
rs919462369 Chr.18: 22414898-22414898 on Build GRCh38
rs1012655037 Chr.18: 22414703-22414703 on Build GRCh38
rs138031196 Chr.18: 22415000-22415000 on Build GRCh38
rs976238185 Chr.18: 22414884-22414884 on Build GRCh38
rs988094960 Chr.18: 22414552-22414552 on Build GRCh38
rs991706422 Chr.18: 22414557-22414557 on Build GRCh38
rs1002629093 Chr.18: 22414752-22414752 on Build GRCh38
rs574738175 Chr.18: 22414996-22414996 on Build GRCh38
rs903152308 Chr.18: 22414991-22414991 on Build GRCh38
rs960283101 Chr.18: 22414555-22414555 on Build GRCh38
rs896011112 Chr.18: 22415058-22415058 on Build GRCh38
rs753820822 Chr.18: 22415045-22415045 on Build GRCh38
rs376406141 Chr.18: 22414549-22414549 on Build GRCh38
rs186641739 Chr.18: 22414935-22414935 on Build GRCh38
rs770908989 Chr.18: 22414750-22414750 on Build GRCh38
rs984581031 Chr.18: 22414648-22414648 on Build GRCh38
rs563470895 Chr.18: 22415017-22415017 on Build GRCh38
rs767150096 Chr.18: 22414877-22414877 on Build GRCh38
rs778627854 Chr.18: 22414819-22414821 on Build GRCh38
rs182822855 Chr.18: 22414828-22414828 on Build GRCh38
rs947302275 Chr.18: 22414963-22414963 on Build GRCh38
rs749353418 Chr.18: 22414639-22414639 on Build GRCh38
rs889153350 Chr.18: 22414754-22414754 on Build GRCh38
rs998914084 Chr.18: 22414805-22414805 on Build GRCh38
rs201905802 Chr.18: 22414823-22414823 on Build GRCh38
rs866607736 Chr.18: 22414705-22414705 on Build GRCh38
rs568389124 Chr.18: 22414655-22414655 on Build GRCh38
rs528123714 Chr.18: 22414553-22414553 on Build GRCh38
rs763511326 Chr.18: 22414822-22414822 on Build GRCh38
rs910652846 Chr.18: 22414630-22414630 on Build GRCh38
rs547894384 Chr.18: 22414612-22414612 on Build GRCh38
rs768821344 Chr.18: 22414698-22414698 on Build GRCh38
rs762510386 Chr.18: 22414820-22414820 on Build GRCh38
rs770601836 Chr.18: 22414772-22414772 on Build GRCh38
rs748324567 Chr.18: 22414704-22414704 on Build GRCh38
rs1023205671 Chr.18: 22414879-22414880 on Build GRCh38
rs140073810 Chr.18: 22414755-22414755 on Build GRCh38
rs761448071 Chr.18: 22414880-22414880 on Build GRCh38
rs1043035797 Chr.18: 22414790-22414790 on Build GRCh38
rs1040568720 Chr.18: 22414640-22414640 on Build GRCh38
rs759850543 Chr.18: 22414589-22414589 on Build GRCh38
rs1011472117 Chr.18: 22414810-22414810 on Build GRCh38
rs371106221 Chr.18: 22414955-22414955 on Build GRCh38
rs759565248 Chr.18: 22414779-22414779 on Build GRCh38
rs968142532 Chr.18: 22414598-22414598 on Build GRCh38
rs986651937 Chr.18: 22414624-22414624 on Build GRCh38
rs986168727 Chr.18: 22414842-22414842 on Build GRCh38
rs929576259 Chr.18: 22414669-22414669 on Build GRCh38
rs775921235 Chr.18: 22414596-22414596 on Build GRCh38
rs778059356 Chr.18: 22414691-22414691 on Build GRCh38
rs1044070143 Chr.18: 22415020-22415020 on Build GRCh38
rs573137202 Chr.18: 22414900-22414900 on Build GRCh38
rs539541123 Chr.18: 22414865-22414865 on Build GRCh38
rs903287220 Chr.18: 22414794-22414794 on Build GRCh38
rs575430629 Chr.18: 22414962-22414962 on Build GRCh38
rs191777397 Chr.18: 22414701-22414701 on Build GRCh38
rs955954493 Chr.18: 22414774-22414774 on Build GRCh38
rs940423502 Chr.18: 22414657-22414657 on Build GRCh38
rs200258851 Chr.18: 22415016-22415017 on Build GRCh38
rs1044508771 Chr.18: 22414706-22414706 on Build GRCh38
rs1043455223 Chr.18: 22414965-22414965 on Build GRCh38
rs867810661 Chr.18: 22414809-22414809 on Build GRCh38

View Full Product Details