Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15369995-15370522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
UniGene
Hs.594496
Gene Chromosome Location
Chr.19: 15353521-15379815 on Build GRCh38
Transcripts
NM_005858.3 XM_011527624.2 XM_017026141.1

SNP Detail

SNP ID Chromosome Location
rs969094832 Chr.19: 15370060-15370060 on Build GRCh38
rs768655589 Chr.19: 15370302-15370302 on Build GRCh38
rs1023859948 Chr.19: 15370262-15370262 on Build GRCh38
rs999720528 Chr.19: 15370022-15370022 on Build GRCh38
rs764675155 Chr.19: 15370143-15370143 on Build GRCh38
rs555700616 Chr.19: 15370267-15370267 on Build GRCh38
rs765397928 Chr.19: 15370184-15370184 on Build GRCh38
rs750785419 Chr.19: 15370230-15370230 on Build GRCh38
rs754591935 Chr.19: 15370165-15370165 on Build GRCh38
rs573292378 Chr.19: 15370121-15370121 on Build GRCh38
rs896662450 Chr.19: 15370219-15370219 on Build GRCh38
rs34063092 Chr.19: 15370170-15370170 on Build GRCh38
rs138223241 Chr.19: 15370179-15370179 on Build GRCh38
rs897856609 Chr.19: 15370033-15370033 on Build GRCh38
rs1027758237 Chr.19: 15370046-15370046 on Build GRCh38
rs965223563 Chr.19: 15370429-15370429 on Build GRCh38
rs142190306 Chr.19: 15370168-15370168 on Build GRCh38
rs1021229207 Chr.19: 15370438-15370438 on Build GRCh38
rs951200244 Chr.19: 15370410-15370410 on Build GRCh38
rs1002386569 Chr.19: 15370065-15370065 on Build GRCh38
rs528242714 Chr.19: 15370015-15370015 on Build GRCh38
rs932022124 Chr.19: 15370154-15370154 on Build GRCh38
rs531514937 Chr.19: 15370487-15370487 on Build GRCh38
rs369648439 Chr.19: 15370110-15370110 on Build GRCh38
rs887916838 Chr.19: 15370059-15370059 on Build GRCh38
rs935294468 Chr.19: 15370465-15370465 on Build GRCh38
rs375868421 Chr.19: 15370109-15370109 on Build GRCh38
rs970025475 Chr.19: 15370337-15370337 on Build GRCh38
rs773748067 Chr.19: 15370102-15370102 on Build GRCh38
rs568309228 Chr.19: 15370031-15370031 on Build GRCh38
rs372946120 Chr.19: 15370220-15370220 on Build GRCh38
rs747019407 Chr.19: 15370175-15370175 on Build GRCh38
rs144668817 Chr.19: 15370139-15370139 on Build GRCh38
rs576420944 Chr.19: 15370488-15370488 on Build GRCh38
rs766206094 Chr.19: 15370120-15370120 on Build GRCh38
rs887872485 Chr.19: 15370364-15370364 on Build GRCh38
rs551576876 Chr.19: 15370491-15370491 on Build GRCh38
rs780444718 Chr.19: 15370153-15370153 on Build GRCh38
rs756344624 Chr.19: 15370095-15370095 on Build GRCh38
rs541425327 Chr.19: 15370035-15370035 on Build GRCh38
rs748913936 Chr.19: 15370164-15370164 on Build GRCh38
rs921217179 Chr.19: 15370431-15370431 on Build GRCh38
rs929338048 Chr.19: 15370350-15370350 on Build GRCh38
rs897807991 Chr.19: 15370331-15370331 on Build GRCh38
rs748053285 Chr.19: 15370108-15370108 on Build GRCh38
rs764335914 Chr.19: 15370205-15370205 on Build GRCh38
rs762909315 Chr.19: 15370204-15370204 on Build GRCh38
rs943815455 Chr.19: 15370123-15370123 on Build GRCh38
rs760443333 Chr.19: 15370119-15370119 on Build GRCh38
rs758590764 Chr.19: 15370217-15370218 on Build GRCh38
rs752784479 Chr.19: 15370197-15370203 on Build GRCh38
rs373189435 Chr.19: 15370134-15370134 on Build GRCh38
rs770651728 Chr.19: 15370176-15370176 on Build GRCh38
rs781728362 Chr.19: 15370156-15370156 on Build GRCh38
rs562708256 Chr.19: 15370471-15370471 on Build GRCh38
rs575505641 Chr.19: 15370376-15370376 on Build GRCh38
rs201525366 Chr.19: 15370133-15370133 on Build GRCh38
rs367961984 Chr.19: 15370044-15370044 on Build GRCh38
rs975223145 Chr.19: 15370430-15370430 on Build GRCh38
rs1032463093 Chr.19: 15370457-15370457 on Build GRCh38
rs1049581188 Chr.19: 15370359-15370359 on Build GRCh38
rs1036931669 Chr.19: 15370290-15370290 on Build GRCh38
rs932437658 Chr.19: 15370191-15370191 on Build GRCh38
rs966744017 Chr.19: 15370439-15370439 on Build GRCh38
rs370392213 Chr.19: 15370193-15370193 on Build GRCh38
rs921155994 Chr.19: 15370150-15370150 on Build GRCh38
rs974124565 Chr.19: 15370098-15370098 on Build GRCh38
rs566857422 Chr.19: 15370214-15370214 on Build GRCh38
rs953810209 Chr.19: 15370479-15370479 on Build GRCh38
rs947281135 Chr.19: 15370436-15370436 on Build GRCh38
rs919711442 Chr.19: 15370099-15370099 on Build GRCh38
rs1033840415 Chr.19: 15370412-15370412 on Build GRCh38
rs183201344 Chr.19: 15370419-15370419 on Build GRCh38
rs1000539826 Chr.19: 15370443-15370443 on Build GRCh38
rs752151622 Chr.19: 15370147-15370147 on Build GRCh38
rs188090160 Chr.19: 15370425-15370425 on Build GRCh38
rs749344859 Chr.19: 15370161-15370161 on Build GRCh38
rs776411886 Chr.19: 15370177-15370177 on Build GRCh38
rs752748972 Chr.19: 15370140-15370140 on Build GRCh38
rs752098721 Chr.19: 15370213-15370213 on Build GRCh38
rs943943693 Chr.19: 15370243-15370243 on Build GRCh38
rs778447463 Chr.19: 15370167-15370167 on Build GRCh38
rs746804221 Chr.19: 15370067-15370067 on Build GRCh38
rs550289387 Chr.19: 15370244-15370244 on Build GRCh38
rs548090543 Chr.19: 15370020-15370020 on Build GRCh38
rs765325976 Chr.19: 15370184-15370184 on Build GRCh38
rs1027057851 Chr.19: 15370366-15370366 on Build GRCh38

View Full Product Details