Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15355027-15355512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
UniGene
Hs.594496
Gene Chromosome Location
Chr.19: 15353521-15379815 on Build GRCh38
Transcripts
NM_005858.3 XM_011527624.2 XM_017026141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709701 c.1864G>A p.E622K Chr.19: 15355130-15355130 on Build GRCh38
5626737 c.1858C>G p.Q620E Chr.19: 15355136-15355136 on Build GRCh38
5587153 c.1785C>T p.P595P Chr.19: 15355209-15355209 on Build GRCh38
709702 c.1942G>A p.E648K Chr.19: 15355052-15355052 on Build GRCh38
355656 c.1801G>A p.G601R Chr.19: 15355193-15355193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755822440 Chr.19: 15355289-15355289 on Build GRCh38
rs747928960 Chr.19: 15355248-15355248 on Build GRCh38
rs909104923 Chr.19: 15355262-15355262 on Build GRCh38
rs148784064 Chr.19: 15355096-15355096 on Build GRCh38
rs148913978 Chr.19: 15355266-15355266 on Build GRCh38
rs777457097 Chr.19: 15355079-15355079 on Build GRCh38
rs529002085 Chr.19: 15355425-15355425 on Build GRCh38
rs762037871 Chr.19: 15355194-15355194 on Build GRCh38
rs774075327 Chr.19: 15355193-15355193 on Build GRCh38
rs200241726 Chr.19: 15355252-15355252 on Build GRCh38
rs751777249 Chr.19: 15355142-15355142 on Build GRCh38
rs140661207 Chr.19: 15355155-15355155 on Build GRCh38
rs146444610 Chr.19: 15355174-15355174 on Build GRCh38
rs377642478 Chr.19: 15355410-15355410 on Build GRCh38
rs868558866 Chr.19: 15355220-15355220 on Build GRCh38
rs950117605 Chr.19: 15355331-15355331 on Build GRCh38
rs140732196 Chr.19: 15355185-15355185 on Build GRCh38
rs755046164 Chr.19: 15355395-15355398 on Build GRCh38
rs756048570 Chr.19: 15355125-15355125 on Build GRCh38
rs368685776 Chr.19: 15355338-15355338 on Build GRCh38
rs551451888 Chr.19: 15355095-15355095 on Build GRCh38
rs61757661 Chr.19: 15355208-15355208 on Build GRCh38
rs764484665 Chr.19: 15355286-15355286 on Build GRCh38
rs750181254 Chr.19: 15355104-15355104 on Build GRCh38
rs370557589 Chr.19: 15355398-15355398 on Build GRCh38
rs571638097 Chr.19: 15355130-15355130 on Build GRCh38
rs773549065 Chr.19: 15355255-15355255 on Build GRCh38
rs758666379 Chr.19: 15355272-15355272 on Build GRCh38
rs968789930 Chr.19: 15355433-15355433 on Build GRCh38
rs756893665 Chr.19: 15355228-15355228 on Build GRCh38
rs753296829 Chr.19: 15355324-15355324 on Build GRCh38
rs777574889 Chr.19: 15355168-15355168 on Build GRCh38
rs1017698390 Chr.19: 15355198-15355198 on Build GRCh38
rs141434767 Chr.19: 15355392-15355392 on Build GRCh38
rs375669146 Chr.19: 15355186-15355186 on Build GRCh38
rs775222031 Chr.19: 15355127-15355127 on Build GRCh38
rs376050255 Chr.19: 15355169-15355169 on Build GRCh38
rs199554386 Chr.19: 15355373-15355373 on Build GRCh38
rs771278987 Chr.19: 15355368-15355368 on Build GRCh38
rs766284070 Chr.19: 15355291-15355294 on Build GRCh38
rs774133689 Chr.19: 15355246-15355246 on Build GRCh38
rs576285040 Chr.19: 15355223-15355223 on Build GRCh38
rs1044518760 Chr.19: 15355105-15355105 on Build GRCh38
rs921338802 Chr.19: 15355211-15355211 on Build GRCh38
rs147487490 Chr.19: 15355362-15355362 on Build GRCh38
rs765693748 Chr.19: 15355270-15355270 on Build GRCh38
rs375883591 Chr.19: 15355156-15355156 on Build GRCh38
rs143611999 Chr.19: 15355356-15355356 on Build GRCh38
rs778741136 Chr.19: 15355335-15355335 on Build GRCh38
rs771586432 Chr.19: 15355051-15355051 on Build GRCh38
rs765983271 Chr.19: 15355397-15355397 on Build GRCh38
rs778528798 Chr.19: 15355402-15355402 on Build GRCh38
rs757266360 Chr.19: 15355391-15355391 on Build GRCh38
rs755492152 Chr.19: 15355212-15355212 on Build GRCh38
rs753977154 Chr.19: 15355306-15355308 on Build GRCh38
rs950466518 Chr.19: 15355480-15355480 on Build GRCh38
rs1045858384 Chr.19: 15355150-15355150 on Build GRCh38
rs978784742 Chr.19: 15355462-15355462 on Build GRCh38
rs375385159 Chr.19: 15355237-15355237 on Build GRCh38
rs142407870 Chr.19: 15355104-15355104 on Build GRCh38
rs758421010 Chr.19: 15355417-15355417 on Build GRCh38
rs769964243 Chr.19: 15355389-15355389 on Build GRCh38
rs200385214 Chr.19: 15355148-15355148 on Build GRCh38
rs201462022 Chr.19: 15355287-15355287 on Build GRCh38
rs950616458 Chr.19: 15355166-15355166 on Build GRCh38
rs372519515 Chr.19: 15355383-15355383 on Build GRCh38
rs745567000 Chr.19: 15355230-15355230 on Build GRCh38
rs760863073 Chr.19: 15355203-15355203 on Build GRCh38
rs536442358 Chr.19: 15355173-15355173 on Build GRCh38
rs748168148 Chr.19: 15355336-15355336 on Build GRCh38
rs138139780 Chr.19: 15355325-15355325 on Build GRCh38
rs978900886 Chr.19: 15355231-15355231 on Build GRCh38
rs111237991 Chr.19: 15355429-15355429 on Build GRCh38
rs749235652 Chr.19: 15355238-15355238 on Build GRCh38
rs1026051602 Chr.19: 15355455-15355455 on Build GRCh38
rs779482413 Chr.19: 15355308-15355308 on Build GRCh38
rs144064099 Chr.19: 15355165-15355165 on Build GRCh38
rs755591473 Chr.19: 15355232-15355232 on Build GRCh38
rs763327144 Chr.19: 15355077-15355077 on Build GRCh38
rs760253297 Chr.19: 15355395-15355395 on Build GRCh38
rs915900145 Chr.19: 15355280-15355280 on Build GRCh38
rs370821966 Chr.19: 15355418-15355418 on Build GRCh38
rs116901926 Chr.19: 15355149-15355149 on Build GRCh38
rs150076099 Chr.19: 15355229-15355229 on Build GRCh38
rs374086322 Chr.19: 15355175-15355175 on Build GRCh38
rs767857353 Chr.19: 15355195-15355195 on Build GRCh38
rs755337627 Chr.19: 15355163-15355163 on Build GRCh38
rs768497457 Chr.19: 15355190-15355190 on Build GRCh38
rs533968274 Chr.19: 15355137-15355137 on Build GRCh38
rs975549921 Chr.19: 15355206-15355206 on Build GRCh38
rs762011903 Chr.19: 15355143-15355143 on Build GRCh38
rs376225309 Chr.19: 15355390-15355390 on Build GRCh38
rs745410158 Chr.19: 15355176-15355176 on Build GRCh38
rs763846151 Chr.19: 15355131-15355131 on Build GRCh38
rs538149553 Chr.19: 15355080-15355080 on Build GRCh38
rs769723956 Chr.19: 15355107-15355107 on Build GRCh38
rs754389775 Chr.19: 15355209-15355209 on Build GRCh38
rs773387673 Chr.19: 15355199-15355199 on Build GRCh38
rs997222030 Chr.19: 15355431-15355431 on Build GRCh38
rs764648704 Chr.19: 15355375-15355375 on Build GRCh38
rs200362293 Chr.19: 15355269-15355269 on Build GRCh38
rs770469145 Chr.19: 15355090-15355090 on Build GRCh38
rs765576072 Chr.19: 15355213-15355213 on Build GRCh38
rs767626174 Chr.19: 15355147-15355147 on Build GRCh38
rs138652097 Chr.19: 15355265-15355265 on Build GRCh38
rs147009364 Chr.19: 15355294-15355294 on Build GRCh38
rs370347087 Chr.19: 15355188-15355188 on Build GRCh38

View Full Product Details