Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41475278-41475839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
UniGene
Hs.302019
Gene Chromosome Location
Chr.X: 41447460-41476414 on Build GRCh38
Transcripts
NM_022567.2 XM_017029709.1

SNP Detail

SNP ID Chromosome Location
rs921009553 Chr.X: 41475303-41475303 on Build GRCh38
rs1035997383 Chr.X: 41475582-41475582 on Build GRCh38
rs781292304 Chr.X: 41475560-41475560 on Build GRCh38
rs775322672 Chr.X: 41475771-41475771 on Build GRCh38
rs1057515885 Chr.X: 41475634-41475635 on Build GRCh38
rs1046198597 Chr.X: 41475542-41475542 on Build GRCh38
rs145454086 Chr.X: 41475673-41475673 on Build GRCh38
rs3021319 Chr.X: 41475408-41475408 on Build GRCh38
rs777385685 Chr.X: 41475491-41475491 on Build GRCh38
rs901380876 Chr.X: 41475721-41475721 on Build GRCh38
rs999570482 Chr.X: 41475735-41475735 on Build GRCh38
rs1019733370 Chr.X: 41475335-41475335 on Build GRCh38
rs182092478 Chr.X: 41475697-41475697 on Build GRCh38
rs1006981323 Chr.X: 41475668-41475669 on Build GRCh38
rs989389536 Chr.X: 41475446-41475446 on Build GRCh38
rs749061819 Chr.X: 41475811-41475811 on Build GRCh38
rs201620180 Chr.X: 41475622-41475622 on Build GRCh38
rs191505759 Chr.X: 41475519-41475519 on Build GRCh38
rs978120347 Chr.X: 41475354-41475354 on Build GRCh38
rs780639009 Chr.X: 41475643-41475643 on Build GRCh38
rs1023577653 Chr.X: 41475774-41475774 on Build GRCh38
rs947920765 Chr.X: 41475537-41475537 on Build GRCh38
rs935165720 Chr.X: 41475306-41475306 on Build GRCh38
rs747097738 Chr.X: 41475416-41475416 on Build GRCh38
rs927758898 Chr.X: 41475567-41475567 on Build GRCh38
rs965450101 Chr.X: 41475338-41475338 on Build GRCh38
rs958198171 Chr.X: 41475442-41475442 on Build GRCh38
rs1043078639 Chr.X: 41475659-41475659 on Build GRCh38
rs1052338157 Chr.X: 41475365-41475365 on Build GRCh38
rs898806102 Chr.X: 41475606-41475606 on Build GRCh38
rs1019670909 Chr.X: 41475680-41475680 on Build GRCh38
rs58930096 Chr.X: 41475313-41475313 on Build GRCh38
rs147706112 Chr.X: 41475769-41475769 on Build GRCh38
rs1048667828 Chr.X: 41475627-41475627 on Build GRCh38
rs1019785721 Chr.X: 41475663-41475663 on Build GRCh38
rs893711795 Chr.X: 41475374-41475374 on Build GRCh38
rs769905108 Chr.X: 41475770-41475770 on Build GRCh38
rs748708637 Chr.X: 41475622-41475622 on Build GRCh38
rs781670309 Chr.X: 41475739-41475739 on Build GRCh38
rs927806332 Chr.X: 41475297-41475297 on Build GRCh38
rs889753775 Chr.X: 41475661-41475661 on Build GRCh38
rs879163416 Chr.X: 41475756-41475756 on Build GRCh38
rs769865394 Chr.X: 41475770-41475771 on Build GRCh38
rs940438059 Chr.X: 41475569-41475569 on Build GRCh38
rs1007101717 Chr.X: 41475652-41475652 on Build GRCh38
rs141612904 Chr.X: 41475635-41475635 on Build GRCh38
rs1025929587 Chr.X: 41475765-41475765 on Build GRCh38
rs1031438339 Chr.X: 41475741-41475741 on Build GRCh38

View Full Product Details