Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24485122-24485618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593717 c.901C>A p.Q301K Chr.8: 24485302-24485302 on Build GRCh38
750027 c.960G>C p.K320N Chr.8: 24485361-24485361 on Build GRCh38
142119 c.905G>A p.G302E Chr.8: 24485306-24485306 on Build GRCh38
6271009 c.898G>T p.V300L Chr.8: 24485299-24485299 on Build GRCh38
6582820 c.911C>T p.S304F Chr.8: 24485312-24485312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768933719 Chr.8: 24485307-24485307 on Build GRCh38
rs551119685 Chr.8: 24485430-24485430 on Build GRCh38
rs993101593 Chr.8: 24485572-24485572 on Build GRCh38
rs757361568 Chr.8: 24485234-24485234 on Build GRCh38
rs1005795887 Chr.8: 24485167-24485167 on Build GRCh38
rs926157073 Chr.8: 24485222-24485222 on Build GRCh38
rs138307487 Chr.8: 24485539-24485539 on Build GRCh38
rs536486452 Chr.8: 24485493-24485493 on Build GRCh38
rs750946570 Chr.8: 24485319-24485319 on Build GRCh38
rs774610129 Chr.8: 24485254-24485254 on Build GRCh38
rs934663316 Chr.8: 24485429-24485429 on Build GRCh38
rs759379831 Chr.8: 24485297-24485297 on Build GRCh38
rs943314322 Chr.8: 24485354-24485354 on Build GRCh38
rs779172102 Chr.8: 24485405-24485405 on Build GRCh38
rs906031283 Chr.8: 24485507-24485507 on Build GRCh38
rs764432252 Chr.8: 24485324-24485324 on Build GRCh38
rs771540419 Chr.8: 24485187-24485187 on Build GRCh38
rs910476256 Chr.8: 24485345-24485345 on Build GRCh38
rs754168177 Chr.8: 24485364-24485364 on Build GRCh38
rs371098153 Chr.8: 24485249-24485249 on Build GRCh38
rs765224250 Chr.8: 24485348-24485348 on Build GRCh38
rs747879325 Chr.8: 24485262-24485262 on Build GRCh38
rs777410073 Chr.8: 24485386-24485386 on Build GRCh38
rs761215307 Chr.8: 24485342-24485343 on Build GRCh38
rs778266701 Chr.8: 24485410-24485410 on Build GRCh38
rs1027963653 Chr.8: 24485486-24485486 on Build GRCh38
rs950128825 Chr.8: 24485577-24485577 on Build GRCh38
rs78175273 Chr.8: 24485461-24485461 on Build GRCh38
rs375234010 Chr.8: 24485272-24485272 on Build GRCh38
rs116249853 Chr.8: 24485409-24485409 on Build GRCh38
rs370216086 Chr.8: 24485390-24485390 on Build GRCh38
rs766685748 Chr.8: 24485351-24485351 on Build GRCh38
rs761402023 Chr.8: 24485257-24485257 on Build GRCh38
rs926159823 Chr.8: 24485526-24485526 on Build GRCh38
rs555691404 Chr.8: 24485216-24485216 on Build GRCh38
rs954426900 Chr.8: 24485584-24485584 on Build GRCh38
rs549240870 Chr.8: 24485256-24485256 on Build GRCh38
rs755281210 Chr.8: 24485385-24485385 on Build GRCh38
rs758805523 Chr.8: 24485367-24485367 on Build GRCh38
rs148851846 Chr.8: 24485299-24485299 on Build GRCh38
rs370395528 Chr.8: 24485317-24485317 on Build GRCh38
rs918978735 Chr.8: 24485286-24485286 on Build GRCh38
rs1053204235 Chr.8: 24485437-24485437 on Build GRCh38
rs576787102 Chr.8: 24485588-24485588 on Build GRCh38
rs963060807 Chr.8: 24485201-24485201 on Build GRCh38
rs558226401 Chr.8: 24485583-24485583 on Build GRCh38
rs934792349 Chr.8: 24485226-24485226 on Build GRCh38
rs917413653 Chr.8: 24485552-24485552 on Build GRCh38
rs898840002 Chr.8: 24485540-24485540 on Build GRCh38
rs534097804 Chr.8: 24485562-24485562 on Build GRCh38
rs377593221 Chr.8: 24485329-24485329 on Build GRCh38
rs34584974 Chr.8: 24485217-24485218 on Build GRCh38
rs757414622 Chr.8: 24485347-24485347 on Build GRCh38
rs531031077 Chr.8: 24485162-24485162 on Build GRCh38
rs746292424 Chr.8: 24485246-24485246 on Build GRCh38
rs375246641 Chr.8: 24485382-24485382 on Build GRCh38
rs779485811 Chr.8: 24485242-24485242 on Build GRCh38
rs920916567 Chr.8: 24485159-24485159 on Build GRCh38
rs760505732 Chr.8: 24485323-24485323 on Build GRCh38
rs750992978 Chr.8: 24485352-24485352 on Build GRCh38
rs554894875 Chr.8: 24485500-24485500 on Build GRCh38
rs978992190 Chr.8: 24485513-24485513 on Build GRCh38
rs971817188 Chr.8: 24485280-24485280 on Build GRCh38
rs561278538 Chr.8: 24485265-24485265 on Build GRCh38
rs748812532 Chr.8: 24485274-24485274 on Build GRCh38
rs774415510 Chr.8: 24485292-24485292 on Build GRCh38
rs770363794 Chr.8: 24485290-24485290 on Build GRCh38
rs759683231 Chr.8: 24485456-24485456 on Build GRCh38
rs895878500 Chr.8: 24485453-24485453 on Build GRCh38
rs780503187 Chr.8: 24485380-24485380 on Build GRCh38
rs1006243092 Chr.8: 24485528-24485528 on Build GRCh38
rs932928636 Chr.8: 24485180-24485180 on Build GRCh38
rs780265327 Chr.8: 24485253-24485253 on Build GRCh38
rs571627810 Chr.8: 24485381-24485381 on Build GRCh38
rs112014682 Chr.8: 24485259-24485259 on Build GRCh38
rs1015418651 Chr.8: 24485388-24485390 on Build GRCh38
rs927284567 Chr.8: 24485587-24485587 on Build GRCh38
rs748937166 Chr.8: 24485402-24485402 on Build GRCh38
rs768180264 Chr.8: 24485318-24485319 on Build GRCh38

View Full Product Details