Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32408339-32408880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
753667 c.277G>A p.A93T Chr.9: 32408524-32408524 on Build GRCh38
753668 c.277G>A p.A93T Chr.9: 32408524-32408524 on Build GRCh38
5063881 c.301C>T p.R101C Chr.9: 32408548-32408548 on Build GRCh38
6260359 c.280G>T p.V94L Chr.9: 32408527-32408527 on Build GRCh38
7152719 c.286G>C p.D96H Chr.9: 32408533-32408533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376285476 Chr.9: 32408666-32408666 on Build GRCh38
rs372286488 Chr.9: 32408523-32408523 on Build GRCh38
rs570128113 Chr.9: 32408794-32408794 on Build GRCh38
rs772570000 Chr.9: 32408587-32408587 on Build GRCh38
rs183565316 Chr.9: 32408758-32408758 on Build GRCh38
rs765104900 Chr.9: 32408432-32408432 on Build GRCh38
rs988647864 Chr.9: 32408708-32408708 on Build GRCh38
rs761289690 Chr.9: 32408680-32408680 on Build GRCh38
rs572632758 Chr.9: 32408376-32408376 on Build GRCh38
rs773570766 Chr.9: 32408592-32408592 on Build GRCh38
rs770567107 Chr.9: 32408535-32408535 on Build GRCh38
rs527589976 Chr.9: 32408683-32408683 on Build GRCh38
rs935167801 Chr.9: 32408667-32408667 on Build GRCh38
rs913317898 Chr.9: 32408772-32408772 on Build GRCh38
rs550022329 Chr.9: 32408773-32408773 on Build GRCh38
rs41304757 Chr.9: 32408614-32408614 on Build GRCh38
rs1012006480 Chr.9: 32408375-32408375 on Build GRCh38
rs753979935 Chr.9: 32408611-32408611 on Build GRCh38
rs759263331 Chr.9: 32408498-32408500 on Build GRCh38
rs780739416 Chr.9: 32408544-32408544 on Build GRCh38
rs766587886 Chr.9: 32408603-32408603 on Build GRCh38
rs765069096 Chr.9: 32408647-32408649 on Build GRCh38
rs1001783226 Chr.9: 32408439-32408439 on Build GRCh38
rs367875724 Chr.9: 32408662-32408662 on Build GRCh38
rs891853460 Chr.9: 32408374-32408374 on Build GRCh38
rs910843737 Chr.9: 32408416-32408416 on Build GRCh38
rs758468204 Chr.9: 32408524-32408524 on Build GRCh38
rs943015511 Chr.9: 32408438-32408438 on Build GRCh38
rs202186814 Chr.9: 32408634-32408634 on Build GRCh38
rs752647033 Chr.9: 32408680-32408680 on Build GRCh38
rs775035570 Chr.9: 32408554-32408554 on Build GRCh38
rs1037397382 Chr.9: 32408443-32408443 on Build GRCh38
rs779470196 Chr.9: 32408676-32408676 on Build GRCh38
rs963698506 Chr.9: 32408626-32408626 on Build GRCh38
rs779177499 Chr.9: 32408520-32408520 on Build GRCh38
rs906112331 Chr.9: 32408429-32408429 on Build GRCh38
rs758345130 Chr.9: 32408646-32408646 on Build GRCh38
rs745573950 Chr.9: 32408546-32408546 on Build GRCh38
rs371431165 Chr.9: 32408682-32408682 on Build GRCh38
rs746785385 Chr.9: 32408533-32408533 on Build GRCh38
rs769267795 Chr.9: 32408548-32408548 on Build GRCh38
rs536277060 Chr.9: 32408593-32408593 on Build GRCh38
rs955039228 Chr.9: 32408793-32408793 on Build GRCh38
rs1021081102 Chr.9: 32408718-32408718 on Build GRCh38
rs547756031 Chr.9: 32408717-32408717 on Build GRCh38
rs976692554 Chr.9: 32408648-32408648 on Build GRCh38
rs759834876 Chr.9: 32408702-32408702 on Build GRCh38
rs146885249 Chr.9: 32408814-32408814 on Build GRCh38
rs769498731 Chr.9: 32408674-32408674 on Build GRCh38
rs142930707 Chr.9: 32408531-32408531 on Build GRCh38
rs766526154 Chr.9: 32408489-32408489 on Build GRCh38
rs10123372 Chr.9: 32408498-32408498 on Build GRCh38
rs536710907 Chr.9: 32408394-32408395 on Build GRCh38
rs947456726 Chr.9: 32408812-32408812 on Build GRCh38
rs190946964 Chr.9: 32408723-32408723 on Build GRCh38
rs879860721 Chr.9: 32408689-32408689 on Build GRCh38
rs895641100 Chr.9: 32408570-32408570 on Build GRCh38
rs755048487 Chr.9: 32408503-32408503 on Build GRCh38
rs760881923 Chr.9: 32408485-32408485 on Build GRCh38
rs774807099 Chr.9: 32408467-32408467 on Build GRCh38
rs905898594 Chr.9: 32408829-32408829 on Build GRCh38
rs773625616 Chr.9: 32408679-32408679 on Build GRCh38
rs561752787 Chr.9: 32408645-32408645 on Build GRCh38
rs745490001 Chr.9: 32408673-32408673 on Build GRCh38
rs866422191 Chr.9: 32408549-32408550 on Build GRCh38
rs186585841 Chr.9: 32408479-32408479 on Build GRCh38
rs748807960 Chr.9: 32408677-32408677 on Build GRCh38
rs987679514 Chr.9: 32408795-32408795 on Build GRCh38
rs541737026 Chr.9: 32408408-32408408 on Build GRCh38
rs368594616 Chr.9: 32408513-32408513 on Build GRCh38
rs368917410 Chr.9: 32408669-32408669 on Build GRCh38
rs762301470 Chr.9: 32408575-32408575 on Build GRCh38
rs762352779 Chr.9: 32408475-32408475 on Build GRCh38
rs984195413 Chr.9: 32408557-32408557 on Build GRCh38
rs964989268 Chr.9: 32408844-32408844 on Build GRCh38
rs1052244452 Chr.9: 32408707-32408707 on Build GRCh38

View Full Product Details